Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99307357-99307855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH1B
Entrez Gene ID
125
Gene Name
alcohol dehydrogenase 1B (class I), beta polypeptide
Gene Aliases
ADH2, HEL-S-117
UniGene
Hs.4
Gene Chromosome Location
Chr.4: 99306387-99321442 on Build GRCh38
Transcripts
NM_000668.5 NM_001286650.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
394790 c.1127G>C p.*376S Chr.4: 99307841-99307841 on Build GRCh38
4150394 c.1122G>A p.T374T Chr.4: 99307846-99307846 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs900369458 Chr.4: 99307524-99307524 on Build GRCh38
rs1004553830 Chr.4: 99307592-99307592 on Build GRCh38
rs927011467 Chr.4: 99307658-99307658 on Build GRCh38
rs374909016 Chr.4: 99307793-99307793 on Build GRCh38
rs1008772663 Chr.4: 99307686-99307686 on Build GRCh38
rs955158919 Chr.4: 99307655-99307655 on Build GRCh38
rs918023649 Chr.4: 99307546-99307546 on Build GRCh38
rs528222703 Chr.4: 99307724-99307724 on Build GRCh38
rs964212715 Chr.4: 99307703-99307703 on Build GRCh38
rs964265014 Chr.4: 99307411-99307411 on Build GRCh38
rs763273083 Chr.4: 99307822-99307822 on Build GRCh38
rs905740481 Chr.4: 99307572-99307572 on Build GRCh38
rs987000359 Chr.4: 99307402-99307402 on Build GRCh38
rs561530000 Chr.4: 99307579-99307579 on Build GRCh38
rs1045600042 Chr.4: 99307568-99307568 on Build GRCh38
rs949896138 Chr.4: 99307547-99307547 on Build GRCh38
rs548914849 Chr.4: 99307460-99307460 on Build GRCh38
rs963135976 Chr.4: 99307625-99307625 on Build GRCh38
rs375251171 Chr.4: 99307520-99307520 on Build GRCh38
rs764467403 Chr.4: 99307825-99307825 on Build GRCh38
rs1038330683 Chr.4: 99307597-99307597 on Build GRCh38
rs183895202 Chr.4: 99307603-99307603 on Build GRCh38
rs111229252 Chr.4: 99307564-99307564 on Build GRCh38
rs28914788 Chr.4: 99307734-99307734 on Build GRCh38
rs778750682 Chr.4: 99307573-99307573 on Build GRCh38
rs995951299 Chr.4: 99307534-99307534 on Build GRCh38
rs973126594 Chr.4: 99307653-99307653 on Build GRCh38
rs1000877928 Chr.4: 99307478-99307478 on Build GRCh38
rs757036512 Chr.4: 99307448-99307448 on Build GRCh38
rs979639583 Chr.4: 99307713-99307713 on Build GRCh38
rs1018395124 Chr.4: 99307688-99307688 on Build GRCh38
rs775974635 Chr.4: 99307805-99307805 on Build GRCh38
rs993872145 Chr.4: 99307643-99307643 on Build GRCh38
rs979692434 Chr.4: 99307442-99307442 on Build GRCh38
rs563121276 Chr.4: 99307407-99307407 on Build GRCh38
rs925277832 Chr.4: 99307505-99307505 on Build GRCh38
rs770345544 Chr.4: 99307804-99307804 on Build GRCh38
rs918178900 Chr.4: 99307732-99307732 on Build GRCh38
rs1042742756 Chr.4: 99307409-99307409 on Build GRCh38
rs955709970 Chr.4: 99307378-99307378 on Build GRCh38
rs199635405 Chr.4: 99307803-99307803 on Build GRCh38
rs200290314 Chr.4: 99307503-99307503 on Build GRCh38
rs1053571652 Chr.4: 99307504-99307504 on Build GRCh38
rs28914789 Chr.4: 99307381-99307382 on Build GRCh38
rs544936759 Chr.4: 99307406-99307406 on Build GRCh38
rs530584711 Chr.4: 99307419-99307419 on Build GRCh38
rs925560127 Chr.4: 99307727-99307727 on Build GRCh38
rs1025786461 Chr.4: 99307647-99307647 on Build GRCh38
rs17033 Chr.4: 99307788-99307788 on Build GRCh38
rs867026472 Chr.4: 99307781-99307781 on Build GRCh38
rs1052927975 Chr.4: 99307532-99307532 on Build GRCh38
rs936659841 Chr.4: 99307824-99307824 on Build GRCh38
rs990984385 Chr.4: 99307694-99307694 on Build GRCh38
rs771991530 Chr.4: 99307619-99307619 on Build GRCh38

View Full Product Details