Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99318695-99319191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH1B
Entrez Gene ID
125
Gene Name
alcohol dehydrogenase 1B (class I), beta polypeptide
Gene Aliases
ADH2, HEL-S-117
UniGene
Hs.4
Gene Chromosome Location
Chr.4: 99306387-99321442 on Build GRCh38
Transcripts
NM_000668.5 NM_001286650.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1618193 c.120+1G>A p.? Chr.4: 99318784-99318784 on Build GRCh38
3665424 c.107_120del14 p.E36fs*11 Chr.4: 99318785-99318798 on Build GRCh38
2951312 c.112C>T p.R38C Chr.4: 99318793-99318793 on Build GRCh38
3664638 c.85G>T p.V29F Chr.4: 99318820-99318820 on Build GRCh38
480670 c.114C>G p.R38R Chr.4: 99318791-99318791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781090391 Chr.4: 99318739-99318739 on Build GRCh38
rs1037809204 Chr.4: 99318832-99318832 on Build GRCh38
rs770117532 Chr.4: 99318793-99318793 on Build GRCh38
rs779569999 Chr.4: 99318751-99318751 on Build GRCh38
rs140880364 Chr.4: 99319021-99319021 on Build GRCh38
rs574604027 Chr.4: 99318977-99318977 on Build GRCh38
rs761102982 Chr.4: 99318843-99318843 on Build GRCh38
rs778258146 Chr.4: 99318775-99318775 on Build GRCh38
rs937508469 Chr.4: 99319076-99319076 on Build GRCh38
rs374975751 Chr.4: 99318854-99318854 on Build GRCh38
rs746299652 Chr.4: 99318911-99318911 on Build GRCh38
rs1013429974 Chr.4: 99319124-99319124 on Build GRCh38
rs753422844 Chr.4: 99318850-99318850 on Build GRCh38
rs762577054 Chr.4: 99318973-99318984 on Build GRCh38
rs780298656 Chr.4: 99319007-99319007 on Build GRCh38
rs186222387 Chr.4: 99318994-99318994 on Build GRCh38
rs768982796 Chr.4: 99318942-99318942 on Build GRCh38
rs757716482 Chr.4: 99318967-99318967 on Build GRCh38
rs933516634 Chr.4: 99318982-99318982 on Build GRCh38
rs201421877 Chr.4: 99318809-99318809 on Build GRCh38
rs555863882 Chr.4: 99318804-99318804 on Build GRCh38
rs765192867 Chr.4: 99318772-99318772 on Build GRCh38
rs149752262 Chr.4: 99319131-99319131 on Build GRCh38
rs72552716 Chr.4: 99318855-99318856 on Build GRCh38
rs375965352 Chr.4: 99318893-99318893 on Build GRCh38
rs999328789 Chr.4: 99319025-99319025 on Build GRCh38
rs750301947 Chr.4: 99318834-99318834 on Build GRCh38
rs969180605 Chr.4: 99318952-99318953 on Build GRCh38
rs28913912 Chr.4: 99318959-99318959 on Build GRCh38
rs763134176 Chr.4: 99318799-99318799 on Build GRCh38
rs775325816 Chr.4: 99318814-99318814 on Build GRCh38
rs773106812 Chr.4: 99318900-99318900 on Build GRCh38
rs776448706 Chr.4: 99319013-99319013 on Build GRCh38
rs28913913 Chr.4: 99318741-99318741 on Build GRCh38
rs747289055 Chr.4: 99319126-99319126 on Build GRCh38
rs562054182 Chr.4: 99319006-99319006 on Build GRCh38
rs751573568 Chr.4: 99318806-99318806 on Build GRCh38
rs1057236364 Chr.4: 99319060-99319060 on Build GRCh38
rs760523527 Chr.4: 99318962-99318962 on Build GRCh38
rs750468573 Chr.4: 99318975-99318975 on Build GRCh38
rs889374434 Chr.4: 99318997-99318997 on Build GRCh38
rs771467302 Chr.4: 99318896-99318896 on Build GRCh38
rs755834727 Chr.4: 99318744-99318744 on Build GRCh38
rs775558705 Chr.4: 99318795-99318795 on Build GRCh38
rs759320890 Chr.4: 99319017-99319017 on Build GRCh38
rs758986990 Chr.4: 99318763-99318763 on Build GRCh38
rs767328444 Chr.4: 99318831-99318831 on Build GRCh38
rs763221207 Chr.4: 99318936-99318936 on Build GRCh38
rs941134545 Chr.4: 99319037-99319037 on Build GRCh38
rs775782629 Chr.4: 99318927-99318927 on Build GRCh38
rs769068976 Chr.4: 99319000-99319000 on Build GRCh38
rs906020550 Chr.4: 99318842-99318842 on Build GRCh38
rs1229983 Chr.4: 99318845-99318845 on Build GRCh38
rs1044777158 Chr.4: 99319053-99319053 on Build GRCh38
rs748253259 Chr.4: 99319003-99319003 on Build GRCh38
rs200481568 Chr.4: 99318790-99318790 on Build GRCh38
rs749561302 Chr.4: 99319041-99319041 on Build GRCh38
rs771307803 Chr.4: 99318791-99318791 on Build GRCh38
rs777100313 Chr.4: 99318792-99318792 on Build GRCh38
rs78501598 Chr.4: 99318784-99318784 on Build GRCh38
rs771980997 Chr.4: 99319004-99319004 on Build GRCh38
rs778345918 Chr.4: 99318861-99318861 on Build GRCh38
rs774616214 Chr.4: 99318954-99318954 on Build GRCh38
rs376664416 Chr.4: 99318970-99318970 on Build GRCh38
rs114261077 Chr.4: 99318976-99318976 on Build GRCh38
rs373976723 Chr.4: 99318965-99318965 on Build GRCh38
rs1018144926 Chr.4: 99318966-99318966 on Build GRCh38
rs780618392 Chr.4: 99318989-99318989 on Build GRCh38
rs201450842 Chr.4: 99318764-99318764 on Build GRCh38
rs368357625 Chr.4: 99318914-99318914 on Build GRCh38
rs113075608 Chr.4: 99318859-99318859 on Build GRCh38
rs190699633 Chr.4: 99318957-99318957 on Build GRCh38
rs1010251860 Chr.4: 99319137-99319139 on Build GRCh38
rs115839326 Chr.4: 99319135-99319135 on Build GRCh38
rs55963727 Chr.4: 99318907-99318907 on Build GRCh38
rs761983697 Chr.4: 99318955-99318955 on Build GRCh38
rs866687371 Chr.4: 99319141-99319141 on Build GRCh38
rs766317953 Chr.4: 99319011-99319011 on Build GRCh38
rs757830318 Chr.4: 99318974-99318974 on Build GRCh38
rs747473547 Chr.4: 99318776-99318776 on Build GRCh38
rs564372124 Chr.4: 99318963-99318963 on Build GRCh38
rs896281868 Chr.4: 99319082-99319082 on Build GRCh38
rs774704345 Chr.4: 99319001-99319001 on Build GRCh38

View Full Product Details