Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 102884943-102885483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CISD2
Entrez Gene ID
493856
Gene Name
CDGSH iron sulfur domain 2
Gene Aliases
ERIS, Miner1, NAF-1, WFS2, ZCD2
UniGene
Hs.444955
Gene Chromosome Location
Chr.4: 102868978-102892807 on Build GRCh38
Transcripts
NM_001008388.4 NM_001100874.2 NR_047513.1 NR_047515.1

Gene Symbol
SLC9B1
Entrez Gene ID
150159
Gene Name
solute carrier family 9 member B1
Gene Aliases
NHA1, NHEDC1
UniGene
Hs.666728
Gene Chromosome Location
Chr.4: 102885048-103019739 on Build GRCh38
Transcripts
NM_001008388.4 NM_001100874.2 NR_047513.1 NR_047515.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4653754 c.307C>T p.R103C Chr.4: 102885419-102885419 on Build GRCh38
6629341 c.119G>A p.R40Q Chr.4: 102885231-102885231 on Build GRCh38
1426149 c.110A>G p.E37G Chr.4: 102885222-102885222 on Build GRCh38
6098795 c.180C>T p.L60L Chr.4: 102885292-102885292 on Build GRCh38
1049601 c.118C>T p.R40W Chr.4: 102885230-102885230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970406087 Chr.4: 102885085-102885085 on Build GRCh38
rs772159844 Chr.4: 102885302-102885302 on Build GRCh38
rs754165955 Chr.4: 102885448-102885448 on Build GRCh38
rs760194746 Chr.4: 102885203-102885203 on Build GRCh38
rs63749888 Chr.4: 102885221-102885221 on Build GRCh38
rs749424337 Chr.4: 102885172-102885172 on Build GRCh38
rs970411640 Chr.4: 102885318-102885318 on Build GRCh38
rs756559515 Chr.4: 102885415-102885415 on Build GRCh38
rs768049003 Chr.4: 102885409-102885409 on Build GRCh38
rs375816656 Chr.4: 102885307-102885307 on Build GRCh38
rs370219158 Chr.4: 102885368-102885368 on Build GRCh38
rs35282200 Chr.4: 102885154-102885155 on Build GRCh38
rs775550185 Chr.4: 102885169-102885169 on Build GRCh38
rs779698439 Chr.4: 102885194-102885194 on Build GRCh38
rs764899307 Chr.4: 102885371-102885371 on Build GRCh38
rs905455060 Chr.4: 102885009-102885009 on Build GRCh38
rs554973807 Chr.4: 102885199-102885199 on Build GRCh38
rs905836999 Chr.4: 102885376-102885376 on Build GRCh38
rs890534906 Chr.4: 102885154-102885154 on Build GRCh38
rs142368879 Chr.4: 102885150-102885151 on Build GRCh38
rs768170324 Chr.4: 102885455-102885455 on Build GRCh38
rs868634687 Chr.4: 102885351-102885351 on Build GRCh38
rs763904771 Chr.4: 102885232-102885232 on Build GRCh38
rs770471638 Chr.4: 102885278-102885278 on Build GRCh38
rs752222512 Chr.4: 102885375-102885375 on Build GRCh38
rs139460212 Chr.4: 102885450-102885450 on Build GRCh38
rs371712100 Chr.4: 102885256-102885256 on Build GRCh38
rs766086013 Chr.4: 102885338-102885338 on Build GRCh38
rs766107276 Chr.4: 102885220-102885220 on Build GRCh38
rs375375350 Chr.4: 102885188-102885188 on Build GRCh38
rs781626534 Chr.4: 102885263-102885263 on Build GRCh38
rs532591680 Chr.4: 102885003-102885003 on Build GRCh38
rs1026429946 Chr.4: 102885138-102885138 on Build GRCh38
rs746277672 Chr.4: 102885270-102885270 on Build GRCh38
rs988521297 Chr.4: 102885151-102885151 on Build GRCh38
rs760499330 Chr.4: 102885209-102885209 on Build GRCh38
rs765050847 Chr.4: 102885443-102885443 on Build GRCh38
rs756402482 Chr.4: 102885277-102885277 on Build GRCh38
rs75722531 Chr.4: 102885127-102885127 on Build GRCh38
rs550618386 Chr.4: 102885080-102885080 on Build GRCh38
rs757788592 Chr.4: 102885257-102885257 on Build GRCh38
rs190212874 Chr.4: 102885207-102885207 on Build GRCh38
rs753403300 Chr.4: 102885230-102885230 on Build GRCh38
rs780294206 Chr.4: 102885447-102885447 on Build GRCh38
rs1037719932 Chr.4: 102885025-102885025 on Build GRCh38
rs369622813 Chr.4: 102885454-102885454 on Build GRCh38
rs1006260464 Chr.4: 102885019-102885019 on Build GRCh38
rs764643608 Chr.4: 102885251-102885251 on Build GRCh38
rs750923436 Chr.4: 102885412-102885412 on Build GRCh38
rs199702688 Chr.4: 102885295-102885295 on Build GRCh38
rs146299528 Chr.4: 102885231-102885231 on Build GRCh38
rs780388471 Chr.4: 102885282-102885282 on Build GRCh38
rs141987332 Chr.4: 102885187-102885187 on Build GRCh38
rs993494308 Chr.4: 102885133-102885133 on Build GRCh38
rs770939981 Chr.4: 102885151-102885152 on Build GRCh38
rs779230538 Chr.4: 102885453-102885453 on Build GRCh38
rs796666622 Chr.4: 102885152-102885153 on Build GRCh38
rs142253163 Chr.4: 102885294-102885294 on Build GRCh38
rs1008100897 Chr.4: 102885157-102885157 on Build GRCh38
rs545863501 Chr.4: 102885063-102885063 on Build GRCh38
rs375529885 Chr.4: 102885352-102885352 on Build GRCh38
rs577064952 Chr.4: 102885337-102885337 on Build GRCh38
rs981360085 Chr.4: 102885091-102885091 on Build GRCh38
rs938229185 Chr.4: 102885011-102885011 on Build GRCh38
rs772249990 Chr.4: 102885457-102885457 on Build GRCh38
rs1022958722 Chr.4: 102885037-102885037 on Build GRCh38
rs866399747 Chr.4: 102885276-102885276 on Build GRCh38
rs374058087 Chr.4: 102885284-102885284 on Build GRCh38
rs151223492 Chr.4: 102885406-102885406 on Build GRCh38

View Full Product Details