Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 102889987-102890499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CISD2
Entrez Gene ID
493856
Gene Name
CDGSH iron sulfur domain 2
Gene Aliases
ERIS, Miner1, NAF-1, WFS2, ZCD2
UniGene
Hs.444955
Gene Chromosome Location
Chr.4: 102868978-102892807 on Build GRCh38
Transcripts
NM_001008388.4 NM_001100874.2 NR_047513.1 NR_047515.1

Gene Symbol
SLC9B1
Entrez Gene ID
150159
Gene Name
solute carrier family 9 member B1
Gene Aliases
NHA1, NHEDC1
UniGene
Hs.666728
Gene Chromosome Location
Chr.4: 102885048-103019739 on Build GRCh38
Transcripts
NM_001008388.4 NM_001100874.2 NR_047513.1 NR_047515.1

SNP Detail

SNP ID Chromosome Location
rs223310 Chr.4: 102890220-102890220 on Build GRCh38
rs566418032 Chr.4: 102890156-102890156 on Build GRCh38
rs541067227 Chr.4: 102890300-102890300 on Build GRCh38
rs867480356 Chr.4: 102890203-102890203 on Build GRCh38
rs565303571 Chr.4: 102890358-102890361 on Build GRCh38
rs989489225 Chr.4: 102890259-102890259 on Build GRCh38
rs1027685863 Chr.4: 102890203-102890205 on Build GRCh38
rs776228466 Chr.4: 102890301-102890301 on Build GRCh38
rs577542582 Chr.4: 102890446-102890446 on Build GRCh38
rs944448634 Chr.4: 102890358-102890359 on Build GRCh38
rs190765542 Chr.4: 102890251-102890251 on Build GRCh38
rs559218368 Chr.4: 102890428-102890428 on Build GRCh38
rs915172408 Chr.4: 102890290-102890290 on Build GRCh38
rs1050706627 Chr.4: 102890323-102890323 on Build GRCh38
rs545747426 Chr.4: 102890309-102890311 on Build GRCh38
rs113008381 Chr.4: 102890227-102890227 on Build GRCh38
rs555830953 Chr.4: 102890285-102890285 on Build GRCh38
rs113787390 Chr.4: 102890107-102890107 on Build GRCh38
rs897572830 Chr.4: 102890418-102890418 on Build GRCh38
rs1008629240 Chr.4: 102890052-102890052 on Build GRCh38
rs1050257791 Chr.4: 102890012-102890012 on Build GRCh38
rs142131629 Chr.4: 102890454-102890457 on Build GRCh38
rs956666137 Chr.4: 102890232-102890232 on Build GRCh38
rs573910212 Chr.4: 102890298-102890298 on Build GRCh38
rs890234405 Chr.4: 102890046-102890046 on Build GRCh38
rs1023785427 Chr.4: 102890217-102890217 on Build GRCh38
rs994426600 Chr.4: 102890202-102890202 on Build GRCh38
rs181627301 Chr.4: 102890462-102890462 on Build GRCh38
rs991423246 Chr.4: 102890214-102890214 on Build GRCh38
rs540036719 Chr.4: 102890192-102890192 on Build GRCh38
rs911701368 Chr.4: 102890338-102890338 on Build GRCh38
rs541623382 Chr.4: 102890153-102890153 on Build GRCh38
rs532358002 Chr.4: 102890456-102890457 on Build GRCh38
rs1014612636 Chr.4: 102890112-102890112 on Build GRCh38

View Full Product Details