Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 103078127-103078634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDH2
Entrez Gene ID
56898
Gene Name
3-hydroxybutyrate dehydrogenase, type 2
Gene Aliases
DHRS6, EFA6R, PRO20933, SDR15C1, UCPA-OR, UNQ6308
UniGene
Hs.124696
Gene Chromosome Location
Chr.4: 103077618-103099867 on Build GRCh38
Transcripts
NM_020139.3 XM_005263140.3 XM_006714274.3 XM_011532127.2 XM_011532128.2 XM_017008461.1 XM_017008462.1

SNP Detail

SNP ID Chromosome Location
rs369331486 Chr.4: 103078190-103078190 on Build GRCh38
rs917695429 Chr.4: 103078214-103078214 on Build GRCh38
rs903531140 Chr.4: 103078285-103078285 on Build GRCh38
rs955473202 Chr.4: 103078596-103078596 on Build GRCh38
rs1000904780 Chr.4: 103078296-103078296 on Build GRCh38
rs997047416 Chr.4: 103078505-103078505 on Build GRCh38
rs535859805 Chr.4: 103078366-103078366 on Build GRCh38
rs900058468 Chr.4: 103078413-103078413 on Build GRCh38
rs192568126 Chr.4: 103078543-103078543 on Build GRCh38
rs773666244 Chr.4: 103078348-103078348 on Build GRCh38
rs753171328 Chr.4: 103078424-103078424 on Build GRCh38
rs768065384 Chr.4: 103078191-103078191 on Build GRCh38
rs945120886 Chr.4: 103078217-103078217 on Build GRCh38
rs1054725661 Chr.4: 103078367-103078367 on Build GRCh38
rs1042076861 Chr.4: 103078247-103078247 on Build GRCh38
rs1056683723 Chr.4: 103078164-103078164 on Build GRCh38
rs558758993 Chr.4: 103078571-103078575 on Build GRCh38
rs769251042 Chr.4: 103078579-103078583 on Build GRCh38

View Full Product Details