Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 103082392-103082942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDH2
Entrez Gene ID
56898
Gene Name
3-hydroxybutyrate dehydrogenase, type 2
Gene Aliases
DHRS6, EFA6R, PRO20933, SDR15C1, UCPA-OR, UNQ6308
UniGene
Hs.124696
Gene Chromosome Location
Chr.4: 103077618-103099867 on Build GRCh38
Transcripts
NM_020139.3 XM_005263140.3 XM_006714274.3 XM_011532127.2 XM_011532128.2 XM_017008461.1 XM_017008462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6611143 c.545C>T p.T182M Chr.4: 103082917-103082917 on Build GRCh38
6098782 c.533G>T p.G178V Chr.4: 103082929-103082929 on Build GRCh38
6611144 c.561A>G p.E187E Chr.4: 103082901-103082901 on Build GRCh38
7258199 c.572C>T p.A191V Chr.4: 103082890-103082890 on Build GRCh38
6287817 c.572C>A p.A191D Chr.4: 103082890-103082890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904677551 Chr.4: 103082805-103082805 on Build GRCh38
rs1022810736 Chr.4: 103082647-103082647 on Build GRCh38
rs771025882 Chr.4: 103082821-103082821 on Build GRCh38
rs545957427 Chr.4: 103082684-103082684 on Build GRCh38
rs60298331 Chr.4: 103082657-103082657 on Build GRCh38
rs901909064 Chr.4: 103082462-103082462 on Build GRCh38
rs763730896 Chr.4: 103082876-103082876 on Build GRCh38
rs762505122 Chr.4: 103082636-103082636 on Build GRCh38
rs767335800 Chr.4: 103082720-103082720 on Build GRCh38
rs1043220178 Chr.4: 103082797-103082797 on Build GRCh38
rs149044895 Chr.4: 103082787-103082787 on Build GRCh38
rs754888416 Chr.4: 103082691-103082691 on Build GRCh38
rs969287856 Chr.4: 103082644-103082644 on Build GRCh38
rs914223944 Chr.4: 103082735-103082735 on Build GRCh38
rs80208844 Chr.4: 103082690-103082690 on Build GRCh38
rs981222850 Chr.4: 103082664-103082664 on Build GRCh38
rs999087357 Chr.4: 103082812-103082812 on Build GRCh38
rs887790399 Chr.4: 103082863-103082863 on Build GRCh38
rs999036453 Chr.4: 103082489-103082489 on Build GRCh38
rs56412006 Chr.4: 103082836-103082836 on Build GRCh38
rs918952120 Chr.4: 103082710-103082710 on Build GRCh38
rs762803668 Chr.4: 103082867-103082867 on Build GRCh38
rs79460377 Chr.4: 103082907-103082907 on Build GRCh38
rs775208549 Chr.4: 103082850-103082850 on Build GRCh38
rs186476150 Chr.4: 103082727-103082727 on Build GRCh38
rs60495707 Chr.4: 103082916-103082916 on Build GRCh38
rs560818402 Chr.4: 103082648-103082648 on Build GRCh38
rs531650464 Chr.4: 103082709-103082709 on Build GRCh38
rs932965121 Chr.4: 103082775-103082775 on Build GRCh38
rs1031652257 Chr.4: 103082492-103082492 on Build GRCh38
rs901960168 Chr.4: 103082815-103082815 on Build GRCh38
rs1044383055 Chr.4: 103082761-103082761 on Build GRCh38
rs1051554873 Chr.4: 103082781-103082781 on Build GRCh38
rs952042083 Chr.4: 103082628-103082628 on Build GRCh38
rs576841758 Chr.4: 103082742-103082742 on Build GRCh38
rs143019882 Chr.4: 103082793-103082793 on Build GRCh38
rs1021712180 Chr.4: 103082452-103082452 on Build GRCh38
rs776654660 Chr.4: 103082824-103082824 on Build GRCh38
rs573572548 Chr.4: 103082528-103082528 on Build GRCh38
rs542194299 Chr.4: 103082555-103082555 on Build GRCh38
rs112941797 Chr.4: 103082464-103082464 on Build GRCh38
rs755443565 Chr.4: 103082865-103082865 on Build GRCh38
rs765603488 Chr.4: 103082450-103082450 on Build GRCh38
rs774074597 Chr.4: 103082894-103082894 on Build GRCh38
rs909010415 Chr.4: 103082675-103082675 on Build GRCh38
rs960296376 Chr.4: 103082527-103082527 on Build GRCh38
rs191505987 Chr.4: 103082773-103082773 on Build GRCh38

View Full Product Details