Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 103106062-103106550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPE
Entrez Gene ID
1062
Gene Name
centromere protein E
Gene Aliases
CENP-E, KIF10, MCPH13, PPP1R61
UniGene
Hs.75573
Gene Chromosome Location
Chr.4: 103105806-103198409 on Build GRCh38
Transcripts
NM_001286734.1 NM_001813.2 XM_011531544.2 XM_011531545.2 XM_011531546.2 XM_011531547.2 XM_011531548.2 XM_011531549.2 XM_017007659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6165924 c.8106G>C p.*2702Y Chr.4: 103106222-103106222 on Build GRCh38
1671177 c.8079G>T p.K2693N Chr.4: 103106249-103106249 on Build GRCh38
4977425 c.8032G>C p.E2678Q Chr.4: 103106296-103106296 on Build GRCh38
4776328 c.8029G>A p.A2677T Chr.4: 103106299-103106299 on Build GRCh38
4836659 c.8061G>A p.W2687* Chr.4: 103106267-103106267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747847978 Chr.4: 103106278-103106278 on Build GRCh38
rs1025104116 Chr.4: 103106378-103106378 on Build GRCh38
rs143981900 Chr.4: 103106089-103106089 on Build GRCh38
rs772570716 Chr.4: 103106171-103106171 on Build GRCh38
rs61756293 Chr.4: 103106248-103106248 on Build GRCh38
rs892567208 Chr.4: 103106340-103106340 on Build GRCh38
rs974217690 Chr.4: 103106098-103106098 on Build GRCh38
rs951119051 Chr.4: 103106452-103106452 on Build GRCh38
rs548608282 Chr.4: 103106402-103106402 on Build GRCh38
rs763324073 Chr.4: 103106220-103106220 on Build GRCh38
rs779866972 Chr.4: 103106260-103106260 on Build GRCh38
rs753178851 Chr.4: 103106218-103106218 on Build GRCh38
rs894977469 Chr.4: 103106456-103106456 on Build GRCh38
rs371147610 Chr.4: 103106189-103106189 on Build GRCh38
rs936643135 Chr.4: 103106512-103106512 on Build GRCh38
rs573901768 Chr.4: 103106103-103106103 on Build GRCh38
rs756144864 Chr.4: 103106259-103106259 on Build GRCh38
rs907328834 Chr.4: 103106386-103106386 on Build GRCh38
rs763608299 Chr.4: 103106143-103106143 on Build GRCh38
rs1045691643 Chr.4: 103106372-103106372 on Build GRCh38
rs560935001 Chr.4: 103106406-103106406 on Build GRCh38
rs775887389 Chr.4: 103106305-103106305 on Build GRCh38
rs936144531 Chr.4: 103106252-103106252 on Build GRCh38
rs530431919 Chr.4: 103106388-103106388 on Build GRCh38
rs1039662971 Chr.4: 103106263-103106263 on Build GRCh38
rs772841784 Chr.4: 103106282-103106282 on Build GRCh38
rs541298252 Chr.4: 103106133-103106133 on Build GRCh38
rs912450297 Chr.4: 103106243-103106243 on Build GRCh38
rs912396601 Chr.4: 103106469-103106469 on Build GRCh38
rs145876863 Chr.4: 103106229-103106229 on Build GRCh38
rs771832973 Chr.4: 103106279-103106279 on Build GRCh38
rs994649680 Chr.4: 103106475-103106475 on Build GRCh38
rs76191558 Chr.4: 103106341-103106341 on Build GRCh38
rs973785232 Chr.4: 103106441-103106441 on Build GRCh38
rs763413612 Chr.4: 103106306-103106306 on Build GRCh38
rs1054260235 Chr.4: 103106303-103106303 on Build GRCh38
rs1033966669 Chr.4: 103106418-103106418 on Build GRCh38
rs562994759 Chr.4: 103106343-103106343 on Build GRCh38
rs936713162 Chr.4: 103106277-103106277 on Build GRCh38
rs781298329 Chr.4: 103106249-103106249 on Build GRCh38
rs566157637 Chr.4: 103106521-103106521 on Build GRCh38
rs929772460 Chr.4: 103106167-103106168 on Build GRCh38
rs368293438 Chr.4: 103106292-103106292 on Build GRCh38
rs370178536 Chr.4: 103106136-103106136 on Build GRCh38
rs17215862 Chr.4: 103106487-103106487 on Build GRCh38
rs754849965 Chr.4: 103106265-103106265 on Build GRCh38
rs749198775 Chr.4: 103106264-103106264 on Build GRCh38
rs778819691 Chr.4: 103106274-103106274 on Build GRCh38
rs946071087 Chr.4: 103106347-103106347 on Build GRCh38
rs921213889 Chr.4: 103106500-103106500 on Build GRCh38
rs751815365 Chr.4: 103106230-103106230 on Build GRCh38
rs965772335 Chr.4: 103106145-103106145 on Build GRCh38
rs1027121928 Chr.4: 103106445-103106445 on Build GRCh38
rs1055125867 Chr.4: 103106276-103106276 on Build GRCh38
rs983091636 Chr.4: 103106134-103106134 on Build GRCh38
rs1002404995 Chr.4: 103106387-103106387 on Build GRCh38
rs376192945 Chr.4: 103106209-103106209 on Build GRCh38
rs553391225 Chr.4: 103106496-103106496 on Build GRCh38
rs114178434 Chr.4: 103106242-103106242 on Build GRCh38
rs762082215 Chr.4: 103106357-103106357 on Build GRCh38
rs1055842027 Chr.4: 103106443-103106443 on Build GRCh38
rs770419707 Chr.4: 103106304-103106304 on Build GRCh38
rs1002019376 Chr.4: 103106435-103106435 on Build GRCh38
rs746503087 Chr.4: 103106297-103106297 on Build GRCh38
rs760143314 Chr.4: 103106196-103106196 on Build GRCh38

View Full Product Details