Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 104472031-104472537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXXC4
Entrez Gene ID
80319
Gene Name
CXXC finger protein 4
Gene Aliases
IDAX
UniGene
Hs.12248
Gene Chromosome Location
Chr.4: 104468306-104494967 on Build GRCh38
Transcripts
NM_025212.3 NR_132741.1 XM_011532284.2 XM_017008652.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5931474 c.1072C>T p.P358S Chr.4: 104472354-104472354 on Build GRCh38
6638691 c.1060-5_1060-4insT p.? Chr.4: 104472370-104472371 on Build GRCh38
6638690 c.553-5_553-4insT p.? Chr.4: 104472370-104472371 on Build GRCh38
4862703 c.1075A>T p.S359C Chr.4: 104472351-104472351 on Build GRCh38
3824886 c.1077C>T p.S359S Chr.4: 104472349-104472349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758301586 Chr.4: 104472398-104472398 on Build GRCh38
rs763750056 Chr.4: 104472348-104472348 on Build GRCh38
rs925401992 Chr.4: 104472097-104472097 on Build GRCh38
rs4413407 Chr.4: 104472204-104472204 on Build GRCh38
rs752219245 Chr.4: 104472335-104472335 on Build GRCh38
rs778563364 Chr.4: 104472354-104472354 on Build GRCh38
rs962116661 Chr.4: 104472421-104472421 on Build GRCh38
rs750882743 Chr.4: 104472317-104472317 on Build GRCh38
rs4282179 Chr.4: 104472163-104472163 on Build GRCh38
rs1052981124 Chr.4: 104472103-104472103 on Build GRCh38
rs548424806 Chr.4: 104472472-104472472 on Build GRCh38
rs537461126 Chr.4: 104472441-104472441 on Build GRCh38
rs574570374 Chr.4: 104472382-104472382 on Build GRCh38
rs767057348 Chr.4: 104472321-104472321 on Build GRCh38
rs1045065504 Chr.4: 104472118-104472118 on Build GRCh38
rs896711163 Chr.4: 104472098-104472098 on Build GRCh38
rs924327127 Chr.4: 104472508-104472508 on Build GRCh38
rs918000318 Chr.4: 104472112-104472112 on Build GRCh38
rs755731838 Chr.4: 104472341-104472341 on Build GRCh38
rs150596658 Chr.4: 104472438-104472438 on Build GRCh38
rs34493088 Chr.4: 104472172-104472173 on Build GRCh38
rs775467301 Chr.4: 104472370-104472371 on Build GRCh38
rs568367499 Chr.4: 104472509-104472509 on Build GRCh38
rs763540720 Chr.4: 104472318-104472318 on Build GRCh38
rs968334764 Chr.4: 104472498-104472498 on Build GRCh38
rs71600293 Chr.4: 104472172-104472172 on Build GRCh38
rs369441679 Chr.4: 104472346-104472346 on Build GRCh38
rs1038209759 Chr.4: 104472201-104472201 on Build GRCh38
rs773103551 Chr.4: 104472442-104472442 on Build GRCh38
rs141779912 Chr.4: 104472427-104472427 on Build GRCh38
rs1045032985 Chr.4: 104472143-104472143 on Build GRCh38
rs181795114 Chr.4: 104472093-104472093 on Build GRCh38
rs753440112 Chr.4: 104472349-104472349 on Build GRCh38
rs994007947 Chr.4: 104472251-104472251 on Build GRCh38
rs765523363 Chr.4: 104472315-104472315 on Build GRCh38
rs997282368 Chr.4: 104472423-104472423 on Build GRCh38
rs953307678 Chr.4: 104472431-104472431 on Build GRCh38
rs776742962 Chr.4: 104472279-104472279 on Build GRCh38
rs547628940 Chr.4: 104472202-104472202 on Build GRCh38
rs771598998 Chr.4: 104472083-104472086 on Build GRCh38
rs757022413 Chr.4: 104472352-104472352 on Build GRCh38
rs762198997 Chr.4: 104472295-104472295 on Build GRCh38
rs748325838 Chr.4: 104472224-104472224 on Build GRCh38
rs527832213 Chr.4: 104472187-104472187 on Build GRCh38
rs1006181186 Chr.4: 104472254-104472254 on Build GRCh38
rs979641077 Chr.4: 104472075-104472075 on Build GRCh38
rs1001834683 Chr.4: 104472175-104472176 on Build GRCh38
rs769933616 Chr.4: 104472422-104472422 on Build GRCh38
rs745646592 Chr.4: 104472115-104472115 on Build GRCh38
rs745734320 Chr.4: 104472370-104472370 on Build GRCh38
rs532113204 Chr.4: 104472291-104472291 on Build GRCh38
rs977092201 Chr.4: 104472507-104472507 on Build GRCh38

View Full Product Details