Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105636191-105636694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF38
Entrez Gene ID
54848
Gene Name
Rho guanine nucleotide exchange factor 38
Gene Aliases
-
UniGene
Hs.585224
Gene Chromosome Location
Chr.4: 105552603-105682539 on Build GRCh38
Transcripts
NM_001242729.1 XM_006714244.3 XM_011532050.2 XM_011532051.2

SNP Detail

SNP ID Chromosome Location
rs1012895858 Chr.4: 105636287-105636287 on Build GRCh38
rs1018459031 Chr.4: 105636228-105636230 on Build GRCh38
rs187414672 Chr.4: 105636282-105636282 on Build GRCh38
rs928966549 Chr.4: 105636454-105636454 on Build GRCh38
rs1022982922 Chr.4: 105636311-105636311 on Build GRCh38
rs971769772 Chr.4: 105636404-105636404 on Build GRCh38
rs893242077 Chr.4: 105636276-105636276 on Build GRCh38
rs941953211 Chr.4: 105636336-105636336 on Build GRCh38
rs902995989 Chr.4: 105636473-105636473 on Build GRCh38
rs766258086 Chr.4: 105636442-105636442 on Build GRCh38
rs563163703 Chr.4: 105636446-105636446 on Build GRCh38
rs910257241 Chr.4: 105636464-105636464 on Build GRCh38
rs1054301349 Chr.4: 105636562-105636562 on Build GRCh38
rs920322047 Chr.4: 105636434-105636434 on Build GRCh38
rs926505864 Chr.4: 105636314-105636314 on Build GRCh38
rs755982394 Chr.4: 105636361-105636361 on Build GRCh38
rs944612868 Chr.4: 105636465-105636465 on Build GRCh38
rs867043765 Chr.4: 105636609-105636609 on Build GRCh38
rs1001114535 Chr.4: 105636618-105636618 on Build GRCh38
rs1032377978 Chr.4: 105636245-105636245 on Build GRCh38
rs973470092 Chr.4: 105636359-105636359 on Build GRCh38
rs143040504 Chr.4: 105636243-105636243 on Build GRCh38
rs10016369 Chr.4: 105636575-105636575 on Build GRCh38
rs551377841 Chr.4: 105636265-105636265 on Build GRCh38
rs970518546 Chr.4: 105636295-105636298 on Build GRCh38
rs972019140 Chr.4: 105636313-105636313 on Build GRCh38
rs957004619 Chr.4: 105636252-105636252 on Build GRCh38
rs961596829 Chr.4: 105636403-105636403 on Build GRCh38
rs1016170706 Chr.4: 105636372-105636372 on Build GRCh38
rs111720147 Chr.4: 105636493-105636493 on Build GRCh38
rs767224160 Chr.4: 105636463-105636463 on Build GRCh38
rs572608005 Chr.4: 105636228-105636228 on Build GRCh38
rs1039878523 Chr.4: 105636549-105636549 on Build GRCh38
rs907236610 Chr.4: 105636651-105636651 on Build GRCh38
rs17261094 Chr.4: 105636272-105636272 on Build GRCh38
rs1023386024 Chr.4: 105636623-105636623 on Build GRCh38
rs1040318632 Chr.4: 105636471-105636471 on Build GRCh38
rs981758045 Chr.4: 105636355-105636355 on Build GRCh38
rs775443482 Chr.4: 105636478-105636478 on Build GRCh38
rs532090666 Chr.4: 105636374-105636374 on Build GRCh38
rs1013010588 Chr.4: 105636620-105636620 on Build GRCh38

View Full Product Details