Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107950113-107950641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2U1
Entrez Gene ID
113612
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
P450TEC, SPG49, SPG56
UniGene
Hs.109087
Gene Chromosome Location
Chr.4: 107931549-107953461 on Build GRCh38
Transcripts
NM_183075.2 XM_005262717.2 XM_005262720.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4121217 c.1570C>A p.L524M Chr.4: 107950358-107950358 on Build GRCh38
4851115 c.1634G>T p.*545L Chr.4: 107950422-107950422 on Build GRCh38
5014444 c.1497A>C p.E499D Chr.4: 107950285-107950285 on Build GRCh38
5681317 c.1603C>T p.H535Y Chr.4: 107950391-107950391 on Build GRCh38
4527564 c.1473G>T p.M491I Chr.4: 107950261-107950261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772136947 Chr.4: 107950418-107950418 on Build GRCh38
rs762873672 Chr.4: 107950251-107950251 on Build GRCh38
rs758717282 Chr.4: 107950221-107950222 on Build GRCh38
rs773047007 Chr.4: 107950372-107950372 on Build GRCh38
rs977349364 Chr.4: 107950421-107950421 on Build GRCh38
rs141431913 Chr.4: 107950250-107950250 on Build GRCh38
rs780578327 Chr.4: 107950336-107950336 on Build GRCh38
rs770825730 Chr.4: 107950393-107950393 on Build GRCh38
rs774394422 Chr.4: 107950260-107950260 on Build GRCh38
rs940928624 Chr.4: 107950253-107950253 on Build GRCh38
rs370914955 Chr.4: 107950444-107950444 on Build GRCh38
rs995234654 Chr.4: 107950307-107950307 on Build GRCh38
rs564888809 Chr.4: 107950140-107950140 on Build GRCh38
rs148315333 Chr.4: 107950249-107950249 on Build GRCh38
rs538538497 Chr.4: 107950442-107950442 on Build GRCh38
rs201919750 Chr.4: 107950434-107950434 on Build GRCh38
rs1028966851 Chr.4: 107950313-107950313 on Build GRCh38
rs763979652 Chr.4: 107950181-107950181 on Build GRCh38
rs183327899 Chr.4: 107950457-107950457 on Build GRCh38
rs553600162 Chr.4: 107950556-107950556 on Build GRCh38
rs530434750 Chr.4: 107950141-107950141 on Build GRCh38
rs372600671 Chr.4: 107950321-107950321 on Build GRCh38
rs760235478 Chr.4: 107950493-107950493 on Build GRCh38
rs571567845 Chr.4: 107950491-107950491 on Build GRCh38
rs80203932 Chr.4: 107950555-107950555 on Build GRCh38
rs1038198482 Chr.4: 107950573-107950573 on Build GRCh38
rs747768954 Chr.4: 107950352-107950352 on Build GRCh38
rs752207434 Chr.4: 107950465-107950465 on Build GRCh38
rs759654099 Chr.4: 107950271-107950271 on Build GRCh38
rs776306146 Chr.4: 107950211-107950211 on Build GRCh38
rs780269996 Chr.4: 107950452-107950453 on Build GRCh38
rs368543204 Chr.4: 107950280-107950280 on Build GRCh38
rs747776098 Chr.4: 107950232-107950232 on Build GRCh38
rs576923939 Chr.4: 107950419-107950419 on Build GRCh38
rs979798894 Chr.4: 107950557-107950557 on Build GRCh38
rs752917310 Chr.4: 107950281-107950281 on Build GRCh38
rs760935150 Chr.4: 107950424-107950424 on Build GRCh38
rs762225242 Chr.4: 107950438-107950438 on Build GRCh38
rs79009744 Chr.4: 107950454-107950454 on Build GRCh38
rs757661797 Chr.4: 107950559-107950560 on Build GRCh38
rs144144031 Chr.4: 107950403-107950403 on Build GRCh38
rs191760145 Chr.4: 107950201-107950201 on Build GRCh38
rs551591505 Chr.4: 107950469-107950469 on Build GRCh38
rs749132333 Chr.4: 107950376-107950376 on Build GRCh38
rs754308246 Chr.4: 107950312-107950312 on Build GRCh38
rs377630638 Chr.4: 107950492-107950492 on Build GRCh38
rs769574546 Chr.4: 107950236-107950236 on Build GRCh38
rs139224667 Chr.4: 107950324-107950324 on Build GRCh38
rs544449820 Chr.4: 107950192-107950192 on Build GRCh38
rs557596915 Chr.4: 107950547-107950547 on Build GRCh38
rs915631856 Chr.4: 107950173-107950173 on Build GRCh38
rs377467252 Chr.4: 107950427-107950427 on Build GRCh38
rs373549777 Chr.4: 107950360-107950360 on Build GRCh38
rs1044155099 Chr.4: 107950177-107950177 on Build GRCh38
rs765717394 Chr.4: 107950443-107950443 on Build GRCh38
rs536290976 Chr.4: 107950567-107950567 on Build GRCh38
rs1033325675 Chr.4: 107950435-107950435 on Build GRCh38
rs371017677 Chr.4: 107950458-107950458 on Build GRCh38
rs1036607521 Chr.4: 107950269-107950269 on Build GRCh38
rs537070614 Chr.4: 107950531-107950531 on Build GRCh38
rs529806409 Chr.4: 107950433-107950433 on Build GRCh38
rs991278699 Chr.4: 107950149-107950149 on Build GRCh38
rs759494725 Chr.4: 107950412-107950412 on Build GRCh38
rs150818323 Chr.4: 107950322-107950322 on Build GRCh38
rs756308496 Chr.4: 107950295-107950295 on Build GRCh38
rs188684590 Chr.4: 107950568-107950568 on Build GRCh38
rs750021329 Chr.4: 107950518-107950518 on Build GRCh38
rs866680080 Chr.4: 107950546-107950546 on Build GRCh38
rs750753356 Chr.4: 107950213-107950213 on Build GRCh38
rs764456836 Chr.4: 107950297-107950297 on Build GRCh38
rs796932488 Chr.4: 107950618-107950618 on Build GRCh38
rs981151496 Chr.4: 107950501-107950501 on Build GRCh38
rs754487808 Chr.4: 107950334-107950334 on Build GRCh38
rs764362029 Chr.4: 107950314-107950314 on Build GRCh38
rs956311267 Chr.4: 107950497-107950497 on Build GRCh38

View Full Product Details