Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107951462-107952000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2U1
Entrez Gene ID
113612
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
P450TEC, SPG49, SPG56
UniGene
Hs.109087
Gene Chromosome Location
Chr.4: 107931549-107953461 on Build GRCh38
Transcripts
NM_183075.2 XM_005262717.2 XM_005262720.2

SNP Detail

SNP ID Chromosome Location
rs1002752895 Chr.4: 107951485-107951485 on Build GRCh38
rs948784401 Chr.4: 107951760-107951760 on Build GRCh38
rs1034582864 Chr.4: 107951520-107951520 on Build GRCh38
rs190255112 Chr.4: 107951765-107951765 on Build GRCh38
rs914648196 Chr.4: 107951726-107951726 on Build GRCh38
rs913271052 Chr.4: 107951919-107951919 on Build GRCh38
rs925194925 Chr.4: 107951875-107951875 on Build GRCh38
rs562528248 Chr.4: 107951643-107951643 on Build GRCh38
rs976271080 Chr.4: 107951670-107951670 on Build GRCh38
rs946105657 Chr.4: 107951936-107951936 on Build GRCh38
rs990661012 Chr.4: 107951723-107951723 on Build GRCh38
rs111975834 Chr.4: 107951697-107951697 on Build GRCh38
rs115289507 Chr.4: 107951895-107951895 on Build GRCh38
rs1026716809 Chr.4: 107951560-107951560 on Build GRCh38
rs995703983 Chr.4: 107951557-107951557 on Build GRCh38
rs966267601 Chr.4: 107951663-107951663 on Build GRCh38
rs756575095 Chr.4: 107951866-107951866 on Build GRCh38
rs896837946 Chr.4: 107951962-107951962 on Build GRCh38
rs907405660 Chr.4: 107951498-107951498 on Build GRCh38
rs562739132 Chr.4: 107951518-107951518 on Build GRCh38
rs1036950753 Chr.4: 107951957-107951957 on Build GRCh38
rs778490219 Chr.4: 107951899-107951899 on Build GRCh38
rs1045806803 Chr.4: 107951967-107951967 on Build GRCh38
rs907356903 Chr.4: 107951970-107951970 on Build GRCh38
rs1015278778 Chr.4: 107951490-107951490 on Build GRCh38
rs1001712858 Chr.4: 107951499-107951499 on Build GRCh38
rs995587818 Chr.4: 107951582-107951582 on Build GRCh38
rs935984369 Chr.4: 107951878-107951878 on Build GRCh38
rs952342320 Chr.4: 107951607-107951607 on Build GRCh38
rs530900852 Chr.4: 107951637-107951637 on Build GRCh38
rs1044254478 Chr.4: 107951791-107951791 on Build GRCh38
rs951200839 Chr.4: 107951620-107951620 on Build GRCh38
rs896812741 Chr.4: 107951492-107951492 on Build GRCh38
rs1045873577 Chr.4: 107951493-107951493 on Build GRCh38
rs567192119 Chr.4: 107951854-107951854 on Build GRCh38
rs986588948 Chr.4: 107951630-107951630 on Build GRCh38
rs561220127 Chr.4: 107951665-107951665 on Build GRCh38
rs986330425 Chr.4: 107951642-107951642 on Build GRCh38
rs367855923 Chr.4: 107951682-107951682 on Build GRCh38
rs551151040 Chr.4: 107951571-107951571 on Build GRCh38
rs975307062 Chr.4: 107951861-107951861 on Build GRCh38
rs578150397 Chr.4: 107951715-107951715 on Build GRCh38
rs934673632 Chr.4: 107951713-107951713 on Build GRCh38

View Full Product Details