Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 108819169-108819669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL25A1
Entrez Gene ID
84570
Gene Name
collagen type XXV alpha 1
Gene Aliases
AMY, CFEOM5, CLAC, CLAC-P, CLACP
UniGene
Hs.658842
Gene Chromosome Location
Chr.4: 108808725-109302737 on Build GRCh38
Transcripts
NM_198721.3 NR_045756.2 XM_011532333.2 XM_011532334.2 XM_011532335.2 XM_011532338.2 XM_011532355.2 XM_011532356.2 XM_011532358.2 XM_017008735.1 XM_017008736.1 XM_017008737.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7197464 c.1882G>A p.G628S Chr.4: 108819293-108819293 on Build GRCh38
4881082 c.1853G>A p.R618H Chr.4: 108819322-108819322 on Build GRCh38
1426303 c.1857C>T p.G619G Chr.4: 108819318-108819318 on Build GRCh38
5594435 c.1874G>A p.G625E Chr.4: 108819301-108819301 on Build GRCh38
7362147 c.1889C>A p.P630H Chr.4: 108819286-108819286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904442482 Chr.4: 108819387-108819387 on Build GRCh38
rs750157931 Chr.4: 108819406-108819408 on Build GRCh38
rs576609210 Chr.4: 108819197-108819197 on Build GRCh38
rs774443798 Chr.4: 108819279-108819279 on Build GRCh38
rs947308629 Chr.4: 108819512-108819512 on Build GRCh38
rs188006559 Chr.4: 108819234-108819234 on Build GRCh38
rs776471790 Chr.4: 108819379-108819379 on Build GRCh38
rs954366919 Chr.4: 108819507-108819507 on Build GRCh38
rs768479643 Chr.4: 108819198-108819198 on Build GRCh38
rs77735410 Chr.4: 108819633-108819633 on Build GRCh38
rs745467559 Chr.4: 108819318-108819318 on Build GRCh38
rs200170620 Chr.4: 108819307-108819307 on Build GRCh38
rs200572202 Chr.4: 108819255-108819255 on Build GRCh38
rs767820097 Chr.4: 108819431-108819431 on Build GRCh38
rs761852364 Chr.4: 108819410-108819410 on Build GRCh38
rs766794669 Chr.4: 108819545-108819545 on Build GRCh38
rs781602978 Chr.4: 108819286-108819286 on Build GRCh38
rs563494638 Chr.4: 108819405-108819405 on Build GRCh38
rs367989402 Chr.4: 108819355-108819355 on Build GRCh38
rs1014255734 Chr.4: 108819632-108819632 on Build GRCh38
rs879056115 Chr.4: 108819455-108819455 on Build GRCh38
rs370156547 Chr.4: 108819199-108819199 on Build GRCh38
rs754158534 Chr.4: 108819551-108819551 on Build GRCh38
rs945876842 Chr.4: 108819295-108819295 on Build GRCh38
rs921611080 Chr.4: 108819495-108819495 on Build GRCh38
rs182735131 Chr.4: 108819392-108819392 on Build GRCh38
rs750738801 Chr.4: 108819243-108819243 on Build GRCh38
rs374473952 Chr.4: 108819208-108819208 on Build GRCh38
rs760047042 Chr.4: 108819566-108819566 on Build GRCh38
rs962047274 Chr.4: 108819400-108819400 on Build GRCh38
rs762032720 Chr.4: 108819218-108819218 on Build GRCh38
rs778525929 Chr.4: 108819317-108819317 on Build GRCh38
rs551331811 Chr.4: 108819618-108819618 on Build GRCh38
rs111547386 Chr.4: 108819569-108819569 on Build GRCh38
rs987216972 Chr.4: 108819525-108819525 on Build GRCh38
rs933076213 Chr.4: 108819272-108819272 on Build GRCh38
rs571186649 Chr.4: 108819627-108819627 on Build GRCh38
rs938365924 Chr.4: 108819553-108819553 on Build GRCh38
rs556699235 Chr.4: 108819193-108819193 on Build GRCh38
rs376464320 Chr.4: 108819222-108819222 on Build GRCh38
rs556786185 Chr.4: 108819634-108819634 on Build GRCh38
rs1017061906 Chr.4: 108819467-108819467 on Build GRCh38
rs548823316 Chr.4: 108819414-108819414 on Build GRCh38
rs773637035 Chr.4: 108819483-108819483 on Build GRCh38
rs768615936 Chr.4: 108819357-108819357 on Build GRCh38
rs372333956 Chr.4: 108819596-108819596 on Build GRCh38
rs745977960 Chr.4: 108819215-108819216 on Build GRCh38
rs187875371 Chr.4: 108819402-108819402 on Build GRCh38
rs1025721572 Chr.4: 108819420-108819420 on Build GRCh38
rs771718659 Chr.4: 108819322-108819322 on Build GRCh38
rs1056934253 Chr.4: 108819220-108819220 on Build GRCh38
rs190975073 Chr.4: 108819247-108819247 on Build GRCh38
rs192553199 Chr.4: 108819446-108819446 on Build GRCh38
rs149003520 Chr.4: 108819371-108819371 on Build GRCh38
rs374349920 Chr.4: 108819264-108819264 on Build GRCh38
rs924267933 Chr.4: 108819533-108819533 on Build GRCh38
rs761108752 Chr.4: 108819493-108819493 on Build GRCh38
rs772008330 Chr.4: 108819369-108819369 on Build GRCh38
rs1041687710 Chr.4: 108819532-108819532 on Build GRCh38
rs925780182 Chr.4: 108819594-108819594 on Build GRCh38
rs765683428 Chr.4: 108819567-108819567 on Build GRCh38
rs1055697153 Chr.4: 108819609-108819609 on Build GRCh38
rs752034234 Chr.4: 108819418-108819418 on Build GRCh38
rs534290264 Chr.4: 108819464-108819464 on Build GRCh38
rs779859944 Chr.4: 108819323-108819323 on Build GRCh38
rs753360764 Chr.4: 108819291-108819291 on Build GRCh38
rs766722448 Chr.4: 108819472-108819472 on Build GRCh38
rs112656770 Chr.4: 108819408-108819408 on Build GRCh38

View Full Product Details