Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109943135-109943656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGF
Entrez Gene ID
1950
Gene Name
epidermal growth factor
Gene Aliases
HOMG4, URG
UniGene
Hs.419815
Gene Chromosome Location
Chr.4: 109912883-110013079 on Build GRCh38
Transcripts
NM_001178130.2 NM_001178131.2 NM_001963.5 XM_005262796.2 XM_011531707.1 XM_017007845.1 XM_017007846.1 XM_017007847.1 XM_017007848.1 XM_017007849.1 XM_017007850.1 XM_017007851.1 XM_017007852.1 XM_017007853.1 XM_017007854.1 XM_017007855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652153 c.379A>C p.N127H Chr.4: 109943305-109943305 on Build GRCh38
343039 c.430G>T p.D144Y Chr.4: 109943356-109943356 on Build GRCh38
6652154 c.370A>C p.N124H Chr.4: 109943296-109943296 on Build GRCh38
1050010 c.345G>T p.E115D Chr.4: 109943271-109943271 on Build GRCh38
3916953 c.440G>A p.G147E Chr.4: 109943366-109943366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758375964 Chr.4: 109943317-109943317 on Build GRCh38
rs765143879 Chr.4: 109943269-109943269 on Build GRCh38
rs559242261 Chr.4: 109943173-109943173 on Build GRCh38
rs746617418 Chr.4: 109943383-109943384 on Build GRCh38
rs150361543 Chr.4: 109943390-109943390 on Build GRCh38
rs988319915 Chr.4: 109943217-109943217 on Build GRCh38
rs763832012 Chr.4: 109943255-109943255 on Build GRCh38
rs955222384 Chr.4: 109943211-109943211 on Build GRCh38
rs1056649870 Chr.4: 109943380-109943380 on Build GRCh38
rs772979123 Chr.4: 109943398-109943398 on Build GRCh38
rs755406765 Chr.4: 109943357-109943357 on Build GRCh38
rs942849036 Chr.4: 109943182-109943182 on Build GRCh38
rs765257185 Chr.4: 109943395-109943395 on Build GRCh38
rs926881361 Chr.4: 109943276-109943276 on Build GRCh38
rs372078351 Chr.4: 109943475-109943475 on Build GRCh38
rs775481271 Chr.4: 109943308-109943310 on Build GRCh38
rs763112495 Chr.4: 109943402-109943402 on Build GRCh38
rs922894532 Chr.4: 109943267-109943267 on Build GRCh38
rs138035348 Chr.4: 109943391-109943391 on Build GRCh38
rs761571360 Chr.4: 109943393-109943393 on Build GRCh38
rs766651293 Chr.4: 109943342-109943342 on Build GRCh38
rs911433301 Chr.4: 109943176-109943176 on Build GRCh38
rs746520838 Chr.4: 109943483-109943483 on Build GRCh38
rs994472084 Chr.4: 109943559-109943560 on Build GRCh38
rs997137966 Chr.4: 109943581-109943581 on Build GRCh38
rs758427373 Chr.4: 109943624-109943624 on Build GRCh38
rs771984189 Chr.4: 109943224-109943226 on Build GRCh38
rs1006233916 Chr.4: 109943563-109943563 on Build GRCh38
rs555212460 Chr.4: 109943259-109943259 on Build GRCh38
rs987984687 Chr.4: 109943601-109943601 on Build GRCh38
rs1043341662 Chr.4: 109943268-109943268 on Build GRCh38
rs1025671554 Chr.4: 109943559-109943559 on Build GRCh38
rs199526360 Chr.4: 109943379-109943379 on Build GRCh38
rs749713346 Chr.4: 109943469-109943469 on Build GRCh38
rs1001381702 Chr.4: 109943407-109943407 on Build GRCh38
rs756517950 Chr.4: 109943461-109943461 on Build GRCh38
rs566242327 Chr.4: 109943532-109943532 on Build GRCh38
rs528285663 Chr.4: 109943192-109943192 on Build GRCh38
rs544673643 Chr.4: 109943214-109943214 on Build GRCh38
rs11568899 Chr.4: 109943589-109943589 on Build GRCh38
rs755316861 Chr.4: 109943431-109943431 on Build GRCh38
rs550344691 Chr.4: 109943369-109943369 on Build GRCh38
rs9991664 Chr.4: 109943377-109943377 on Build GRCh38
rs778121711 Chr.4: 109943375-109943375 on Build GRCh38
rs771807712 Chr.4: 109943204-109943204 on Build GRCh38
rs1048316110 Chr.4: 109943456-109943456 on Build GRCh38
rs778375711 Chr.4: 109943466-109943466 on Build GRCh38
rs534546146 Chr.4: 109943360-109943360 on Build GRCh38
rs202179418 Chr.4: 109943225-109943225 on Build GRCh38
rs558262413 Chr.4: 109943600-109943600 on Build GRCh38
rs764873696 Chr.4: 109943488-109943488 on Build GRCh38
rs530407670 Chr.4: 109943353-109943353 on Build GRCh38
rs766200265 Chr.4: 109943474-109943474 on Build GRCh38
rs767836927 Chr.4: 109943459-109943459 on Build GRCh38
rs952688852 Chr.4: 109943561-109943561 on Build GRCh38
rs899768598 Chr.4: 109943580-109943580 on Build GRCh38
rs759070960 Chr.4: 109943460-109943460 on Build GRCh38
rs751835222 Chr.4: 109943412-109943412 on Build GRCh38
rs779367241 Chr.4: 109943482-109943482 on Build GRCh38
rs775234641 Chr.4: 109943386-109943386 on Build GRCh38
rs946419697 Chr.4: 109943263-109943263 on Build GRCh38
rs960001956 Chr.4: 109943515-109943515 on Build GRCh38
rs750562434 Chr.4: 109943272-109943272 on Build GRCh38
rs11568898 Chr.4: 109943567-109943567 on Build GRCh38
rs117552261 Chr.4: 109943440-109943440 on Build GRCh38
rs766311143 Chr.4: 109943408-109943408 on Build GRCh38
rs143595598 Chr.4: 109943410-109943410 on Build GRCh38
rs148281400 Chr.4: 109943374-109943374 on Build GRCh38
rs144013010 Chr.4: 109943404-109943404 on Build GRCh38
rs775967170 Chr.4: 109943482-109943482 on Build GRCh38
rs761675450 Chr.4: 109943264-109943264 on Build GRCh38

View Full Product Details