Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109944949-109945348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGF
Entrez Gene ID
1950
Gene Name
epidermal growth factor
Gene Aliases
HOMG4, URG
UniGene
Hs.419815
Gene Chromosome Location
Chr.4: 109912883-110013079 on Build GRCh38
Transcripts
NM_001178130.2 NM_001178131.2 NM_001963.5 XM_005262796.2 XM_011531707.1 XM_017007845.1 XM_017007846.1 XM_017007847.1 XM_017007848.1 XM_017007849.1 XM_017007850.1 XM_017007851.1 XM_017007852.1 XM_017007853.1 XM_017007854.1 XM_017007855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4886717 c.819C>G p.N273K Chr.4: 109945154-109945154 on Build GRCh38
6402833 c.940G>A p.E314K Chr.4: 109945275-109945275 on Build GRCh38
6652148 c.746T>C p.L249S Chr.4: 109945081-109945081 on Build GRCh38
6392367 c.765T>A p.F255L Chr.4: 109945100-109945100 on Build GRCh38
3824948 c.785C>G p.S262* Chr.4: 109945120-109945120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763602539 Chr.4: 109945224-109945224 on Build GRCh38
rs975174696 Chr.4: 109945057-109945057 on Build GRCh38
rs780021823 Chr.4: 109945174-109945174 on Build GRCh38
rs140068596 Chr.4: 109945259-109945259 on Build GRCh38
rs747087585 Chr.4: 109945176-109945176 on Build GRCh38
rs758470342 Chr.4: 109945173-109945173 on Build GRCh38
rs146585233 Chr.4: 109945226-109945226 on Build GRCh38
rs955232184 Chr.4: 109945108-109945108 on Build GRCh38
rs185882668 Chr.4: 109945006-109945006 on Build GRCh38
rs11568911 Chr.4: 109945104-109945104 on Build GRCh38
rs756341864 Chr.4: 109945072-109945072 on Build GRCh38
rs920332542 Chr.4: 109945300-109945300 on Build GRCh38
rs760127815 Chr.4: 109945222-109945222 on Build GRCh38
rs373341220 Chr.4: 109945172-109945172 on Build GRCh38
rs757131048 Chr.4: 109945107-109945107 on Build GRCh38
rs890665967 Chr.4: 109945111-109945111 on Build GRCh38
rs11568912 Chr.4: 109945297-109945297 on Build GRCh38
rs773710034 Chr.4: 109945067-109945067 on Build GRCh38
rs758238904 Chr.4: 109945261-109945261 on Build GRCh38
rs4698755 Chr.4: 109945022-109945022 on Build GRCh38
rs867950317 Chr.4: 109945126-109945126 on Build GRCh38
rs746904541 Chr.4: 109945281-109945281 on Build GRCh38
rs565605864 Chr.4: 109945042-109945042 on Build GRCh38
rs370617801 Chr.4: 109945062-109945062 on Build GRCh38
rs765524556 Chr.4: 109945065-109945065 on Build GRCh38
rs776728511 Chr.4: 109945192-109945192 on Build GRCh38
rs774883058 Chr.4: 109945216-109945216 on Build GRCh38
rs149782177 Chr.4: 109945262-109945262 on Build GRCh38
rs778787208 Chr.4: 109945170-109945170 on Build GRCh38
rs748331617 Chr.4: 109945193-109945193 on Build GRCh38
rs763686145 Chr.4: 109945096-109945096 on Build GRCh38
rs769999140 Chr.4: 109945196-109945196 on Build GRCh38
rs4698756 Chr.4: 109945286-109945286 on Build GRCh38
rs1009067759 Chr.4: 109945128-109945128 on Build GRCh38
rs191577694 Chr.4: 109945292-109945292 on Build GRCh38
rs748211601 Chr.4: 109945287-109945287 on Build GRCh38
rs913827959 Chr.4: 109945165-109945165 on Build GRCh38
rs139711858 Chr.4: 109945201-109945201 on Build GRCh38
rs757041257 Chr.4: 109945244-109945244 on Build GRCh38
rs776871101 Chr.4: 109945043-109945043 on Build GRCh38
rs754573805 Chr.4: 109945308-109945308 on Build GRCh38
rs370244204 Chr.4: 109945285-109945285 on Build GRCh38
rs752195298 Chr.4: 109945089-109945089 on Build GRCh38
rs747101578 Chr.4: 109945030-109945030 on Build GRCh38
rs771238363 Chr.4: 109945299-109945299 on Build GRCh38
rs761412607 Chr.4: 109945030-109945031 on Build GRCh38
rs764972836 Chr.4: 109945257-109945257 on Build GRCh38
rs780307505 Chr.4: 109945091-109945091 on Build GRCh38
rs763517791 Chr.4: 109945071-109945071 on Build GRCh38
rs766856972 Chr.4: 109945075-109945075 on Build GRCh38
rs755122917 Chr.4: 109945296-109945296 on Build GRCh38
rs996244030 Chr.4: 109945058-109945058 on Build GRCh38
rs940394433 Chr.4: 109945016-109945016 on Build GRCh38
rs967447422 Chr.4: 109945213-109945213 on Build GRCh38
rs774736161 Chr.4: 109945320-109945320 on Build GRCh38
rs376262854 Chr.4: 109945212-109945212 on Build GRCh38
rs35191533 Chr.4: 109945210-109945210 on Build GRCh38
rs768730588 Chr.4: 109945177-109945177 on Build GRCh38

View Full Product Details