Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109963170-109963718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGF
Entrez Gene ID
1950
Gene Name
epidermal growth factor
Gene Aliases
HOMG4, URG
UniGene
Hs.419815
Gene Chromosome Location
Chr.4: 109912883-110013079 on Build GRCh38
Transcripts
NM_001178130.2 NM_001178131.2 NM_001963.5 XM_005262796.2 XM_011531707.1 XM_017007845.1 XM_017007846.1 XM_017007847.1 XM_017007848.1 XM_017007849.1 XM_017007850.1 XM_017007851.1 XM_017007852.1 XM_017007853.1 XM_017007854.1 XM_017007855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3006366 c.1418delA p.S475fs*43 Chr.4: 109963278-109963278 on Build GRCh38
4893327 c.1417G>A p.E473K Chr.4: 109963277-109963277 on Build GRCh38
5865568 c.1357C>T p.P453S Chr.4: 109963217-109963217 on Build GRCh38
5865569 c.1413_1414GG>AA p.D472N Chr.4: 109963273-109963274 on Build GRCh38
6056322 c.1326C>T p.P442P Chr.4: 109963186-109963186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11568953 Chr.4: 109963240-109963240 on Build GRCh38
rs1033308841 Chr.4: 109963198-109963198 on Build GRCh38
rs905991842 Chr.4: 109963468-109963468 on Build GRCh38
rs922651469 Chr.4: 109963338-109963338 on Build GRCh38
rs988161626 Chr.4: 109963341-109963341 on Build GRCh38
rs964205080 Chr.4: 109963280-109963280 on Build GRCh38
rs775591256 Chr.4: 109963349-109963349 on Build GRCh38
rs370511854 Chr.4: 109963346-109963346 on Build GRCh38
rs745921822 Chr.4: 109963201-109963201 on Build GRCh38
rs955468090 Chr.4: 109963605-109963605 on Build GRCh38
rs758455864 Chr.4: 109963333-109963333 on Build GRCh38
rs564693339 Chr.4: 109963507-109963507 on Build GRCh38
rs1021410971 Chr.4: 109963670-109963670 on Build GRCh38
rs142751045 Chr.4: 109963214-109963214 on Build GRCh38
rs913889158 Chr.4: 109963347-109963347 on Build GRCh38
rs528376562 Chr.4: 109963374-109963374 on Build GRCh38
rs192894154 Chr.4: 109963368-109963368 on Build GRCh38
rs964689729 Chr.4: 109963681-109963681 on Build GRCh38
rs11568954 Chr.4: 109963332-109963333 on Build GRCh38
rs542925186 Chr.4: 109963529-109963529 on Build GRCh38
rs556603989 Chr.4: 109963342-109963342 on Build GRCh38
rs542559958 Chr.4: 109963387-109963387 on Build GRCh38
rs984682205 Chr.4: 109963610-109963610 on Build GRCh38
rs900012640 Chr.4: 109963563-109963563 on Build GRCh38
rs1013187520 Chr.4: 109963535-109963535 on Build GRCh38
rs1017152201 Chr.4: 109963534-109963534 on Build GRCh38
rs1006218646 Chr.4: 109963522-109963522 on Build GRCh38
rs11568957 Chr.4: 109963612-109963612 on Build GRCh38
rs11568956 Chr.4: 109963589-109963589 on Build GRCh38
rs149166157 Chr.4: 109963472-109963472 on Build GRCh38
rs779196084 Chr.4: 109963213-109963214 on Build GRCh38
rs777131517 Chr.4: 109963229-109963229 on Build GRCh38
rs542126113 Chr.4: 109963372-109963372 on Build GRCh38
rs886776171 Chr.4: 109963514-109963514 on Build GRCh38
rs112168633 Chr.4: 109963223-109963223 on Build GRCh38
rs755866001 Chr.4: 109963293-109963293 on Build GRCh38
rs746855652 Chr.4: 109963345-109963345 on Build GRCh38
rs185353207 Chr.4: 109963508-109963508 on Build GRCh38
rs745635681 Chr.4: 109963636-109963637 on Build GRCh38
rs190053627 Chr.4: 109963587-109963587 on Build GRCh38
rs895594167 Chr.4: 109963474-109963474 on Build GRCh38
rs779967710 Chr.4: 109963340-109963341 on Build GRCh38
rs777776376 Chr.4: 109963335-109963335 on Build GRCh38
rs747624637 Chr.4: 109963333-109963333 on Build GRCh38
rs149448320 Chr.4: 109963276-109963276 on Build GRCh38
rs754468394 Chr.4: 109963249-109963249 on Build GRCh38
rs11568955 Chr.4: 109963401-109963401 on Build GRCh38
rs749082116 Chr.4: 109963339-109963339 on Build GRCh38
rs528025061 Chr.4: 109963471-109963471 on Build GRCh38
rs377308920 Chr.4: 109963574-109963574 on Build GRCh38
rs201101453 Chr.4: 109963264-109963264 on Build GRCh38
rs41434845 Chr.4: 109963340-109963340 on Build GRCh38
rs975502951 Chr.4: 109963299-109963299 on Build GRCh38
rs762377419 Chr.4: 109963230-109963230 on Build GRCh38
rs991882907 Chr.4: 109963224-109963224 on Build GRCh38
rs1043613743 Chr.4: 109963405-109963405 on Build GRCh38
rs765706561 Chr.4: 109963232-109963232 on Build GRCh38
rs150311153 Chr.4: 109963227-109963227 on Build GRCh38
rs527489767 Chr.4: 109963682-109963682 on Build GRCh38

View Full Product Details