Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 113174331-113174843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANK2
Entrez Gene ID
287
Gene Name
ankyrin 2, neuronal
Gene Aliases
ANK-2, LQT4, brank-2
UniGene
Hs.620557
Gene Chromosome Location
Chr.4: 112706083-113383740 on Build GRCh38
Transcripts
NM_001127493.1 NM_001148.4 NM_020977.3 XM_005262945.2 XM_017008067.1 XM_017008068.1 XM_017008069.1 XM_017008070.1 XM_017008071.1 XM_017008072.1 XM_017008073.1 XM_017008074.1 XM_017008075.1 XM_017008076.1 XM_017008077.1 XM_017008078.1 XM_017008079.1 XM_017008080.1 XM_017008081.1 XM_017008082.1 XM_017008083.1 XM_017008084.1 XM_017008085.1 XM_017008086.1 XM_017008087.1 XM_017008088.1 XM_017008089.1 XM_017008090.1 XM_017008091.1 XM_017008092.1 XM_017008093.1 XM_017008094.1 XM_017008095.1 XM_017008096.1 XM_017008097.1 XM_017008098.1 XM_017008099.1 XM_017008100.1 XM_017008101.1 XM_017008102.1 XM_017008103.1 XM_017008104.1 XM_017008105.1 XM_017008106.1 XM_017008107.1 XM_017008108.1 XM_017008109.1 XM_017008110.1 XM_017008111.1 XM_017008112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3781906 c.22-5C>T p.? Chr.4: 113174411-113174411 on Build GRCh38
7253882 c.93G>T p.K31N Chr.4: 113174487-113174487 on Build GRCh38
7253881 c.156G>T p.K52N Chr.4: 113174487-113174487 on Build GRCh38
4661556 c.128A>G p.N43S Chr.4: 113174459-113174459 on Build GRCh38
6050485 c.108C>A p.I36I Chr.4: 113174502-113174502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111908922 Chr.4: 113174552-113174552 on Build GRCh38
rs138293707 Chr.4: 113174595-113174595 on Build GRCh38
rs749635039 Chr.4: 113174493-113174493 on Build GRCh38
rs371475487 Chr.4: 113174396-113174396 on Build GRCh38
rs1014640205 Chr.4: 113174690-113174690 on Build GRCh38
rs28725346 Chr.4: 113174682-113174682 on Build GRCh38
rs763460244 Chr.4: 113174403-113174403 on Build GRCh38
rs766679849 Chr.4: 113174387-113174387 on Build GRCh38
rs150688564 Chr.4: 113174731-113174731 on Build GRCh38
rs554150200 Chr.4: 113174654-113174654 on Build GRCh38
rs896235867 Chr.4: 113174697-113174697 on Build GRCh38
rs937199337 Chr.4: 113174655-113174655 on Build GRCh38
rs750209101 Chr.4: 113174421-113174421 on Build GRCh38
rs776004030 Chr.4: 113174539-113174539 on Build GRCh38
rs786205718 Chr.4: 113174507-113174507 on Build GRCh38
rs561002554 Chr.4: 113174754-113174754 on Build GRCh38
rs879244459 Chr.4: 113174490-113174490 on Build GRCh38
rs781510145 Chr.4: 113174366-113174366 on Build GRCh38
rs991278014 Chr.4: 113174673-113174673 on Build GRCh38
rs769893243 Chr.4: 113174489-113174489 on Build GRCh38
rs117768632 Chr.4: 113174559-113174559 on Build GRCh38
rs150986299 Chr.4: 113174463-113174463 on Build GRCh38
rs139910370 Chr.4: 113174772-113174772 on Build GRCh38
rs748468378 Chr.4: 113174483-113174483 on Build GRCh38
rs781246712 Chr.4: 113174688-113174688 on Build GRCh38
rs769178589 Chr.4: 113174741-113174741 on Build GRCh38
rs372560419 Chr.4: 113174411-113174411 on Build GRCh38
rs369879683 Chr.4: 113174553-113174553 on Build GRCh38
rs555003393 Chr.4: 113174471-113174471 on Build GRCh38
rs1032598276 Chr.4: 113174773-113174773 on Build GRCh38
rs757292523 Chr.4: 113174630-113174630 on Build GRCh38
rs780966324 Chr.4: 113174601-113174601 on Build GRCh38
rs758379675 Chr.4: 113174427-113174427 on Build GRCh38
rs765150752 Chr.4: 113174418-113174418 on Build GRCh38
rs770197287 Chr.4: 113174378-113174378 on Build GRCh38
rs1049003531 Chr.4: 113174642-113174642 on Build GRCh38
rs370825390 Chr.4: 113174369-113174369 on Build GRCh38
rs969646633 Chr.4: 113174417-113174417 on Build GRCh38
rs545319394 Chr.4: 113174639-113174639 on Build GRCh38
rs899869365 Chr.4: 113174634-113174634 on Build GRCh38
rs755162060 Chr.4: 113174549-113174549 on Build GRCh38
rs771378991 Chr.4: 113174504-113174504 on Build GRCh38
rs956862168 Chr.4: 113174818-113174818 on Build GRCh38
rs145272651 Chr.4: 113174461-113174461 on Build GRCh38
rs377612257 Chr.4: 113174522-113174522 on Build GRCh38
rs369019700 Chr.4: 113174397-113174397 on Build GRCh38
rs768219534 Chr.4: 113174533-113174533 on Build GRCh38
rs866875104 Chr.4: 113174433-113174433 on Build GRCh38
rs1017008834 Chr.4: 113174510-113174510 on Build GRCh38
rs1047496087 Chr.4: 113174465-113174465 on Build GRCh38
rs775017092 Chr.4: 113174505-113174505 on Build GRCh38
rs969070662 Chr.4: 113174744-113174744 on Build GRCh38
rs878944908 Chr.4: 113174361-113174361 on Build GRCh38
rs369877280 Chr.4: 113174440-113174440 on Build GRCh38
rs753544572 Chr.4: 113174404-113174404 on Build GRCh38
rs997006405 Chr.4: 113174544-113174544 on Build GRCh38
rs377576219 Chr.4: 113174412-113174412 on Build GRCh38
rs558666470 Chr.4: 113174589-113174589 on Build GRCh38
rs375688098 Chr.4: 113174371-113174371 on Build GRCh38
rs548690392 Chr.4: 113174617-113174617 on Build GRCh38
rs960362147 Chr.4: 113174358-113174358 on Build GRCh38
rs201304241 Chr.4: 113174642-113174642 on Build GRCh38
rs887127609 Chr.4: 113174745-113174745 on Build GRCh38

View Full Product Details