Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 121678293-121678795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA5
Entrez Gene ID
308
Gene Name
annexin A5
Gene Aliases
ANX5, ENX2, HEL-S-7, PP4, RPRGL3
UniGene
Hs.480653
Gene Chromosome Location
Chr.4: 121667997-121696992 on Build GRCh38
Transcripts
NM_001154.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6873215 c.395-1G>A p.? Chr.4: 121678495-121678495 on Build GRCh38
3008372 c.421G>A p.V141M Chr.4: 121678468-121678468 on Build GRCh38
5915266 c.400G>T p.G134C Chr.4: 121678489-121678489 on Build GRCh38
3008370 c.454A>G p.M152V Chr.4: 121678435-121678435 on Build GRCh38
1426627 c.451C>T p.R151W Chr.4: 121678438-121678438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776789298 Chr.4: 121678361-121678361 on Build GRCh38
rs935407486 Chr.4: 121678575-121678575 on Build GRCh38
rs886830394 Chr.4: 121678452-121678452 on Build GRCh38
rs934838426 Chr.4: 121678577-121678577 on Build GRCh38
rs774548386 Chr.4: 121678400-121678400 on Build GRCh38
rs570249127 Chr.4: 121678460-121678460 on Build GRCh38
rs912362974 Chr.4: 121678587-121678587 on Build GRCh38
rs968435196 Chr.4: 121678533-121678533 on Build GRCh38
rs780076315 Chr.4: 121678438-121678438 on Build GRCh38
rs750003688 Chr.4: 121678412-121678412 on Build GRCh38
rs112191309 Chr.4: 121678664-121678664 on Build GRCh38
rs970418444 Chr.4: 121678497-121678497 on Build GRCh38
rs539231447 Chr.4: 121678729-121678729 on Build GRCh38
rs761349302 Chr.4: 121678502-121678502 on Build GRCh38
rs374246149 Chr.4: 121678407-121678407 on Build GRCh38
rs933317139 Chr.4: 121678703-121678703 on Build GRCh38
rs77516666 Chr.4: 121678386-121678386 on Build GRCh38
rs760151688 Chr.4: 121678473-121678473 on Build GRCh38
rs764592261 Chr.4: 121678411-121678411 on Build GRCh38
rs979485608 Chr.4: 121678570-121678570 on Build GRCh38
rs754732080 Chr.4: 121678454-121678454 on Build GRCh38
rs924876606 Chr.4: 121678553-121678553 on Build GRCh38
rs749865660 Chr.4: 121678517-121678517 on Build GRCh38
rs755758865 Chr.4: 121678745-121678745 on Build GRCh38
rs567626342 Chr.4: 121678482-121678482 on Build GRCh38
rs536268036 Chr.4: 121678579-121678579 on Build GRCh38
rs142008590 Chr.4: 121678457-121678457 on Build GRCh38
rs982126449 Chr.4: 121678508-121678508 on Build GRCh38
rs754079941 Chr.4: 121678367-121678367 on Build GRCh38
rs565836612 Chr.4: 121678682-121678682 on Build GRCh38
rs765842815 Chr.4: 121678535-121678535 on Build GRCh38
rs886294267 Chr.4: 121678456-121678456 on Build GRCh38
rs200076201 Chr.4: 121678483-121678483 on Build GRCh38
rs765003645 Chr.4: 121678485-121678485 on Build GRCh38
rs757880125 Chr.4: 121678525-121678525 on Build GRCh38
rs563255553 Chr.4: 121678340-121678340 on Build GRCh38
rs760027157 Chr.4: 121678403-121678403 on Build GRCh38
rs202083195 Chr.4: 121678469-121678469 on Build GRCh38
rs571398297 Chr.4: 121678408-121678408 on Build GRCh38
rs1039128905 Chr.4: 121678601-121678601 on Build GRCh38
rs3733649 Chr.4: 121678600-121678600 on Build GRCh38
rs763820845 Chr.4: 121678425-121678427 on Build GRCh38
rs368943069 Chr.4: 121678449-121678449 on Build GRCh38
rs146310531 Chr.4: 121678468-121678468 on Build GRCh38
rs552415548 Chr.4: 121678677-121678677 on Build GRCh38
rs752261506 Chr.4: 121678716-121678716 on Build GRCh38
rs923951554 Chr.4: 121678574-121678574 on Build GRCh38
rs746976066 Chr.4: 121678448-121678448 on Build GRCh38
rs763553599 Chr.4: 121678372-121678372 on Build GRCh38
rs763754425 Chr.4: 121678596-121678596 on Build GRCh38
rs901878985 Chr.4: 121678667-121678667 on Build GRCh38
rs537655077 Chr.4: 121678747-121678747 on Build GRCh38
rs368764657 Chr.4: 121678459-121678459 on Build GRCh38
rs753580828 Chr.4: 121678757-121678757 on Build GRCh38
rs1048135670 Chr.4: 121678713-121678713 on Build GRCh38
rs375266096 Chr.4: 121678540-121678540 on Build GRCh38
rs758259826 Chr.4: 121678437-121678437 on Build GRCh38
rs143768472 Chr.4: 121678599-121678599 on Build GRCh38
rs1035472271 Chr.4: 121678730-121678730 on Build GRCh38
rs748313765 Chr.4: 121678458-121678458 on Build GRCh38
rs1046701956 Chr.4: 121678668-121678668 on Build GRCh38
rs373860357 Chr.4: 121678371-121678371 on Build GRCh38
rs372194066 Chr.4: 121678481-121678481 on Build GRCh38
rs186936197 Chr.4: 121678378-121678378 on Build GRCh38
rs753458508 Chr.4: 121678458-121678459 on Build GRCh38

View Full Product Details