Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 121684387-121684884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA5
Entrez Gene ID
308
Gene Name
annexin A5
Gene Aliases
ANX5, ENX2, HEL-S-7, PP4, RPRGL3
UniGene
Hs.480653
Gene Chromosome Location
Chr.4: 121667997-121696992 on Build GRCh38
Transcripts
NM_001154.3 XM_017008141.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5698878 c.189+1G>T p.? Chr.4: 121684676-121684676 on Build GRCh38
5004391 c.132C>T p.S44S Chr.4: 121684734-121684734 on Build GRCh38
4662161 c.148C>T p.R50C Chr.4: 121684718-121684718 on Build GRCh38
4851971 c.133C>T p.R45* Chr.4: 121684733-121684733 on Build GRCh38
6166131 c.163G>T p.A55S Chr.4: 121684703-121684703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768613921 Chr.4: 121684742-121684742 on Build GRCh38
rs765143178 Chr.4: 121684652-121684652 on Build GRCh38
rs370104706 Chr.4: 121684691-121684691 on Build GRCh38
rs569010676 Chr.4: 121684419-121684419 on Build GRCh38
rs185204121 Chr.4: 121684443-121684443 on Build GRCh38
rs766552765 Chr.4: 121684786-121684786 on Build GRCh38
rs754964566 Chr.4: 121684800-121684800 on Build GRCh38
rs894472681 Chr.4: 121684602-121684602 on Build GRCh38
rs764003339 Chr.4: 121684626-121684626 on Build GRCh38
rs965275217 Chr.4: 121684490-121684490 on Build GRCh38
rs983415166 Chr.4: 121684519-121684519 on Build GRCh38
rs758001979 Chr.4: 121684744-121684745 on Build GRCh38
rs201890674 Chr.4: 121684759-121684759 on Build GRCh38
rs957313205 Chr.4: 121684748-121684748 on Build GRCh38
rs78275252 Chr.4: 121684585-121684585 on Build GRCh38
rs767669099 Chr.4: 121684807-121684807 on Build GRCh38
rs548422052 Chr.4: 121684449-121684449 on Build GRCh38
rs543247319 Chr.4: 121684743-121684743 on Build GRCh38
rs765069940 Chr.4: 121684752-121684752 on Build GRCh38
rs878992406 Chr.4: 121684849-121684849 on Build GRCh38
rs1034796343 Chr.4: 121684589-121684589 on Build GRCh38
rs1014608162 Chr.4: 121684612-121684612 on Build GRCh38
rs571830605 Chr.4: 121684501-121684501 on Build GRCh38
rs73844252 Chr.4: 121684568-121684568 on Build GRCh38
rs927975209 Chr.4: 121684560-121684560 on Build GRCh38
rs751659232 Chr.4: 121684669-121684669 on Build GRCh38
rs991343363 Chr.4: 121684581-121684581 on Build GRCh38
rs766470932 Chr.4: 121684664-121684664 on Build GRCh38
rs746027549 Chr.4: 121684809-121684809 on Build GRCh38
rs761869455 Chr.4: 121684747-121684747 on Build GRCh38
rs376942269 Chr.4: 121684686-121684686 on Build GRCh38
rs950686865 Chr.4: 121684513-121684514 on Build GRCh38
rs878923880 Chr.4: 121684853-121684853 on Build GRCh38
rs761746776 Chr.4: 121684650-121684650 on Build GRCh38
rs778094878 Chr.4: 121684820-121684820 on Build GRCh38
rs1007068620 Chr.4: 121684453-121684453 on Build GRCh38
rs749720098 Chr.4: 121684822-121684822 on Build GRCh38
rs950904156 Chr.4: 121684795-121684795 on Build GRCh38
rs758712188 Chr.4: 121684662-121684662 on Build GRCh38
rs751637939 Chr.4: 121684790-121684790 on Build GRCh38
rs763265975 Chr.4: 121684775-121684775 on Build GRCh38
rs1050475048 Chr.4: 121684436-121684436 on Build GRCh38
rs574246361 Chr.4: 121684718-121684718 on Build GRCh38
rs923826869 Chr.4: 121684648-121684648 on Build GRCh38
rs770983977 Chr.4: 121684687-121684688 on Build GRCh38
rs1043648671 Chr.4: 121684415-121684417 on Build GRCh38
rs750610018 Chr.4: 121684659-121684659 on Build GRCh38
rs1017742920 Chr.4: 121684483-121684483 on Build GRCh38
rs994270024 Chr.4: 121684514-121684514 on Build GRCh38
rs771767958 Chr.4: 121684720-121684720 on Build GRCh38
rs773203351 Chr.4: 121684773-121684773 on Build GRCh38
rs749475839 Chr.4: 121684662-121684664 on Build GRCh38
rs1027945105 Chr.4: 121684504-121684504 on Build GRCh38
rs756625909 Chr.4: 121684810-121684810 on Build GRCh38
rs901378898 Chr.4: 121684508-121684508 on Build GRCh38
rs1039882471 Chr.4: 121684717-121684717 on Build GRCh38
rs778015840 Chr.4: 121684652-121684652 on Build GRCh38
rs753136616 Chr.4: 121684809-121684809 on Build GRCh38
rs935671246 Chr.4: 121684429-121684429 on Build GRCh38
rs888494063 Chr.4: 121684578-121684578 on Build GRCh38
rs776452376 Chr.4: 121684637-121684637 on Build GRCh38
rs756431689 Chr.4: 121684706-121684706 on Build GRCh38
rs759419433 Chr.4: 121684733-121684733 on Build GRCh38
rs142171370 Chr.4: 121684716-121684716 on Build GRCh38
rs1050759371 Chr.4: 121684681-121684681 on Build GRCh38
rs17851664 Chr.4: 121684707-121684707 on Build GRCh38
rs774223040 Chr.4: 121684724-121684724 on Build GRCh38

View Full Product Details