Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122407722-122408241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAD1
Entrez Gene ID
132612
Gene Name
adenosine deaminase domain containing 1
Gene Aliases
Tenr
UniGene
Hs.518957
Gene Chromosome Location
Chr.4: 122378966-122429815 on Build GRCh38
Transcripts
NM_001159285.1 NM_001159295.1 NM_139243.3 XM_005262741.2 XM_005262743.2 XM_005262744.2 XM_005262745.2 XM_011531603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5989440 c.807T>A p.H269Q Chr.4: 122407990-122407990 on Build GRCh38
6469266 c.844C>T p.L282F Chr.4: 122408027-122408027 on Build GRCh38
5400407 c.794G>A p.G265E Chr.4: 122407977-122407977 on Build GRCh38
5665218 c.848+1G>T p.? Chr.4: 122408032-122408032 on Build GRCh38
5400406 c.769T>G p.Y257D Chr.4: 122407952-122407952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762546046 Chr.4: 122407992-122407992 on Build GRCh38
rs111326362 Chr.4: 122407924-122407924 on Build GRCh38
rs902042694 Chr.4: 122407966-122407966 on Build GRCh38
rs766397786 Chr.4: 122407999-122407999 on Build GRCh38
rs368185298 Chr.4: 122408076-122408076 on Build GRCh38
rs753987408 Chr.4: 122408055-122408055 on Build GRCh38
rs17388568 Chr.4: 122408207-122408207 on Build GRCh38
rs766090215 Chr.4: 122407865-122407865 on Build GRCh38
rs201227638 Chr.4: 122407975-122407975 on Build GRCh38
rs746305813 Chr.4: 122407931-122407931 on Build GRCh38
rs138644891 Chr.4: 122408027-122408027 on Build GRCh38
rs1054089234 Chr.4: 122407936-122407936 on Build GRCh38
rs759412069 Chr.4: 122408030-122408030 on Build GRCh38
rs368577732 Chr.4: 122407943-122407943 on Build GRCh38
rs200278158 Chr.4: 122408006-122408006 on Build GRCh38
rs181883965 Chr.4: 122408170-122408170 on Build GRCh38
rs757275029 Chr.4: 122408066-122408066 on Build GRCh38
rs528208894 Chr.4: 122407749-122407749 on Build GRCh38
rs938072416 Chr.4: 122407826-122407826 on Build GRCh38
rs534828477 Chr.4: 122408181-122408181 on Build GRCh38
rs902743801 Chr.4: 122407928-122407928 on Build GRCh38
rs529159036 Chr.4: 122407815-122407816 on Build GRCh38
rs756268316 Chr.4: 122408046-122408046 on Build GRCh38
rs771363390 Chr.4: 122407921-122407921 on Build GRCh38
rs1005186681 Chr.4: 122408149-122408149 on Build GRCh38
rs748197463 Chr.4: 122407757-122407757 on Build GRCh38
rs372034161 Chr.4: 122407986-122407986 on Build GRCh38
rs772077913 Chr.4: 122408211-122408211 on Build GRCh38
rs777997061 Chr.4: 122408164-122408164 on Build GRCh38
rs984731609 Chr.4: 122407793-122407793 on Build GRCh38
rs752542794 Chr.4: 122408099-122408099 on Build GRCh38
rs574555756 Chr.4: 122408216-122408216 on Build GRCh38
rs748283365 Chr.4: 122407871-122407871 on Build GRCh38
rs949572643 Chr.4: 122407880-122407880 on Build GRCh38
rs536908452 Chr.4: 122407870-122407870 on Build GRCh38
rs371541360 Chr.4: 122407917-122407917 on Build GRCh38
rs754714576 Chr.4: 122407867-122407867 on Build GRCh38
rs997274596 Chr.4: 122408061-122408061 on Build GRCh38
rs1017752571 Chr.4: 122408047-122408047 on Build GRCh38
rs779287503 Chr.4: 122407923-122407923 on Build GRCh38
rs754392703 Chr.4: 122408011-122408011 on Build GRCh38
rs779526572 Chr.4: 122408068-122408068 on Build GRCh38
rs950648924 Chr.4: 122407756-122407756 on Build GRCh38
rs559025293 Chr.4: 122408163-122408163 on Build GRCh38
rs892785778 Chr.4: 122407997-122407997 on Build GRCh38
rs756277581 Chr.4: 122408204-122408204 on Build GRCh38
rs767173018 Chr.4: 122408040-122408040 on Build GRCh38
rs374635047 Chr.4: 122407800-122407800 on Build GRCh38
rs1026174889 Chr.4: 122407748-122407748 on Build GRCh38
rs570408145 Chr.4: 122408044-122408044 on Build GRCh38
rs764469971 Chr.4: 122407891-122407892 on Build GRCh38
rs539024989 Chr.4: 122408130-122408130 on Build GRCh38
rs375153552 Chr.4: 122408048-122408048 on Build GRCh38
rs756215147 Chr.4: 122407875-122407875 on Build GRCh38
rs761519708 Chr.4: 122407958-122407958 on Build GRCh38
rs746331227 Chr.4: 122408070-122408070 on Build GRCh38
rs770035334 Chr.4: 122408158-122408158 on Build GRCh38
rs751437462 Chr.4: 122407866-122407866 on Build GRCh38
rs75008096 Chr.4: 122408073-122408073 on Build GRCh38
rs752936217 Chr.4: 122408045-122408045 on Build GRCh38
rs958631981 Chr.4: 122408167-122408167 on Build GRCh38
rs909218768 Chr.4: 122407812-122407812 on Build GRCh38
rs150494354 Chr.4: 122407950-122407950 on Build GRCh38
rs547951542 Chr.4: 122407789-122407789 on Build GRCh38
rs370894579 Chr.4: 122407911-122407911 on Build GRCh38
rs991302368 Chr.4: 122407854-122407854 on Build GRCh38

View Full Product Details