Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122411033-122411565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAD1
Entrez Gene ID
132612
Gene Name
adenosine deaminase domain containing 1
Gene Aliases
Tenr
UniGene
Hs.518957
Gene Chromosome Location
Chr.4: 122378966-122429815 on Build GRCh38
Transcripts
NM_001159285.1 NM_001159295.1 NM_139243.3 XM_005262741.2 XM_005262743.2 XM_005262744.2 XM_005262745.2 XM_011531603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4820525 c.971T>A p.L324H Chr.4: 122411344-122411344 on Build GRCh38
5400408 c.960C>T p.I320I Chr.4: 122411333-122411333 on Build GRCh38
278613 c.905A>C p.K302T Chr.4: 122411278-122411278 on Build GRCh38
301626 c.1019+1_1019+4delGTAA p.? Chr.4: 122411393-122411396 on Build GRCh38
4572865 c.888T>G p.N296K Chr.4: 122411261-122411261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939341516 Chr.4: 122411133-122411133 on Build GRCh38
rs116392967 Chr.4: 122411491-122411491 on Build GRCh38
rs371956235 Chr.4: 122411205-122411205 on Build GRCh38
rs769635674 Chr.4: 122411185-122411185 on Build GRCh38
rs754040910 Chr.4: 122411300-122411300 on Build GRCh38
rs765373740 Chr.4: 122411433-122411433 on Build GRCh38
rs375876455 Chr.4: 122411408-122411408 on Build GRCh38
rs1052660144 Chr.4: 122411159-122411159 on Build GRCh38
rs778818744 Chr.4: 122411315-122411315 on Build GRCh38
rs758774867 Chr.4: 122411335-122411335 on Build GRCh38
rs200348694 Chr.4: 122411260-122411260 on Build GRCh38
rs960673535 Chr.4: 122411456-122411456 on Build GRCh38
rs779996962 Chr.4: 122411372-122411372 on Build GRCh38
rs752425189 Chr.4: 122411283-122411283 on Build GRCh38
rs980636797 Chr.4: 122411531-122411531 on Build GRCh38
rs772172005 Chr.4: 122411290-122411290 on Build GRCh38
rs758489560 Chr.4: 122411439-122411439 on Build GRCh38
rs770312986 Chr.4: 122411222-122411222 on Build GRCh38
rs892136500 Chr.4: 122411160-122411160 on Build GRCh38
rs764067369 Chr.4: 122411102-122411102 on Build GRCh38
rs980818844 Chr.4: 122411443-122411443 on Build GRCh38
rs371568313 Chr.4: 122411405-122411405 on Build GRCh38
rs774372601 Chr.4: 122411224-122411224 on Build GRCh38
rs890089616 Chr.4: 122411402-122411402 on Build GRCh38
rs750404686 Chr.4: 122411438-122411438 on Build GRCh38
rs780700387 Chr.4: 122411347-122411347 on Build GRCh38
rs918267851 Chr.4: 122411069-122411069 on Build GRCh38
rs1008090284 Chr.4: 122411366-122411366 on Build GRCh38
rs1044581563 Chr.4: 122411099-122411099 on Build GRCh38
rs753745368 Chr.4: 122411164-122411164 on Build GRCh38
rs927910102 Chr.4: 122411119-122411119 on Build GRCh38
rs201052303 Chr.4: 122411403-122411403 on Build GRCh38
rs202125972 Chr.4: 122411292-122411292 on Build GRCh38
rs974127300 Chr.4: 122411436-122411436 on Build GRCh38
rs928846180 Chr.4: 122411101-122411101 on Build GRCh38
rs745819418 Chr.4: 122411387-122411387 on Build GRCh38
rs748849398 Chr.4: 122411364-122411364 on Build GRCh38
rs747817489 Chr.4: 122411183-122411183 on Build GRCh38
rs759340721 Chr.4: 122411252-122411252 on Build GRCh38
rs770493348 Chr.4: 122411370-122411370 on Build GRCh38
rs761893234 Chr.4: 122411430-122411430 on Build GRCh38
rs530807444 Chr.4: 122411355-122411355 on Build GRCh38
rs920452061 Chr.4: 122411086-122411086 on Build GRCh38
rs903615340 Chr.4: 122411238-122411238 on Build GRCh38
rs919974492 Chr.4: 122411475-122411475 on Build GRCh38
rs368081439 Chr.4: 122411178-122411178 on Build GRCh38
rs765554978 Chr.4: 122411421-122411422 on Build GRCh38
rs762600481 Chr.4: 122411206-122411206 on Build GRCh38
rs765289119 Chr.4: 122411518-122411518 on Build GRCh38
rs900035695 Chr.4: 122411329-122411329 on Build GRCh38
rs981302015 Chr.4: 122411104-122411104 on Build GRCh38
rs750853726 Chr.4: 122411422-122411423 on Build GRCh38
rs757399093 Chr.4: 122411306-122411306 on Build GRCh38
rs1043889105 Chr.4: 122411322-122411322 on Build GRCh38
rs757395686 Chr.4: 122411417-122411417 on Build GRCh38
rs142713989 Chr.4: 122411354-122411354 on Build GRCh38
rs542529171 Chr.4: 122411332-122411332 on Build GRCh38
rs562545888 Chr.4: 122411174-122411174 on Build GRCh38
rs928782125 Chr.4: 122411541-122411541 on Build GRCh38
rs1044411260 Chr.4: 122411539-122411539 on Build GRCh38
rs997061538 Chr.4: 122411282-122411282 on Build GRCh38
rs928048111 Chr.4: 122411452-122411452 on Build GRCh38
rs988122326 Chr.4: 122411490-122411490 on Build GRCh38
rs143785712 Chr.4: 122411349-122411349 on Build GRCh38
rs762357002 Chr.4: 122411255-122411256 on Build GRCh38
rs995196815 Chr.4: 122411421-122411421 on Build GRCh38

View Full Product Details