Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122896840-122897384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF2
Entrez Gene ID
2247
Gene Name
fibroblast growth factor 2
Gene Aliases
BFGF, FGF-2, FGFB, HBGF-2
UniGene
Hs.284244
Gene Chromosome Location
Chr.4: 122826708-122898235 on Build GRCh38
Transcripts
NM_002006.4 NM_007083.4 NM_198041.2

Gene Symbol
NUDT6
Entrez Gene ID
11162
Gene Name
nudix hydrolase 6
Gene Aliases
ASFGF2, FGF-AS, FGF2AS, GFG-1, GFG1
UniGene
Hs.558459
Gene Chromosome Location
Chr.4: 122892644-122923004 on Build GRCh38
Transcripts
NM_002006.4 NM_007083.4 NM_198041.2

SNP Detail

SNP ID Chromosome Location
rs968761944 Chr.4: 122897132-122897132 on Build GRCh38
rs902568482 Chr.4: 122896948-122896948 on Build GRCh38
rs761610705 Chr.4: 122896909-122896909 on Build GRCh38
rs949683611 Chr.4: 122897255-122897255 on Build GRCh38
rs1055419691 Chr.4: 122896918-122896918 on Build GRCh38
rs750100571 Chr.4: 122896904-122896904 on Build GRCh38
rs913091467 Chr.4: 122897126-122897126 on Build GRCh38
rs557179954 Chr.4: 122896886-122896886 on Build GRCh38
rs909374689 Chr.4: 122897169-122897169 on Build GRCh38
rs115581650 Chr.4: 122897117-122897117 on Build GRCh38
rs916815778 Chr.4: 122897225-122897225 on Build GRCh38
rs942260245 Chr.4: 122896910-122896910 on Build GRCh38
rs765435562 Chr.4: 122897275-122897276 on Build GRCh38
rs1476216 Chr.4: 122897143-122897143 on Build GRCh38
rs571992641 Chr.4: 122897076-122897076 on Build GRCh38
rs76656561 Chr.4: 122896889-122896889 on Build GRCh38
rs530807859 Chr.4: 122897338-122897338 on Build GRCh38
rs991400504 Chr.4: 122897189-122897189 on Build GRCh38
rs1476217 Chr.4: 122897356-122897356 on Build GRCh38
rs538183843 Chr.4: 122897333-122897333 on Build GRCh38
rs923961176 Chr.4: 122897300-122897300 on Build GRCh38
rs1053831439 Chr.4: 122897318-122897318 on Build GRCh38
rs1476215 Chr.4: 122897116-122897116 on Build GRCh38
rs912165524 Chr.4: 122897301-122897301 on Build GRCh38
rs45598031 Chr.4: 122896985-122896985 on Build GRCh38
rs935415415 Chr.4: 122897314-122897314 on Build GRCh38
rs7683093 Chr.4: 122896930-122896930 on Build GRCh38
rs545579663 Chr.4: 122896974-122896974 on Build GRCh38
rs576914614 Chr.4: 122896933-122896933 on Build GRCh38
rs193177947 Chr.4: 122897145-122897145 on Build GRCh38
rs963506997 Chr.4: 122897170-122897170 on Build GRCh38
rs776616085 Chr.4: 122896934-122896934 on Build GRCh38
rs3747676 Chr.4: 122896919-122896919 on Build GRCh38
rs373076210 Chr.4: 122896975-122896975 on Build GRCh38
rs754667365 Chr.4: 122897048-122897048 on Build GRCh38
rs45594531 Chr.4: 122897092-122897092 on Build GRCh38
rs983240927 Chr.4: 122897151-122897151 on Build GRCh38
rs1046852113 Chr.4: 122896943-122896943 on Build GRCh38
rs983588864 Chr.4: 122896900-122896900 on Build GRCh38
rs932218561 Chr.4: 122896903-122896903 on Build GRCh38
rs932009163 Chr.4: 122897259-122897259 on Build GRCh38
rs1046380015 Chr.4: 122897290-122897290 on Build GRCh38
rs536593273 Chr.4: 122897153-122897153 on Build GRCh38
rs1052417278 Chr.4: 122897298-122897298 on Build GRCh38
rs752640425 Chr.4: 122897113-122897113 on Build GRCh38
rs552375660 Chr.4: 122897350-122897350 on Build GRCh38

View Full Product Details