Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 139044943-139045449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOCT
Entrez Gene ID
25819
Gene Name
nocturnin
Gene Aliases
CCR4L, CCRN4L, Ccr4c, NOC
UniGene
Hs.639842
Gene Chromosome Location
Chr.4: 139015759-139045939 on Build GRCh38
Transcripts
NM_012118.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1727159 c.1105T>C p.S369P Chr.4: 139045283-139045283 on Build GRCh38
7248887 c.1238A>G p.D413G Chr.4: 139045416-139045416 on Build GRCh38
7209554 c.1222T>C p.F408L Chr.4: 139045400-139045400 on Build GRCh38
5303077 c.1176C>T p.L392L Chr.4: 139045354-139045354 on Build GRCh38
5644343 c.869G>A p.R290H Chr.4: 139045047-139045047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748771340 Chr.4: 139045037-139045037 on Build GRCh38
rs1012075580 Chr.4: 139045193-139045193 on Build GRCh38
rs368722959 Chr.4: 139045103-139045103 on Build GRCh38
rs763885264 Chr.4: 139045261-139045261 on Build GRCh38
rs144267239 Chr.4: 139045015-139045015 on Build GRCh38
rs371042290 Chr.4: 139045025-139045025 on Build GRCh38
rs371368774 Chr.4: 139045117-139045117 on Build GRCh38
rs778411432 Chr.4: 139045151-139045151 on Build GRCh38
rs373770561 Chr.4: 139045008-139045008 on Build GRCh38
rs770391718 Chr.4: 139045300-139045300 on Build GRCh38
rs745430849 Chr.4: 139045155-139045155 on Build GRCh38
rs761638930 Chr.4: 139045338-139045338 on Build GRCh38
rs770981949 Chr.4: 139045384-139045384 on Build GRCh38
rs765227776 Chr.4: 139045273-139045273 on Build GRCh38
rs765412497 Chr.4: 139045089-139045089 on Build GRCh38
rs746017569 Chr.4: 139045398-139045398 on Build GRCh38
rs759772556 Chr.4: 139045327-139045327 on Build GRCh38
rs776520225 Chr.4: 139045354-139045355 on Build GRCh38
rs77592090 Chr.4: 139045292-139045292 on Build GRCh38
rs372649885 Chr.4: 139045375-139045375 on Build GRCh38
rs761728813 Chr.4: 139045268-139045268 on Build GRCh38
rs771159932 Chr.4: 139045239-139045239 on Build GRCh38
rs919299627 Chr.4: 139045058-139045058 on Build GRCh38
rs778849254 Chr.4: 139045387-139045387 on Build GRCh38
rs760474553 Chr.4: 139045254-139045254 on Build GRCh38
rs145515684 Chr.4: 139045099-139045099 on Build GRCh38
rs769508036 Chr.4: 139045421-139045421 on Build GRCh38
rs550157246 Chr.4: 139045348-139045348 on Build GRCh38
rs932133701 Chr.4: 139045017-139045017 on Build GRCh38
rs764569893 Chr.4: 139045007-139045007 on Build GRCh38
rs776409805 Chr.4: 139045331-139045331 on Build GRCh38
rs953118791 Chr.4: 139045388-139045388 on Build GRCh38
rs143546269 Chr.4: 139045282-139045282 on Build GRCh38
rs112187808 Chr.4: 139045263-139045263 on Build GRCh38
rs775739555 Chr.4: 139045401-139045401 on Build GRCh38
rs1039561902 Chr.4: 139045189-139045189 on Build GRCh38
rs748678311 Chr.4: 139045216-139045216 on Build GRCh38
rs555003359 Chr.4: 139045174-139045174 on Build GRCh38
rs759840367 Chr.4: 139044964-139044964 on Build GRCh38
rs747379240 Chr.4: 139045172-139045172 on Build GRCh38
rs773335755 Chr.4: 139045231-139045231 on Build GRCh38
rs778300044 Chr.4: 139045376-139045376 on Build GRCh38
rs557454341 Chr.4: 139045343-139045343 on Build GRCh38
rs777238536 Chr.4: 139045213-139045213 on Build GRCh38
rs370362401 Chr.4: 139044998-139044998 on Build GRCh38
rs772252609 Chr.4: 139045399-139045399 on Build GRCh38
rs763967112 Chr.4: 139045126-139045126 on Build GRCh38
rs761261263 Chr.4: 139044987-139044987 on Build GRCh38
rs374898315 Chr.4: 139045344-139045344 on Build GRCh38
rs774562025 Chr.4: 139045325-139045325 on Build GRCh38
rs773135002 Chr.4: 139045422-139045422 on Build GRCh38
rs749760177 Chr.4: 139045382-139045382 on Build GRCh38
rs144899070 Chr.4: 139045355-139045355 on Build GRCh38
rs772342321 Chr.4: 139045329-139045329 on Build GRCh38
rs750696054 Chr.4: 139045014-139045014 on Build GRCh38
rs754672120 Chr.4: 139045351-139045351 on Build GRCh38
rs756693534 Chr.4: 139045141-139045141 on Build GRCh38
rs777328444 Chr.4: 139045068-139045068 on Build GRCh38
rs769256787 Chr.4: 139045067-139045067 on Build GRCh38
rs764470353 Chr.4: 139045414-139045414 on Build GRCh38
rs765200259 Chr.4: 139045340-139045340 on Build GRCh38
rs894523592 Chr.4: 139045121-139045121 on Build GRCh38
rs570831721 Chr.4: 139045131-139045131 on Build GRCh38
rs775797715 Chr.4: 139044974-139044974 on Build GRCh38
rs531162189 Chr.4: 139045046-139045046 on Build GRCh38
rs779614892 Chr.4: 139045166-139045166 on Build GRCh38
rs141093326 Chr.4: 139045354-139045354 on Build GRCh38
rs765534383 Chr.4: 139045010-139045010 on Build GRCh38
rs903699987 Chr.4: 139045278-139045278 on Build GRCh38
rs534652931 Chr.4: 139045162-139045162 on Build GRCh38
rs770942853 Chr.4: 139045313-139045313 on Build GRCh38
rs756072524 Chr.4: 139045033-139045033 on Build GRCh38
rs749919641 Chr.4: 139045277-139045277 on Build GRCh38
rs150993247 Chr.4: 139045291-139045291 on Build GRCh38
rs756778673 Chr.4: 139045045-139045045 on Build GRCh38
rs751202695 Chr.4: 139045283-139045283 on Build GRCh38
rs754662993 Chr.4: 139045024-139045024 on Build GRCh38
rs184864849 Chr.4: 139045413-139045413 on Build GRCh38
rs375776058 Chr.4: 139045062-139045062 on Build GRCh38
rs371556362 Chr.4: 139045061-139045061 on Build GRCh38
rs1043541532 Chr.4: 139045209-139045209 on Build GRCh38
rs770479937 Chr.4: 139045217-139045217 on Build GRCh38
rs766293473 Chr.4: 139044978-139044978 on Build GRCh38
rs769166706 Chr.4: 139045192-139045192 on Build GRCh38
rs774455427 Chr.4: 139045241-139045241 on Build GRCh38
rs372648550 Chr.4: 139045153-139045153 on Build GRCh38
rs984943906 Chr.4: 139045022-139045022 on Build GRCh38
rs762981302 Chr.4: 139045237-139045237 on Build GRCh38
rs202135520 Chr.4: 139045112-139045112 on Build GRCh38
rs762786108 Chr.4: 139045423-139045423 on Build GRCh38
rs778189361 Chr.4: 139045304-139045304 on Build GRCh38
rs752504077 Chr.4: 139045116-139045116 on Build GRCh38
rs761936630 Chr.4: 139045077-139045077 on Build GRCh38
rs773510297 Chr.4: 139045090-139045090 on Build GRCh38
rs768951060 Chr.4: 139045207-139045207 on Build GRCh38
rs757896028 Chr.4: 139045281-139045281 on Build GRCh38
rs753800509 Chr.4: 139045129-139045129 on Build GRCh38
rs753720979 Chr.4: 139045265-139045265 on Build GRCh38
rs772522544 Chr.4: 139044970-139044970 on Build GRCh38
rs752491438 Chr.4: 139045032-139045032 on Build GRCh38
rs745543266 Chr.4: 139045047-139045047 on Build GRCh38
rs921744903 Chr.4: 139045011-139045011 on Build GRCh38
rs369138900 Chr.4: 139045346-139045346 on Build GRCh38
rs759058033 Chr.4: 139045006-139045006 on Build GRCh38

View Full Product Details