Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 139045477-139046016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOCT
Entrez Gene ID
25819
Gene Name
nocturnin
Gene Aliases
CCR4L, CCRN4L, Ccr4c, NOC
UniGene
Hs.639842
Gene Chromosome Location
Chr.4: 139015759-139045939 on Build GRCh38
Transcripts
NM_012118.3

SNP Detail

SNP ID Chromosome Location
rs143956839 Chr.4: 139045507-139045509 on Build GRCh38
rs537515514 Chr.4: 139045545-139045545 on Build GRCh38
rs546933608 Chr.4: 139045661-139045661 on Build GRCh38
rs765445144 Chr.4: 139045510-139045516 on Build GRCh38
rs866844114 Chr.4: 139045666-139045666 on Build GRCh38
rs910607895 Chr.4: 139045587-139045587 on Build GRCh38
rs749525572 Chr.4: 139045856-139045856 on Build GRCh38
rs977249121 Chr.4: 139045914-139045914 on Build GRCh38
rs1050803462 Chr.4: 139045733-139045733 on Build GRCh38
rs563412654 Chr.4: 139045610-139045610 on Build GRCh38
rs991738463 Chr.4: 139045840-139045840 on Build GRCh38
rs574421684 Chr.4: 139045599-139045599 on Build GRCh38
rs77121168 Chr.4: 139045877-139045877 on Build GRCh38
rs933834912 Chr.4: 139045629-139045629 on Build GRCh38
rs947464721 Chr.4: 139045558-139045558 on Build GRCh38
rs934711710 Chr.4: 139045885-139045885 on Build GRCh38
rs941952540 Chr.4: 139045964-139045964 on Build GRCh38
rs28646895 Chr.4: 139045619-139045619 on Build GRCh38
rs976158885 Chr.4: 139045869-139045869 on Build GRCh38
rs767221587 Chr.4: 139045519-139045520 on Build GRCh38
rs1050355555 Chr.4: 139045946-139045946 on Build GRCh38
rs769574813 Chr.4: 139045507-139045507 on Build GRCh38
rs866466181 Chr.4: 139045535-139045535 on Build GRCh38
rs1052230376 Chr.4: 139045706-139045706 on Build GRCh38
rs964851237 Chr.4: 139045595-139045595 on Build GRCh38
rs924519300 Chr.4: 139045915-139045915 on Build GRCh38
rs770815841 Chr.4: 139045927-139045927 on Build GRCh38
rs1035790946 Chr.4: 139045827-139045827 on Build GRCh38
rs540988141 Chr.4: 139045537-139045537 on Build GRCh38
rs1055757449 Chr.4: 139045543-139045543 on Build GRCh38
rs60662360 Chr.4: 139045534-139045535 on Build GRCh38
rs550766803 Chr.4: 139045686-139045686 on Build GRCh38
rs188543771 Chr.4: 139045603-139045603 on Build GRCh38
rs763044211 Chr.4: 139045510-139045517 on Build GRCh38
rs1015887944 Chr.4: 139045768-139045768 on Build GRCh38
rs564396604 Chr.4: 139045652-139045652 on Build GRCh38
rs978838641 Chr.4: 139045682-139045682 on Build GRCh38
rs777321337 Chr.4: 139045523-139045524 on Build GRCh38
rs754546916 Chr.4: 139045518-139045519 on Build GRCh38
rs760757770 Chr.4: 139045803-139045803 on Build GRCh38
rs530984087 Chr.4: 139045618-139045618 on Build GRCh38
rs773281448 Chr.4: 139045510-139045522 on Build GRCh38
rs546346607 Chr.4: 139045509-139045510 on Build GRCh38
rs186208185 Chr.4: 139045737-139045737 on Build GRCh38
rs934741136 Chr.4: 139045704-139045704 on Build GRCh38
rs1005950050 Chr.4: 139045639-139045639 on Build GRCh38
rs752398312 Chr.4: 139045520-139045521 on Build GRCh38
rs947399835 Chr.4: 139045674-139045674 on Build GRCh38
rs1037305841 Chr.4: 139045983-139045983 on Build GRCh38
rs972172113 Chr.4: 139045663-139045663 on Build GRCh38
rs993708853 Chr.4: 139045503-139045503 on Build GRCh38
rs746800275 Chr.4: 139045926-139045926 on Build GRCh38
rs796384573 Chr.4: 139045675-139045675 on Build GRCh38
rs553261505 Chr.4: 139045788-139045788 on Build GRCh38
rs181207675 Chr.4: 139045714-139045714 on Build GRCh38
rs117927123 Chr.4: 139045738-139045738 on Build GRCh38
rs951995835 Chr.4: 139045575-139045575 on Build GRCh38
rs923118595 Chr.4: 139045910-139045910 on Build GRCh38
rs751377958 Chr.4: 139045510-139045518 on Build GRCh38
rs575004537 Chr.4: 139045884-139045884 on Build GRCh38
rs935968075 Chr.4: 139045916-139045916 on Build GRCh38
rs541235439 Chr.4: 139045719-139045719 on Build GRCh38
rs137915288 Chr.4: 139045662-139045662 on Build GRCh38
rs796306859 Chr.4: 139045969-139045969 on Build GRCh38
rs768565041 Chr.4: 139045547-139045547 on Build GRCh38
rs74826922 Chr.4: 139045509-139045509 on Build GRCh38
rs915541603 Chr.4: 139045944-139045944 on Build GRCh38
rs879424914 Chr.4: 139045671-139045671 on Build GRCh38
rs897537786 Chr.4: 139045796-139045796 on Build GRCh38
rs768960609 Chr.4: 139045868-139045868 on Build GRCh38
rs888996115 Chr.4: 139045959-139045959 on Build GRCh38
rs143949609 Chr.4: 139045857-139045857 on Build GRCh38
rs766034892 Chr.4: 139045510-139045519 on Build GRCh38
rs567744832 Chr.4: 139045665-139045665 on Build GRCh38
rs545533241 Chr.4: 139045913-139045913 on Build GRCh38
rs1052346742 Chr.4: 139045594-139045594 on Build GRCh38
rs866490277 Chr.4: 139045602-139045602 on Build GRCh38
rs984826054 Chr.4: 139045577-139045577 on Build GRCh38
rs112018412 Chr.4: 139045655-139045655 on Build GRCh38
rs924725175 Chr.4: 139045699-139045699 on Build GRCh38
rs923370472 Chr.4: 139045876-139045877 on Build GRCh38
rs755591282 Chr.4: 139045522-139045523 on Build GRCh38
rs976848029 Chr.4: 139045894-139045894 on Build GRCh38
rs1043194856 Chr.4: 139045562-139045562 on Build GRCh38
rs903672067 Chr.4: 139045585-139045585 on Build GRCh38
rs761959376 Chr.4: 139045507-139045508 on Build GRCh38
rs149471540 Chr.4: 139045717-139045717 on Build GRCh38
rs915899918 Chr.4: 139045673-139045673 on Build GRCh38
rs564335847 Chr.4: 139045962-139045962 on Build GRCh38
rs999401603 Chr.4: 139045586-139045586 on Build GRCh38
rs747236099 Chr.4: 139045505-139045505 on Build GRCh38
rs535263276 Chr.4: 139045832-139045832 on Build GRCh38

View Full Product Details