Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 140393746-140394248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLGN
Entrez Gene ID
1047
Gene Name
calmegin
Gene Aliases
-
UniGene
Hs.86368
Gene Chromosome Location
Chr.4: 140388453-140427661 on Build GRCh38
Transcripts
NM_001130675.1 NM_004362.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4600113 c.1357G>C p.A453P Chr.4: 140393834-140393834 on Build GRCh38
6319530 c.1319C>A p.A440D Chr.4: 140393872-140393872 on Build GRCh38
1201368 c.1223T>C p.F408S Chr.4: 140393968-140393968 on Build GRCh38
3600452 c.1180C>T p.P394S Chr.4: 140394011-140394011 on Build GRCh38
3600451 c.1274A>C p.D425A Chr.4: 140393917-140393917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768944085 Chr.4: 140393822-140393822 on Build GRCh38
rs745633273 Chr.4: 140394076-140394076 on Build GRCh38
rs763692996 Chr.4: 140393892-140393892 on Build GRCh38
rs141702107 Chr.4: 140393855-140393855 on Build GRCh38
rs200652126 Chr.4: 140393882-140393882 on Build GRCh38
rs139581390 Chr.4: 140393899-140393899 on Build GRCh38
rs1048056786 Chr.4: 140393886-140393886 on Build GRCh38
rs781415802 Chr.4: 140394053-140394053 on Build GRCh38
rs770728700 Chr.4: 140393862-140393862 on Build GRCh38
rs551992592 Chr.4: 140394179-140394179 on Build GRCh38
rs376420992 Chr.4: 140394152-140394153 on Build GRCh38
rs563971370 Chr.4: 140393971-140393971 on Build GRCh38
rs570309700 Chr.4: 140394202-140394202 on Build GRCh38
rs372000570 Chr.4: 140393931-140393931 on Build GRCh38
rs953158697 Chr.4: 140393839-140393839 on Build GRCh38
rs190724420 Chr.4: 140394132-140394132 on Build GRCh38
rs747092384 Chr.4: 140393819-140393819 on Build GRCh38
rs939656755 Chr.4: 140393929-140393929 on Build GRCh38
rs779869380 Chr.4: 140394085-140394085 on Build GRCh38
rs142857415 Chr.4: 140393942-140393942 on Build GRCh38
rs542190561 Chr.4: 140393907-140393907 on Build GRCh38
rs113934195 Chr.4: 140394149-140394150 on Build GRCh38
rs982648514 Chr.4: 140394122-140394122 on Build GRCh38
rs777017750 Chr.4: 140393854-140393854 on Build GRCh38
rs771819099 Chr.4: 140394079-140394079 on Build GRCh38
rs774127578 Chr.4: 140393986-140393986 on Build GRCh38
rs918628588 Chr.4: 140394197-140394197 on Build GRCh38
rs750744079 Chr.4: 140394026-140394026 on Build GRCh38
rs973224601 Chr.4: 140394193-140394193 on Build GRCh38
rs767869735 Chr.4: 140394203-140394203 on Build GRCh38
rs866469996 Chr.4: 140394011-140394011 on Build GRCh38
rs776065662 Chr.4: 140394017-140394017 on Build GRCh38
rs746368987 Chr.4: 140393802-140393802 on Build GRCh38
rs371614813 Chr.4: 140393995-140393995 on Build GRCh38
rs1032278528 Chr.4: 140394059-140394059 on Build GRCh38
rs757667634 Chr.4: 140393785-140393785 on Build GRCh38
rs186160583 Chr.4: 140394111-140394111 on Build GRCh38
rs984596633 Chr.4: 140394130-140394130 on Build GRCh38
rs181401532 Chr.4: 140394102-140394102 on Build GRCh38
rs759512289 Chr.4: 140393868-140393868 on Build GRCh38
rs76550209 Chr.4: 140394000-140394000 on Build GRCh38
rs369401077 Chr.4: 140393957-140393957 on Build GRCh38
rs960955505 Chr.4: 140394207-140394207 on Build GRCh38
rs780722455 Chr.4: 140393933-140393933 on Build GRCh38
rs77827238 Chr.4: 140393873-140393873 on Build GRCh38
rs138845069 Chr.4: 140394020-140394020 on Build GRCh38
rs564584207 Chr.4: 140393999-140393999 on Build GRCh38
rs752082133 Chr.4: 140394043-140394043 on Build GRCh38
rs369314028 Chr.4: 140393803-140393803 on Build GRCh38
rs765625742 Chr.4: 140393904-140393904 on Build GRCh38
rs1039664461 Chr.4: 140394219-140394219 on Build GRCh38
rs373256495 Chr.4: 140394065-140394065 on Build GRCh38
rs760197947 Chr.4: 140393888-140393888 on Build GRCh38
rs918563581 Chr.4: 140393894-140393894 on Build GRCh38
rs868121960 Chr.4: 140393903-140393903 on Build GRCh38
rs763605792 Chr.4: 140393783-140393783 on Build GRCh38
rs780279369 Chr.4: 140393815-140393815 on Build GRCh38
rs755140861 Chr.4: 140393951-140393951 on Build GRCh38
rs139813038 Chr.4: 140394031-140394031 on Build GRCh38
rs111866427 Chr.4: 140393966-140393966 on Build GRCh38
rs752724334 Chr.4: 140394061-140394061 on Build GRCh38
rs368143808 Chr.4: 140393781-140393781 on Build GRCh38
rs970959029 Chr.4: 140394103-140394103 on Build GRCh38
rs767445232 Chr.4: 140393874-140393874 on Build GRCh38
rs777879388 Chr.4: 140394066-140394066 on Build GRCh38
rs748388584 Chr.4: 140393960-140393960 on Build GRCh38
rs962848367 Chr.4: 140394158-140394158 on Build GRCh38
rs374700061 Chr.4: 140393792-140393792 on Build GRCh38
rs575768191 Chr.4: 140393988-140393988 on Build GRCh38
rs150556621 Chr.4: 140393898-140393898 on Build GRCh38
rs570518802 Chr.4: 140394051-140394051 on Build GRCh38
rs200529128 Chr.4: 140393864-140393864 on Build GRCh38
rs149292968 Chr.4: 140394136-140394136 on Build GRCh38
rs372543725 Chr.4: 140393856-140393856 on Build GRCh38
rs189753269 Chr.4: 140393867-140393867 on Build GRCh38
rs368889697 Chr.4: 140394025-140394025 on Build GRCh38
rs867165670 Chr.4: 140393844-140393844 on Build GRCh38
rs147434096 Chr.4: 140393998-140393998 on Build GRCh38
rs942353847 Chr.4: 140394199-140394199 on Build GRCh38
rs992372237 Chr.4: 140394215-140394215 on Build GRCh38

View Full Product Details