Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145064486-145064993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC10
Entrez Gene ID
10393
Gene Name
anaphase promoting complex subunit 10
Gene Aliases
APC10, DOC1
UniGene
Hs.480876
Gene Chromosome Location
Chr.4: 144967117-145098541 on Build GRCh38
Transcripts
NM_001256706.1 NM_001256707.1 NM_001256708.1 NM_001256709.1 NM_001256710.1 NM_001256711.1 NM_001256712.1 NM_001318367.1 NM_014885.4 NR_046345.1 XM_011531524.2 XM_011531525.2 XM_011531526.2 XM_011531527.2 XM_011531529.2 XM_011531530.2 XM_011531531.2 XM_011531532.2 XM_017007638.1 XM_017007639.1 XM_017007640.1 XM_017007641.1 XM_017007642.1 XM_017007643.1 XM_017007644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7312795 c.269C>T p.T90I Chr.4: 145064630-145064630 on Build GRCh38
1427343 c.267T>C p.Y89Y Chr.4: 145064632-145064632 on Build GRCh38
136444 c.307C>T p.H103Y Chr.4: 145064592-145064592 on Build GRCh38
3600626 c.254C>T p.S85F Chr.4: 145064645-145064645 on Build GRCh38
4637945 c.309C>T p.H103H Chr.4: 145064590-145064590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886105409 Chr.4: 145064951-145064951 on Build GRCh38
rs947408112 Chr.4: 145064814-145064814 on Build GRCh38
rs954909755 Chr.4: 145064603-145064603 on Build GRCh38
rs372866692 Chr.4: 145064863-145064863 on Build GRCh38
rs749794176 Chr.4: 145064538-145064538 on Build GRCh38
rs372401997 Chr.4: 145064590-145064590 on Build GRCh38
rs750440669 Chr.4: 145064607-145064607 on Build GRCh38
rs949978503 Chr.4: 145064869-145064869 on Build GRCh38
rs994359834 Chr.4: 145064907-145064907 on Build GRCh38
rs913220136 Chr.4: 145064793-145064793 on Build GRCh38
rs938598382 Chr.4: 145064781-145064781 on Build GRCh38
rs369523731 Chr.4: 145064719-145064719 on Build GRCh38
rs766028459 Chr.4: 145064620-145064620 on Build GRCh38
rs1023065805 Chr.4: 145064545-145064546 on Build GRCh38
rs969879082 Chr.4: 145064610-145064610 on Build GRCh38
rs979942999 Chr.4: 145064705-145064705 on Build GRCh38
rs1054430568 Chr.4: 145064937-145064937 on Build GRCh38
rs113933578 Chr.4: 145064855-145064855 on Build GRCh38
rs376998760 Chr.4: 145064716-145064716 on Build GRCh38
rs1006305080 Chr.4: 145064967-145064967 on Build GRCh38
rs541388710 Chr.4: 145064917-145064917 on Build GRCh38
rs989024450 Chr.4: 145064655-145064655 on Build GRCh38
rs754700610 Chr.4: 145064673-145064673 on Build GRCh38
rs988752523 Chr.4: 145064790-145064790 on Build GRCh38
rs765089127 Chr.4: 145064605-145064605 on Build GRCh38
rs756571582 Chr.4: 145064720-145064720 on Build GRCh38
rs1000270711 Chr.4: 145064582-145064582 on Build GRCh38
rs761050211 Chr.4: 145064728-145064729 on Build GRCh38
rs759790944 Chr.4: 145064559-145064559 on Build GRCh38
rs143503304 Chr.4: 145064777-145064777 on Build GRCh38
rs778279410 Chr.4: 145064532-145064532 on Build GRCh38
rs764236630 Chr.4: 145064886-145064888 on Build GRCh38
rs34032580 Chr.4: 145064940-145064940 on Build GRCh38
rs747417617 Chr.4: 145064675-145064681 on Build GRCh38
rs761610901 Chr.4: 145064600-145064600 on Build GRCh38
rs901406789 Chr.4: 145064892-145064892 on Build GRCh38
rs748499295 Chr.4: 145064524-145064524 on Build GRCh38
rs920263912 Chr.4: 145064830-145064830 on Build GRCh38
rs1043143304 Chr.4: 145064844-145064844 on Build GRCh38
rs369778610 Chr.4: 145064588-145064588 on Build GRCh38
rs895661070 Chr.4: 145064519-145064519 on Build GRCh38
rs866908810 Chr.4: 145064657-145064657 on Build GRCh38
rs973149618 Chr.4: 145064813-145064813 on Build GRCh38
rs767143852 Chr.4: 145064703-145064703 on Build GRCh38
rs942836814 Chr.4: 145064856-145064856 on Build GRCh38
rs781441929 Chr.4: 145064521-145064521 on Build GRCh38
rs572575836 Chr.4: 145064882-145064882 on Build GRCh38
rs34600335 Chr.4: 145064579-145064580 on Build GRCh38
rs905963724 Chr.4: 145064572-145064572 on Build GRCh38
rs780429965 Chr.4: 145064528-145064528 on Build GRCh38
rs751074331 Chr.4: 145064658-145064658 on Build GRCh38
rs770221015 Chr.4: 145064528-145064528 on Build GRCh38
rs1044647662 Chr.4: 145064870-145064870 on Build GRCh38
rs986269649 Chr.4: 145064765-145064765 on Build GRCh38
rs757727366 Chr.4: 145064722-145064722 on Build GRCh38
rs762962943 Chr.4: 145064612-145064612 on Build GRCh38
rs376827906 Chr.4: 145064540-145064540 on Build GRCh38
rs1000275477 Chr.4: 145064924-145064924 on Build GRCh38
rs774390312 Chr.4: 145064544-145064544 on Build GRCh38
rs1047641622 Chr.4: 145064517-145064517 on Build GRCh38

View Full Product Details