Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145097597-145098091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCE1
Entrez Gene ID
6059
Gene Name
ATP binding cassette subfamily E member 1
Gene Aliases
ABC38, OABP, RLI, RLI1, RNASEL1, RNASELI, RNS4I
UniGene
Hs.12013
Gene Chromosome Location
Chr.4: 145098004-145129524 on Build GRCh38
Transcripts
NM_001040876.1 NM_002940.2 NM_001256706.1 NM_001256707.1 NM_001256708.1 NM_001256709.1 NM_001256710.1 NM_001256711.1 NM_001256712.1 NM_014885.4 NR_046345.1 XM_011531524.2 XM_011531525.2 XM_011531526.2 XM_011531527.2 XM_011531529.2 XM_011531530.2 XM_011531531.2 XM_011531532.2 XM_017007638.1 XM_017007639.1 XM_017007640.1 XM_017007641.1 XM_017007642.1 XM_017007643.1 XM_017007644.1

Gene Symbol
ANAPC10
Entrez Gene ID
10393
Gene Name
anaphase promoting complex subunit 10
Gene Aliases
APC10, DOC1
UniGene
Hs.480876
Gene Chromosome Location
Chr.4: 144967117-145098541 on Build GRCh38
Transcripts
NM_001040876.1 NM_002940.2 NM_001256706.1 NM_001256707.1 NM_001256708.1 NM_001256709.1 NM_001256710.1 NM_001256711.1 NM_001256712.1 NM_014885.4 NR_046345.1 XM_011531524.2 XM_011531525.2 XM_011531526.2 XM_011531527.2 XM_011531529.2 XM_011531530.2 XM_011531531.2 XM_011531532.2 XM_017007638.1 XM_017007639.1 XM_017007640.1 XM_017007641.1 XM_017007642.1 XM_017007643.1 XM_017007644.1

SNP Detail

SNP ID Chromosome Location
rs185818965 Chr.4: 145097647-145097647 on Build GRCh38
rs949077153 Chr.4: 145097905-145097905 on Build GRCh38
rs573120842 Chr.4: 145097882-145097882 on Build GRCh38
rs760717543 Chr.4: 145097670-145097670 on Build GRCh38
rs922910161 Chr.4: 145097946-145097946 on Build GRCh38
rs887591855 Chr.4: 145097712-145097712 on Build GRCh38
rs1050329927 Chr.4: 145097955-145097955 on Build GRCh38
rs143173559 Chr.4: 145097675-145097675 on Build GRCh38
rs996136949 Chr.4: 145098012-145098012 on Build GRCh38
rs900408896 Chr.4: 145098010-145098010 on Build GRCh38
rs1014923991 Chr.4: 145097772-145097772 on Build GRCh38
rs770682613 Chr.4: 145097677-145097677 on Build GRCh38
rs151233911 Chr.4: 145097777-145097777 on Build GRCh38
rs923675811 Chr.4: 145097936-145097936 on Build GRCh38
rs562099261 Chr.4: 145097944-145097944 on Build GRCh38
rs1043449647 Chr.4: 145097664-145097664 on Build GRCh38
rs900417883 Chr.4: 145097703-145097703 on Build GRCh38
rs373642093 Chr.4: 145097884-145097884 on Build GRCh38
rs1006728136 Chr.4: 145097760-145097760 on Build GRCh38
rs936467034 Chr.4: 145097971-145097971 on Build GRCh38
rs995407774 Chr.4: 145098053-145098053 on Build GRCh38
rs991726744 Chr.4: 145097879-145097879 on Build GRCh38
rs1002822589 Chr.4: 145097807-145097807 on Build GRCh38
rs973600432 Chr.4: 145097838-145097838 on Build GRCh38
rs996050224 Chr.4: 145097689-145097689 on Build GRCh38
rs887612591 Chr.4: 145097958-145097958 on Build GRCh38
rs990880372 Chr.4: 145097898-145097898 on Build GRCh38
rs1053664124 Chr.4: 145097993-145097993 on Build GRCh38
rs1004994369 Chr.4: 145097816-145097816 on Build GRCh38
rs539503062 Chr.4: 145097833-145097833 on Build GRCh38
rs1035932882 Chr.4: 145097825-145097825 on Build GRCh38
rs542184823 Chr.4: 145097921-145097921 on Build GRCh38
rs137970745 Chr.4: 145097638-145097638 on Build GRCh38
rs1050706619 Chr.4: 145097691-145097691 on Build GRCh38
rs941871748 Chr.4: 145097991-145097991 on Build GRCh38
rs1455135 Chr.4: 145097667-145097667 on Build GRCh38
rs901745826 Chr.4: 145097684-145097684 on Build GRCh38
rs917608728 Chr.4: 145097904-145097904 on Build GRCh38
rs954762720 Chr.4: 145097811-145097811 on Build GRCh38
rs568110381 Chr.4: 145098003-145098003 on Build GRCh38
rs975727044 Chr.4: 145097945-145097945 on Build GRCh38
rs145916143 Chr.4: 145098019-145098019 on Build GRCh38
rs927535135 Chr.4: 145097841-145097841 on Build GRCh38
rs138810873 Chr.4: 145098038-145098038 on Build GRCh38
rs550063794 Chr.4: 145097917-145097917 on Build GRCh38
rs890437324 Chr.4: 145098035-145098035 on Build GRCh38
rs1015029990 Chr.4: 145097832-145097832 on Build GRCh38
rs914721865 Chr.4: 145097626-145097626 on Build GRCh38
rs34683609 Chr.4: 145098045-145098045 on Build GRCh38
rs996576085 Chr.4: 145097774-145097774 on Build GRCh38
rs959230801 Chr.4: 145097873-145097873 on Build GRCh38
rs923699369 Chr.4: 145097918-145097918 on Build GRCh38
rs1014977796 Chr.4: 145098043-145098043 on Build GRCh38
rs553441793 Chr.4: 145097864-145097864 on Build GRCh38
rs33923494 Chr.4: 145097618-145097618 on Build GRCh38
rs1035666808 Chr.4: 145098061-145098061 on Build GRCh38
rs894441855 Chr.4: 145098069-145098069 on Build GRCh38
rs959022133 Chr.4: 145097847-145097847 on Build GRCh38

View Full Product Details