Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145726825-145727280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf51
Entrez Gene ID
646603
Gene Name
chromosome 4 open reading frame 51
Gene Aliases
-
UniGene
Hs.452865
Gene Chromosome Location
Chr.4: 145680103-145792248 on Build GRCh38
Transcripts
NM_001080531.1 XM_006714289.3 XM_011532196.2 XM_011532197.2 XM_017008547.1 XM_017008548.1 XM_017008549.1 XM_017008550.1 XM_017008551.1 XM_017008552.1 XM_017008553.1 XM_017008554.1 XM_017008555.1 XM_017008556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3724958 c.366+10A>G p.? Chr.4: 145726979-145726979 on Build GRCh38
5995749 c.308-1G>T p.? Chr.4: 145726910-145726910 on Build GRCh38
3428208 c.308G>A p.G103E Chr.4: 145726911-145726911 on Build GRCh38
1283823 c.309A>G p.G103G Chr.4: 145726912-145726912 on Build GRCh38
4913101 c.345A>G p.S115S Chr.4: 145726948-145726948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750584342 Chr.4: 145726900-145726900 on Build GRCh38
rs1032851603 Chr.4: 145727252-145727252 on Build GRCh38
rs142441248 Chr.4: 145726913-145726914 on Build GRCh38
rs537404428 Chr.4: 145727166-145727166 on Build GRCh38
rs761909280 Chr.4: 145726893-145726893 on Build GRCh38
rs951424513 Chr.4: 145727238-145727238 on Build GRCh38
rs756761258 Chr.4: 145726929-145726929 on Build GRCh38
rs752508416 Chr.4: 145726918-145726918 on Build GRCh38
rs911339434 Chr.4: 145726852-145726852 on Build GRCh38
rs527907784 Chr.4: 145726927-145726927 on Build GRCh38
rs367871550 Chr.4: 145726882-145726882 on Build GRCh38
rs752002463 Chr.4: 145727136-145727136 on Build GRCh38
rs376727006 Chr.4: 145726870-145726870 on Build GRCh38
rs767369875 Chr.4: 145726911-145726911 on Build GRCh38
rs535521371 Chr.4: 145727139-145727139 on Build GRCh38
rs748185452 Chr.4: 145727003-145727003 on Build GRCh38
rs941574835 Chr.4: 145726970-145726970 on Build GRCh38
rs146275366 Chr.4: 145727162-145727162 on Build GRCh38
rs184976257 Chr.4: 145726932-145726932 on Build GRCh38
rs765446268 Chr.4: 145726899-145726899 on Build GRCh38
rs752822243 Chr.4: 145726995-145726995 on Build GRCh38
rs565390537 Chr.4: 145726910-145726910 on Build GRCh38
rs756058976 Chr.4: 145726921-145726921 on Build GRCh38
rs922647592 Chr.4: 145727236-145727236 on Build GRCh38
rs778233041 Chr.4: 145727130-145727130 on Build GRCh38
rs1056873809 Chr.4: 145727089-145727089 on Build GRCh38
rs1009903253 Chr.4: 145727095-145727095 on Build GRCh38
rs761124820 Chr.4: 145726874-145726874 on Build GRCh38
rs978524690 Chr.4: 145727245-145727245 on Build GRCh38
rs1001303632 Chr.4: 145727029-145727029 on Build GRCh38
rs547639902 Chr.4: 145726930-145726930 on Build GRCh38
rs747348970 Chr.4: 145726980-145726980 on Build GRCh38
rs1014110541 Chr.4: 145727214-145727214 on Build GRCh38
rs780056782 Chr.4: 145727090-145727090 on Build GRCh38
rs777720642 Chr.4: 145726926-145726926 on Build GRCh38
rs771818362 Chr.4: 145726976-145726976 on Build GRCh38
rs776595771 Chr.4: 145726891-145726891 on Build GRCh38
rs188808477 Chr.4: 145727242-145727242 on Build GRCh38
rs941481410 Chr.4: 145727121-145727124 on Build GRCh38
rs1047314733 Chr.4: 145726931-145726931 on Build GRCh38
rs558038796 Chr.4: 145727208-145727208 on Build GRCh38
rs79616316 Chr.4: 145727154-145727154 on Build GRCh38
rs934234157 Chr.4: 145726953-145726953 on Build GRCh38
rs373518184 Chr.4: 145726985-145726985 on Build GRCh38
rs777124988 Chr.4: 145727000-145727000 on Build GRCh38
rs1020233735 Chr.4: 145727131-145727131 on Build GRCh38
rs577835460 Chr.4: 145727232-145727232 on Build GRCh38

View Full Product Details