Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 147543388-147543922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDNRA
Entrez Gene ID
1909
Gene Name
endothelin receptor type A
Gene Aliases
ET-A, ETA, ETA-R, ETAR, ETRA, MFDA, hET-AR
UniGene
Hs.183713
Gene Chromosome Location
Chr.4: 147480917-147544954 on Build GRCh38
Transcripts
NM_001166055.1 NM_001256283.1 NM_001957.3 NR_045958.1

SNP Detail

SNP ID Chromosome Location
rs10305934 Chr.4: 147543596-147543597 on Build GRCh38
rs558342598 Chr.4: 147543533-147543533 on Build GRCh38
rs200076817 Chr.4: 147543583-147543583 on Build GRCh38
rs973059293 Chr.4: 147543729-147543729 on Build GRCh38
rs201078072 Chr.4: 147543550-147543550 on Build GRCh38
rs200450644 Chr.4: 147543855-147543855 on Build GRCh38
rs201437124 Chr.4: 147543863-147543863 on Build GRCh38
rs202015557 Chr.4: 147543566-147543566 on Build GRCh38
rs201441258 Chr.4: 147543769-147543769 on Build GRCh38
rs1049963 Chr.4: 147543695-147543695 on Build GRCh38
rs5343 Chr.4: 147543840-147543840 on Build GRCh38
rs916351230 Chr.4: 147543827-147543827 on Build GRCh38
rs10305935 Chr.4: 147543664-147543664 on Build GRCh38
rs551964029 Chr.4: 147543665-147543665 on Build GRCh38
rs5341 Chr.4: 147543447-147543447 on Build GRCh38
rs972124854 Chr.4: 147543744-147543744 on Build GRCh38
rs188239028 Chr.4: 147543601-147543601 on Build GRCh38
rs5340 Chr.4: 147543412-147543412 on Build GRCh38
rs200631888 Chr.4: 147543825-147543825 on Build GRCh38
rs541312488 Chr.4: 147543569-147543569 on Build GRCh38
rs555890843 Chr.4: 147543591-147543592 on Build GRCh38
rs10305936 Chr.4: 147543825-147543826 on Build GRCh38
rs1028393375 Chr.4: 147543660-147543660 on Build GRCh38
rs796286349 Chr.4: 147543595-147543596 on Build GRCh38
rs201293932 Chr.4: 147543827-147543827 on Build GRCh38
rs939156106 Chr.4: 147543499-147543499 on Build GRCh38
rs934435678 Chr.4: 147543469-147543469 on Build GRCh38
rs200884045 Chr.4: 147543422-147543422 on Build GRCh38
rs565472753 Chr.4: 147543668-147543668 on Build GRCh38
rs902173380 Chr.4: 147543869-147543869 on Build GRCh38
rs201358413 Chr.4: 147543426-147543426 on Build GRCh38
rs950031861 Chr.4: 147543841-147543841 on Build GRCh38
rs569551850 Chr.4: 147543618-147543618 on Build GRCh38
rs201217576 Chr.4: 147543588-147543588 on Build GRCh38
rs763312481 Chr.4: 147543642-147543642 on Build GRCh38
rs909839858 Chr.4: 147543720-147543721 on Build GRCh38
rs5342 Chr.4: 147543619-147543619 on Build GRCh38
rs200153547 Chr.4: 147543597-147543597 on Build GRCh38
rs893999103 Chr.4: 147543542-147543542 on Build GRCh38
rs202003918 Chr.4: 147543596-147543596 on Build GRCh38
rs761912854 Chr.4: 147543573-147543573 on Build GRCh38
rs386680600 Chr.4: 147543618-147543619 on Build GRCh38
rs148962339 Chr.4: 147543531-147543531 on Build GRCh38
rs999472892 Chr.4: 147543600-147543600 on Build GRCh38
rs962402412 Chr.4: 147543721-147543721 on Build GRCh38
rs200121333 Chr.4: 147543539-147543539 on Build GRCh38
rs200955775 Chr.4: 147543479-147543479 on Build GRCh38
rs1054795653 Chr.4: 147543521-147543521 on Build GRCh38
rs762453267 Chr.4: 147543808-147543808 on Build GRCh38
rs199621989 Chr.4: 147543801-147543801 on Build GRCh38
rs200070806 Chr.4: 147543887-147543887 on Build GRCh38
rs951893858 Chr.4: 147543802-147543802 on Build GRCh38

View Full Product Details