Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 147874841-147875359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP10
Entrez Gene ID
79658
Gene Name
Rho GTPase activating protein 10
Gene Aliases
GRAF2, PS-GAP, PSGAP
UniGene
Hs.368631
Gene Chromosome Location
Chr.4: 147732063-148072781 on Build GRCh38
Transcripts
NM_024605.3 XM_005263215.3 XM_017008602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1051870 c.713G>A p.R238Q Chr.4: 147875031-147875031 on Build GRCh38
1241997 c.805G>A p.E269K Chr.4: 147875123-147875123 on Build GRCh38
1051871 c.785G>T p.R262L Chr.4: 147875103-147875103 on Build GRCh38
6099168 c.749T>C p.M250T Chr.4: 147875067-147875067 on Build GRCh38
1695058 c.769C>T p.P257S Chr.4: 147875087-147875087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772058840 Chr.4: 147875012-147875012 on Build GRCh38
rs200063962 Chr.4: 147875184-147875184 on Build GRCh38
rs533776222 Chr.4: 147875273-147875273 on Build GRCh38
rs901492827 Chr.4: 147875050-147875050 on Build GRCh38
rs565059906 Chr.4: 147875251-147875251 on Build GRCh38
rs368662689 Chr.4: 147874905-147874905 on Build GRCh38
rs928627388 Chr.4: 147874972-147874972 on Build GRCh38
rs772598386 Chr.4: 147875097-147875097 on Build GRCh38
rs371591903 Chr.4: 147875062-147875062 on Build GRCh38
rs113611599 Chr.4: 147875047-147875047 on Build GRCh38
rs960286831 Chr.4: 147875218-147875218 on Build GRCh38
rs747104670 Chr.4: 147875103-147875103 on Build GRCh38
rs150957621 Chr.4: 147875002-147875002 on Build GRCh38
rs913284261 Chr.4: 147875287-147875287 on Build GRCh38
rs756910242 Chr.4: 147875181-147875181 on Build GRCh38
rs762116845 Chr.4: 147875127-147875127 on Build GRCh38
rs760183525 Chr.4: 147875176-147875176 on Build GRCh38
rs752574330 Chr.4: 147875175-147875175 on Build GRCh38
rs564186201 Chr.4: 147875014-147875014 on Build GRCh38
rs978938292 Chr.4: 147875299-147875299 on Build GRCh38
rs538578376 Chr.4: 147875102-147875102 on Build GRCh38
rs1035706165 Chr.4: 147875208-147875208 on Build GRCh38
rs778309775 Chr.4: 147875166-147875166 on Build GRCh38
rs763589122 Chr.4: 147875177-147875177 on Build GRCh38
rs774166818 Chr.4: 147875136-147875136 on Build GRCh38
rs954990555 Chr.4: 147875044-147875044 on Build GRCh38
rs142531496 Chr.4: 147875087-147875087 on Build GRCh38
rs996772704 Chr.4: 147875255-147875255 on Build GRCh38
rs745790711 Chr.4: 147874998-147874998 on Build GRCh38
rs771954703 Chr.4: 147875066-147875069 on Build GRCh38
rs483352827 Chr.4: 147875034-147875034 on Build GRCh38
rs892920534 Chr.4: 147875245-147875245 on Build GRCh38
rs756716146 Chr.4: 147875105-147875105 on Build GRCh38
rs908268011 Chr.4: 147875056-147875056 on Build GRCh38
rs913418911 Chr.4: 147875006-147875006 on Build GRCh38
rs755467012 Chr.4: 147875026-147875026 on Build GRCh38
rs766978867 Chr.4: 147875037-147875037 on Build GRCh38
rs755721833 Chr.4: 147874989-147874989 on Build GRCh38
rs777372402 Chr.4: 147874991-147874991 on Build GRCh38
rs142876032 Chr.4: 147875024-147875024 on Build GRCh38
rs368101248 Chr.4: 147875195-147875195 on Build GRCh38
rs750827875 Chr.4: 147875187-147875187 on Build GRCh38
rs915927270 Chr.4: 147875123-147875123 on Build GRCh38
rs569429656 Chr.4: 147875090-147875090 on Build GRCh38
rs930913612 Chr.4: 147875117-147875117 on Build GRCh38
rs752285517 Chr.4: 147875046-147875046 on Build GRCh38
rs374150548 Chr.4: 147875122-147875122 on Build GRCh38
rs540518228 Chr.4: 147874901-147874901 on Build GRCh38
rs1045819181 Chr.4: 147875174-147875174 on Build GRCh38
rs483352830 Chr.4: 147875140-147875140 on Build GRCh38
rs749700146 Chr.4: 147874998-147874998 on Build GRCh38
rs7663294 Chr.4: 147875313-147875313 on Build GRCh38
rs771271335 Chr.4: 147875000-147875000 on Build GRCh38
rs930672980 Chr.4: 147874921-147874921 on Build GRCh38
rs572146689 Chr.4: 147874884-147874884 on Build GRCh38
rs549588917 Chr.4: 147875083-147875083 on Build GRCh38
rs962546015 Chr.4: 147875089-147875089 on Build GRCh38
rs767514014 Chr.4: 147875157-147875157 on Build GRCh38
rs770683315 Chr.4: 147875134-147875134 on Build GRCh38
rs997288392 Chr.4: 147875066-147875066 on Build GRCh38
rs1007879799 Chr.4: 147875338-147875338 on Build GRCh38
rs894442989 Chr.4: 147874964-147874964 on Build GRCh38
rs968765150 Chr.4: 147875290-147875290 on Build GRCh38
rs1028214830 Chr.4: 147875262-147875262 on Build GRCh38
rs142593681 Chr.4: 147874877-147874877 on Build GRCh38
rs546648027 Chr.4: 147875030-147875030 on Build GRCh38
rs746320175 Chr.4: 147875004-147875004 on Build GRCh38
rs942994858 Chr.4: 147875319-147875319 on Build GRCh38
rs1055720704 Chr.4: 147874950-147874950 on Build GRCh38
rs1014302106 Chr.4: 147874994-147874994 on Build GRCh38
rs776974905 Chr.4: 147875025-147875025 on Build GRCh38
rs753545822 Chr.4: 147875180-147875180 on Build GRCh38
rs759498019 Chr.4: 147875031-147875031 on Build GRCh38
rs988856649 Chr.4: 147875252-147875252 on Build GRCh38

View Full Product Details