Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 151577757-151578266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM160A1
Entrez Gene ID
729830
Gene Name
family with sequence similarity 160 member A1
Gene Aliases
-
UniGene
Hs.633810
Gene Chromosome Location
Chr.4: 151409143-151670503 on Build GRCh38
Transcripts
NM_001109977.1 XM_005263199.1 XM_011532220.1 XM_011532221.2 XM_011532222.2 XM_011532223.2 XM_011532224.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5925109 c.565G>A p.D189N Chr.4: 151577909-151577909 on Build GRCh38
6204968 c.633A>G p.V211V Chr.4: 151577977-151577977 on Build GRCh38
5975023 c.626G>T p.G209V Chr.4: 151577970-151577970 on Build GRCh38
1485734 c.684G>A p.E228E Chr.4: 151578028-151578028 on Build GRCh38
5729221 c.729T>C p.C243C Chr.4: 151578073-151578073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758822978 Chr.4: 151578123-151578123 on Build GRCh38
rs543303426 Chr.4: 151577793-151577793 on Build GRCh38
rs1028774652 Chr.4: 151577810-151577810 on Build GRCh38
rs202028349 Chr.4: 151578043-151578043 on Build GRCh38
rs185952321 Chr.4: 151577991-151577991 on Build GRCh38
rs772660365 Chr.4: 151577933-151577933 on Build GRCh38
rs908964755 Chr.4: 151578228-151578228 on Build GRCh38
rs747116506 Chr.4: 151577862-151577862 on Build GRCh38
rs977002138 Chr.4: 151578128-151578128 on Build GRCh38
rs767498380 Chr.4: 151578011-151578011 on Build GRCh38
rs952550563 Chr.4: 151578238-151578238 on Build GRCh38
rs934883582 Chr.4: 151577998-151577998 on Build GRCh38
rs750867683 Chr.4: 151578107-151578107 on Build GRCh38
rs889790060 Chr.4: 151577812-151577812 on Build GRCh38
rs867037082 Chr.4: 151577843-151577843 on Build GRCh38
rs756998700 Chr.4: 151578072-151578072 on Build GRCh38
rs541204183 Chr.4: 151577910-151577910 on Build GRCh38
rs150755896 Chr.4: 151577874-151577874 on Build GRCh38
rs765035756 Chr.4: 151578152-151578152 on Build GRCh38
rs889832606 Chr.4: 151578134-151578134 on Build GRCh38
rs968789538 Chr.4: 151578055-151578055 on Build GRCh38
rs1053311877 Chr.4: 151578032-151578032 on Build GRCh38
rs972559005 Chr.4: 151577879-151577879 on Build GRCh38
rs560394823 Chr.4: 151578135-151578135 on Build GRCh38
rs1047972070 Chr.4: 151577905-151577905 on Build GRCh38
rs762112920 Chr.4: 151577917-151577917 on Build GRCh38
rs769326756 Chr.4: 151577903-151577903 on Build GRCh38
rs965333577 Chr.4: 151578115-151578115 on Build GRCh38
rs947491028 Chr.4: 151578062-151578062 on Build GRCh38
rs776587071 Chr.4: 151577927-151577927 on Build GRCh38
rs545255097 Chr.4: 151577957-151577957 on Build GRCh38
rs1010062547 Chr.4: 151578052-151578052 on Build GRCh38
rs1019532849 Chr.4: 151577836-151577836 on Build GRCh38
rs752671972 Chr.4: 151578037-151578037 on Build GRCh38
rs951674192 Chr.4: 151578169-151578169 on Build GRCh38
rs868550454 Chr.4: 151577918-151577918 on Build GRCh38
rs563978251 Chr.4: 151578160-151578160 on Build GRCh38
rs905792693 Chr.4: 151577964-151577964 on Build GRCh38
rs138026287 Chr.4: 151578053-151578053 on Build GRCh38
rs897102085 Chr.4: 151578201-151578201 on Build GRCh38
rs555525555 Chr.4: 151577827-151577827 on Build GRCh38
rs867645841 Chr.4: 151577970-151577970 on Build GRCh38
rs879818277 Chr.4: 151578084-151578084 on Build GRCh38
rs762607344 Chr.4: 151577988-151577988 on Build GRCh38
rs1026755479 Chr.4: 151577890-151577890 on Build GRCh38
rs1041420084 Chr.4: 151578194-151578194 on Build GRCh38
rs1011656579 Chr.4: 151577801-151577801 on Build GRCh38
rs769958379 Chr.4: 151577960-151577960 on Build GRCh38
rs976908771 Chr.4: 151577791-151577791 on Build GRCh38
rs778678888 Chr.4: 151578076-151578076 on Build GRCh38
rs77297019 Chr.4: 151578233-151578233 on Build GRCh38
rs531270208 Chr.4: 151578208-151578208 on Build GRCh38
rs760458965 Chr.4: 151578049-151578049 on Build GRCh38
rs1049837743 Chr.4: 151578132-151578132 on Build GRCh38
rs780685999 Chr.4: 151577814-151577814 on Build GRCh38
rs376159656 Chr.4: 151578058-151578058 on Build GRCh38
rs559765365 Chr.4: 151577990-151577990 on Build GRCh38
rs775648585 Chr.4: 151577934-151577936 on Build GRCh38
rs746193101 Chr.4: 151577788-151577788 on Build GRCh38

View Full Product Details