Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 151638537-151639076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM160A1
Entrez Gene ID
729830
Gene Name
family with sequence similarity 160 member A1
Gene Aliases
-
UniGene
Hs.633810
Gene Chromosome Location
Chr.4: 151409143-151670503 on Build GRCh38
Transcripts
NM_001109977.1 XM_005263199.1 XM_011532220.1 XM_011532221.2 XM_011532222.2 XM_011532223.2 XM_011532224.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6325745 c.1182C>T p.L394L Chr.4: 151638712-151638712 on Build GRCh38
1594364 c.1186G>T p.G396C Chr.4: 151638716-151638716 on Build GRCh38
7367104 c.1184T>C p.I395T Chr.4: 151638714-151638714 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs934955312 Chr.4: 151638960-151638960 on Build GRCh38
rs774737019 Chr.4: 151638660-151638661 on Build GRCh38
rs759159827 Chr.4: 151638633-151638633 on Build GRCh38
rs750614722 Chr.4: 151638773-151638773 on Build GRCh38
rs11450944 Chr.4: 151638565-151638566 on Build GRCh38
rs375873845 Chr.4: 151638701-151638701 on Build GRCh38
rs779263864 Chr.4: 151638762-151638762 on Build GRCh38
rs924836486 Chr.4: 151638906-151638906 on Build GRCh38
rs185879278 Chr.4: 151638721-151638721 on Build GRCh38
rs538426374 Chr.4: 151638714-151638714 on Build GRCh38
rs934991644 Chr.4: 151638611-151638611 on Build GRCh38
rs765388862 Chr.4: 151638777-151638778 on Build GRCh38
rs781159818 Chr.4: 151638791-151638791 on Build GRCh38
rs554228682 Chr.4: 151638874-151638874 on Build GRCh38
rs144886285 Chr.4: 151638588-151638589 on Build GRCh38
rs924893829 Chr.4: 151638608-151638608 on Build GRCh38
rs371178348 Chr.4: 151638878-151638878 on Build GRCh38
rs1013348622 Chr.4: 151638649-151638649 on Build GRCh38
rs558342704 Chr.4: 151638984-151638984 on Build GRCh38
rs753966721 Chr.4: 151638723-151638723 on Build GRCh38
rs149114647 Chr.4: 151638973-151638973 on Build GRCh38
rs754780087 Chr.4: 151638989-151638989 on Build GRCh38
rs1030346529 Chr.4: 151638852-151638852 on Build GRCh38
rs961974566 Chr.4: 151638724-151638724 on Build GRCh38
rs548135019 Chr.4: 151638794-151638800 on Build GRCh38
rs910691655 Chr.4: 151638875-151638875 on Build GRCh38
rs917607512 Chr.4: 151639022-151639022 on Build GRCh38
rs867512229 Chr.4: 151638688-151638688 on Build GRCh38
rs1015938069 Chr.4: 151638916-151638916 on Build GRCh38
rs978966637 Chr.4: 151638891-151638891 on Build GRCh38
rs181630452 Chr.4: 151638653-151638653 on Build GRCh38
rs896209777 Chr.4: 151638634-151638634 on Build GRCh38
rs954935318 Chr.4: 151638853-151638853 on Build GRCh38
rs993893614 Chr.4: 151638772-151638772 on Build GRCh38
rs954175830 Chr.4: 151638702-151638702 on Build GRCh38
rs1044852280 Chr.4: 151638659-151638659 on Build GRCh38
rs934137505 Chr.4: 151638728-151638728 on Build GRCh38
rs374097630 Chr.4: 151639009-151639009 on Build GRCh38
rs1043338898 Chr.4: 151638577-151638577 on Build GRCh38
rs748198293 Chr.4: 151638693-151638693 on Build GRCh38
rs552827825 Chr.4: 151638613-151638613 on Build GRCh38
rs568660066 Chr.4: 151638731-151638731 on Build GRCh38
rs758590080 Chr.4: 151638784-151638784 on Build GRCh38
rs536287650 Chr.4: 151638788-151638788 on Build GRCh38

View Full Product Details