Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 151649345-151649860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM160A1
Entrez Gene ID
729830
Gene Name
family with sequence similarity 160 member A1
Gene Aliases
-
UniGene
Hs.633810
Gene Chromosome Location
Chr.4: 151409143-151670503 on Build GRCh38
Transcripts
NM_001109977.1 XM_005263199.1 XM_011532220.1 XM_011532221.2 XM_011532222.2 XM_011532223.2 XM_011532224.2 XM_017008583.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1427559 c.1640C>T p.S547L Chr.4: 151649681-151649681 on Build GRCh38
1427557 c.1588C>A p.P530T Chr.4: 151649629-151649629 on Build GRCh38
3714817 c.1426G>T p.E476* Chr.4: 151649467-151649467 on Build GRCh38
4973913 c.1537C>T p.R513C Chr.4: 151649578-151649578 on Build GRCh38
3696475 c.1430G>A p.R477Q Chr.4: 151649471-151649471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773209777 Chr.4: 151649547-151649547 on Build GRCh38
rs906622352 Chr.4: 151649634-151649634 on Build GRCh38
rs1041546407 Chr.4: 151649399-151649399 on Build GRCh38
rs565972508 Chr.4: 151649454-151649454 on Build GRCh38
rs1041167722 Chr.4: 151649682-151649682 on Build GRCh38
rs755190327 Chr.4: 151649599-151649599 on Build GRCh38
rs995549055 Chr.4: 151649440-151649440 on Build GRCh38
rs368249366 Chr.4: 151649492-151649492 on Build GRCh38
rs190685739 Chr.4: 151649635-151649635 on Build GRCh38
rs757898713 Chr.4: 151649661-151649661 on Build GRCh38
rs181059274 Chr.4: 151649706-151649706 on Build GRCh38
rs567628337 Chr.4: 151649705-151649705 on Build GRCh38
rs762249220 Chr.4: 151649539-151649539 on Build GRCh38
rs558082181 Chr.4: 151649471-151649471 on Build GRCh38
rs549725285 Chr.4: 151649382-151649382 on Build GRCh38
rs534754196 Chr.4: 151649817-151649817 on Build GRCh38
rs372351230 Chr.4: 151649513-151649513 on Build GRCh38
rs376523990 Chr.4: 151649678-151649678 on Build GRCh38
rs766347001 Chr.4: 151649550-151649550 on Build GRCh38
rs1055765574 Chr.4: 151649412-151649412 on Build GRCh38
rs367927475 Chr.4: 151649622-151649622 on Build GRCh38
rs767813801 Chr.4: 151649818-151649818 on Build GRCh38
rs868574661 Chr.4: 151649512-151649512 on Build GRCh38
rs548449268 Chr.4: 151649578-151649578 on Build GRCh38
rs553292974 Chr.4: 151649549-151649549 on Build GRCh38
rs372152751 Chr.4: 151649525-151649525 on Build GRCh38
rs564075271 Chr.4: 151649660-151649660 on Build GRCh38
rs938562700 Chr.4: 151649631-151649631 on Build GRCh38
rs186402735 Chr.4: 151649794-151649794 on Build GRCh38
rs371000085 Chr.4: 151649470-151649470 on Build GRCh38
rs547762488 Chr.4: 151649402-151649402 on Build GRCh38
rs894461676 Chr.4: 151649416-151649416 on Build GRCh38
rs781115863 Chr.4: 151649494-151649494 on Build GRCh38
rs915855942 Chr.4: 151649643-151649643 on Build GRCh38
rs976451668 Chr.4: 151649584-151649584 on Build GRCh38
rs545503967 Chr.4: 151649597-151649597 on Build GRCh38
rs980639739 Chr.4: 151649788-151649788 on Build GRCh38
rs139068371 Chr.4: 151649486-151649486 on Build GRCh38
rs965190351 Chr.4: 151649695-151649695 on Build GRCh38
rs1011159528 Chr.4: 151649506-151649506 on Build GRCh38
rs967067626 Chr.4: 151649805-151649805 on Build GRCh38
rs376521688 Chr.4: 151649782-151649782 on Build GRCh38
rs751260140 Chr.4: 151649825-151649825 on Build GRCh38
rs771761564 Chr.4: 151649482-151649482 on Build GRCh38
rs909748525 Chr.4: 151649614-151649614 on Build GRCh38
rs529140364 Chr.4: 151649744-151649744 on Build GRCh38
rs757301030 Chr.4: 151649415-151649415 on Build GRCh38
rs746680259 Chr.4: 151649674-151649674 on Build GRCh38
rs144995238 Chr.4: 151649672-151649672 on Build GRCh38
rs555443576 Chr.4: 151649404-151649404 on Build GRCh38
rs541223684 Chr.4: 151649545-151649545 on Build GRCh38
rs967119864 Chr.4: 151649600-151649600 on Build GRCh38
rs929936231 Chr.4: 151649607-151649607 on Build GRCh38
rs1048491577 Chr.4: 151649448-151649448 on Build GRCh38
rs10014639 Chr.4: 151649383-151649383 on Build GRCh38
rs1048248230 Chr.4: 151649396-151649396 on Build GRCh38
rs778886880 Chr.4: 151649642-151649642 on Build GRCh38
rs751833203 Chr.4: 151649595-151649595 on Build GRCh38
rs965461953 Chr.4: 151649579-151649579 on Build GRCh38
rs186699437 Chr.4: 151649493-151649493 on Build GRCh38
rs953511032 Chr.4: 151649523-151649523 on Build GRCh38
rs1021248667 Chr.4: 151649773-151649773 on Build GRCh38
rs576478699 Chr.4: 151649474-151649474 on Build GRCh38
rs768399304 Chr.4: 151649502-151649502 on Build GRCh38
rs763507820 Chr.4: 151649698-151649698 on Build GRCh38
rs945870031 Chr.4: 151649395-151649395 on Build GRCh38
rs766848601 Chr.4: 151649712-151649712 on Build GRCh38
rs866465776 Chr.4: 151649575-151649575 on Build GRCh38
rs199603138 Chr.4: 151649681-151649681 on Build GRCh38
rs371778098 Chr.4: 151649623-151649623 on Build GRCh38
rs928466686 Chr.4: 151649612-151649612 on Build GRCh38
rs535625490 Chr.4: 151649401-151649401 on Build GRCh38
rs866837668 Chr.4: 151649663-151649663 on Build GRCh38
rs181358812 Chr.4: 151649490-151649490 on Build GRCh38
rs766577986 Chr.4: 151649743-151649743 on Build GRCh38
rs748517622 Chr.4: 151649697-151649697 on Build GRCh38
rs867904600 Chr.4: 151649487-151649487 on Build GRCh38
rs375182034 Chr.4: 151649608-151649608 on Build GRCh38
rs745351760 Chr.4: 151649481-151649481 on Build GRCh38
rs1022564763 Chr.4: 151649673-151649673 on Build GRCh38
rs1022493461 Chr.4: 151649517-151649517 on Build GRCh38
rs746499636 Chr.4: 151649501-151649501 on Build GRCh38

View Full Product Details