Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154235056-154235553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCHS2
Entrez Gene ID
54798
Gene Name
dachsous cadherin-related 2
Gene Aliases
CDH27, CDHJ, CDHR7, PCDH23, PCDHJ
UniGene
Hs.655664
Gene Chromosome Location
Chr.4: 154234375-154491778 on Build GRCh38
Transcripts
NM_017639.3 XM_011532045.1 XM_011532046.2 XM_011532049.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1319369 c.7841C>T p.P2614L Chr.4: 154235446-154235446 on Build GRCh38
6469350 c.8058G>A p.Q2686Q Chr.4: 154235229-154235229 on Build GRCh38
1203115 c.7808G>A p.C2603Y Chr.4: 154235479-154235479 on Build GRCh38
288546 c.7842G>A p.P2614P Chr.4: 154235445-154235445 on Build GRCh38
7363829 c.8192C>T p.T2731I Chr.4: 154235095-154235095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs267600050 Chr.4: 154235471-154235471 on Build GRCh38
rs947135427 Chr.4: 154235435-154235435 on Build GRCh38
rs780963230 Chr.4: 154235414-154235414 on Build GRCh38
rs867486365 Chr.4: 154235326-154235326 on Build GRCh38
rs760236185 Chr.4: 154235080-154235080 on Build GRCh38
rs778338448 Chr.4: 154235441-154235441 on Build GRCh38
rs895550623 Chr.4: 154235226-154235226 on Build GRCh38
rs765345306 Chr.4: 154235475-154235475 on Build GRCh38
rs747787568 Chr.4: 154235271-154235271 on Build GRCh38
rs764792734 Chr.4: 154235302-154235303 on Build GRCh38
rs760910436 Chr.4: 154235357-154235357 on Build GRCh38
rs756459080 Chr.4: 154235235-154235235 on Build GRCh38
rs769504815 Chr.4: 154235274-154235274 on Build GRCh38
rs775937198 Chr.4: 154235460-154235460 on Build GRCh38
rs61741046 Chr.4: 154235390-154235390 on Build GRCh38
rs201121579 Chr.4: 154235078-154235078 on Build GRCh38
rs772643568 Chr.4: 154235281-154235281 on Build GRCh38
rs151035309 Chr.4: 154235484-154235484 on Build GRCh38
rs143523763 Chr.4: 154235210-154235210 on Build GRCh38
rs200455137 Chr.4: 154235232-154235232 on Build GRCh38
rs777586912 Chr.4: 154235500-154235500 on Build GRCh38
rs201033133 Chr.4: 154235145-154235145 on Build GRCh38
rs771298095 Chr.4: 154235307-154235307 on Build GRCh38
rs759452619 Chr.4: 154235481-154235481 on Build GRCh38
rs535611182 Chr.4: 154235195-154235195 on Build GRCh38
rs749983277 Chr.4: 154235398-154235399 on Build GRCh38
rs61746132 Chr.4: 154235260-154235260 on Build GRCh38
rs190682484 Chr.4: 154235170-154235170 on Build GRCh38
rs781493961 Chr.4: 154235153-154235153 on Build GRCh38
rs776377183 Chr.4: 154235192-154235192 on Build GRCh38
rs750896876 Chr.4: 154235121-154235121 on Build GRCh38
rs377694605 Chr.4: 154235369-154235369 on Build GRCh38
rs1043329599 Chr.4: 154235515-154235515 on Build GRCh38
rs745784975 Chr.4: 154235167-154235167 on Build GRCh38
rs558435505 Chr.4: 154235356-154235356 on Build GRCh38
rs755444808 Chr.4: 154235472-154235473 on Build GRCh38
rs758208401 Chr.4: 154235246-154235246 on Build GRCh38
rs374185470 Chr.4: 154235319-154235319 on Build GRCh38
rs766914817 Chr.4: 154235227-154235227 on Build GRCh38
rs767904791 Chr.4: 154235347-154235347 on Build GRCh38
rs745888948 Chr.4: 154235112-154235112 on Build GRCh38
rs147174513 Chr.4: 154235233-154235233 on Build GRCh38
rs754770354 Chr.4: 154235259-154235259 on Build GRCh38
rs778416571 Chr.4: 154235521-154235521 on Build GRCh38
rs1012908503 Chr.4: 154235101-154235101 on Build GRCh38
rs377590785 Chr.4: 154235237-154235237 on Build GRCh38
rs149548848 Chr.4: 154235446-154235446 on Build GRCh38
rs531958470 Chr.4: 154235149-154235149 on Build GRCh38
rs763501582 Chr.4: 154235125-154235125 on Build GRCh38
rs768356361 Chr.4: 154235090-154235090 on Build GRCh38
rs768958497 Chr.4: 154235466-154235466 on Build GRCh38
rs199868518 Chr.4: 154235429-154235429 on Build GRCh38
rs752779419 Chr.4: 154235154-154235154 on Build GRCh38
rs748810414 Chr.4: 154235298-154235298 on Build GRCh38
rs774521920 Chr.4: 154235228-154235228 on Build GRCh38
rs772655193 Chr.4: 154235526-154235526 on Build GRCh38
rs757388888 Chr.4: 154235380-154235380 on Build GRCh38
rs777426357 Chr.4: 154235431-154235431 on Build GRCh38
rs756295492 Chr.4: 154235161-154235161 on Build GRCh38
rs761334454 Chr.4: 154235100-154235100 on Build GRCh38
rs763448372 Chr.4: 154235219-154235219 on Build GRCh38
rs138106573 Chr.4: 154235247-154235247 on Build GRCh38
rs865847267 Chr.4: 154235348-154235348 on Build GRCh38
rs753968095 Chr.4: 154235244-154235244 on Build GRCh38
rs200990565 Chr.4: 154235229-154235229 on Build GRCh38
rs140493376 Chr.4: 154235253-154235253 on Build GRCh38
rs779644338 Chr.4: 154235179-154235179 on Build GRCh38
rs756927685 Chr.4: 154235218-154235221 on Build GRCh38
rs770448499 Chr.4: 154235440-154235440 on Build GRCh38
rs61738813 Chr.4: 154235095-154235095 on Build GRCh38
rs760003334 Chr.4: 154235331-154235331 on Build GRCh38
rs753511759 Chr.4: 154235505-154235505 on Build GRCh38
rs61741036 Chr.4: 154235445-154235445 on Build GRCh38
rs750536754 Chr.4: 154235476-154235476 on Build GRCh38
rs778042900 Chr.4: 154235164-154235164 on Build GRCh38
rs761139903 Chr.4: 154235463-154235463 on Build GRCh38
rs903090049 Chr.4: 154235516-154235516 on Build GRCh38
rs373906914 Chr.4: 154235483-154235483 on Build GRCh38
rs781634290 Chr.4: 154235518-154235518 on Build GRCh38
rs150299118 Chr.4: 154235079-154235079 on Build GRCh38
rs375679010 Chr.4: 154235411-154235411 on Build GRCh38
rs201468562 Chr.4: 154235250-154235250 on Build GRCh38
rs751490474 Chr.4: 154235407-154235407 on Build GRCh38
rs74491447 Chr.4: 154235522-154235522 on Build GRCh38
rs748710721 Chr.4: 154235438-154235438 on Build GRCh38
rs761991239 Chr.4: 154235472-154235472 on Build GRCh38
rs752028220 Chr.4: 154235139-154235139 on Build GRCh38
rs746831021 Chr.4: 154235186-154235186 on Build GRCh38
rs77305317 Chr.4: 154235358-154235358 on Build GRCh38
rs747943618 Chr.4: 154235425-154235425 on Build GRCh38
rs568618708 Chr.4: 154235187-154235187 on Build GRCh38
rs774958409 Chr.4: 154235318-154235318 on Build GRCh38

View Full Product Details