Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154235793-154236338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCHS2
Entrez Gene ID
54798
Gene Name
dachsous cadherin-related 2
Gene Aliases
CDH27, CDHJ, CDHR7, PCDH23, PCDHJ
UniGene
Hs.655664
Gene Chromosome Location
Chr.4: 154234375-154491778 on Build GRCh38
Transcripts
NM_017639.3 XM_011532045.1 XM_011532046.2 XM_011532049.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1052292 c.7246G>T p.E2416* Chr.4: 154236041-154236041 on Build GRCh38
1203111 c.7255G>T p.D2419Y Chr.4: 154236032-154236032 on Build GRCh38
6217863 c.7121C>T p.P2374L Chr.4: 154236166-154236166 on Build GRCh38
3674119 c.7261T>C p.F2421L Chr.4: 154236026-154236026 on Build GRCh38
6640773 c.7240G>T p.D2414Y Chr.4: 154236047-154236047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372086117 Chr.4: 154236228-154236228 on Build GRCh38
rs1030289940 Chr.4: 154236021-154236021 on Build GRCh38
rs770829294 Chr.4: 154236086-154236086 on Build GRCh38
rs1038293144 Chr.4: 154236302-154236302 on Build GRCh38
rs757002929 Chr.4: 154236075-154236075 on Build GRCh38
rs755310867 Chr.4: 154236283-154236283 on Build GRCh38
rs774544507 Chr.4: 154236050-154236050 on Build GRCh38
rs746126538 Chr.4: 154235882-154235882 on Build GRCh38
rs752832186 Chr.4: 154236293-154236293 on Build GRCh38
rs769774264 Chr.4: 154235906-154235906 on Build GRCh38
rs758712894 Chr.4: 154236029-154236029 on Build GRCh38
rs749226677 Chr.4: 154236231-154236231 on Build GRCh38
rs200042662 Chr.4: 154236008-154236008 on Build GRCh38
rs576097143 Chr.4: 154236253-154236253 on Build GRCh38
rs766143078 Chr.4: 154235828-154235828 on Build GRCh38
rs771114281 Chr.4: 154235991-154235992 on Build GRCh38
rs374382000 Chr.4: 154236043-154236043 on Build GRCh38
rs761024588 Chr.4: 154235985-154235985 on Build GRCh38
rs948460639 Chr.4: 154235877-154235877 on Build GRCh38
rs750803132 Chr.4: 154236192-154236192 on Build GRCh38
rs773400420 Chr.4: 154236269-154236269 on Build GRCh38
rs772019908 Chr.4: 154235887-154235887 on Build GRCh38
rs764602985 Chr.4: 154235999-154235999 on Build GRCh38
rs1044202976 Chr.4: 154235886-154235886 on Build GRCh38
rs774809783 Chr.4: 154236111-154236111 on Build GRCh38
rs572143137 Chr.4: 154236154-154236154 on Build GRCh38
rs779930327 Chr.4: 154236058-154236058 on Build GRCh38
rs1023004902 Chr.4: 154236052-154236052 on Build GRCh38
rs370459330 Chr.4: 154235890-154235890 on Build GRCh38
rs1037859671 Chr.4: 154235955-154235955 on Build GRCh38
rs61743677 Chr.4: 154235863-154235863 on Build GRCh38
rs749518525 Chr.4: 154235961-154235964 on Build GRCh38
rs777884817 Chr.4: 154236080-154236080 on Build GRCh38
rs924369995 Chr.4: 154236085-154236085 on Build GRCh38
rs142781136 Chr.4: 154236237-154236237 on Build GRCh38
rs765745460 Chr.4: 154236261-154236261 on Build GRCh38
rs191279971 Chr.4: 154236042-154236042 on Build GRCh38
rs747125890 Chr.4: 154236066-154236066 on Build GRCh38
rs537787483 Chr.4: 154235866-154235866 on Build GRCh38
rs1057221009 Chr.4: 154235864-154235864 on Build GRCh38
rs751064423 Chr.4: 154235829-154235829 on Build GRCh38
rs749279867 Chr.4: 154236084-154236084 on Build GRCh38
rs772304810 Chr.4: 154236158-154236159 on Build GRCh38
rs759283350 Chr.4: 154235971-154235971 on Build GRCh38
rs1015675487 Chr.4: 154235880-154235880 on Build GRCh38
rs143419229 Chr.4: 154235837-154235837 on Build GRCh38
rs774348520 Chr.4: 154236090-154236090 on Build GRCh38
rs956155063 Chr.4: 154236221-154236221 on Build GRCh38
rs1007504979 Chr.4: 154236115-154236115 on Build GRCh38
rs765140906 Chr.4: 154236126-154236126 on Build GRCh38
rs770667835 Chr.4: 154235954-154235954 on Build GRCh38
rs762362898 Chr.4: 154236174-154236174 on Build GRCh38
rs774118820 Chr.4: 154235956-154235956 on Build GRCh38
rs781472664 Chr.4: 154236230-154236230 on Build GRCh38
rs748029791 Chr.4: 154235892-154235892 on Build GRCh38
rs375918917 Chr.4: 154236248-154236248 on Build GRCh38
rs924554739 Chr.4: 154235827-154235827 on Build GRCh38
rs763406614 Chr.4: 154236274-154236274 on Build GRCh38
rs145835627 Chr.4: 154235969-154235969 on Build GRCh38
rs979017766 Chr.4: 154236166-154236166 on Build GRCh38
rs750308600 Chr.4: 154236163-154236163 on Build GRCh38
rs202103169 Chr.4: 154235984-154235984 on Build GRCh38
rs572379226 Chr.4: 154236063-154236063 on Build GRCh38
rs771764741 Chr.4: 154236107-154236107 on Build GRCh38
rs551336329 Chr.4: 154235847-154235847 on Build GRCh38
rs114015145 Chr.4: 154236252-154236252 on Build GRCh38
rs753125225 Chr.4: 154235855-154235855 on Build GRCh38
rs186377292 Chr.4: 154235821-154235821 on Build GRCh38
rs755055599 Chr.4: 154236226-154236226 on Build GRCh38
rs201072592 Chr.4: 154235920-154235920 on Build GRCh38
rs776188265 Chr.4: 154236258-154236258 on Build GRCh38
rs141998004 Chr.4: 154235871-154235871 on Build GRCh38
rs138746850 Chr.4: 154236013-154236013 on Build GRCh38
rs917040636 Chr.4: 154236183-154236183 on Build GRCh38
rs948563805 Chr.4: 154236235-154236235 on Build GRCh38
rs775514206 Chr.4: 154236202-154236202 on Build GRCh38
rs574592615 Chr.4: 154235983-154235983 on Build GRCh38
rs758491051 Chr.4: 154236193-154236193 on Build GRCh38
rs745905752 Chr.4: 154236101-154236101 on Build GRCh38
rs3749471 Chr.4: 154235986-154235986 on Build GRCh38
rs968763447 Chr.4: 154236157-154236157 on Build GRCh38
rs762864664 Chr.4: 154235929-154235929 on Build GRCh38
rs771580195 Chr.4: 154236239-154236239 on Build GRCh38
rs370450804 Chr.4: 154236194-154236194 on Build GRCh38
rs367885409 Chr.4: 154236172-154236172 on Build GRCh38
rs765579760 Chr.4: 154236187-154236187 on Build GRCh38
rs750629300 Chr.4: 154236025-154236025 on Build GRCh38
rs756705093 Chr.4: 154235856-154235856 on Build GRCh38
rs182645950 Chr.4: 154236260-154236260 on Build GRCh38
rs754157385 Chr.4: 154236007-154236007 on Build GRCh38
rs751785249 Chr.4: 154236275-154236275 on Build GRCh38
rs759109485 Chr.4: 154236091-154236091 on Build GRCh38
rs762627103 Chr.4: 154235822-154235822 on Build GRCh38
rs199850135 Chr.4: 154235870-154235870 on Build GRCh38
rs1029813270 Chr.4: 154236006-154236006 on Build GRCh38
rs112424317 Chr.4: 154236104-154236104 on Build GRCh38
rs751554085 Chr.4: 154236203-154236203 on Build GRCh38

View Full Product Details