Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154566381-154566882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGB
Entrez Gene ID
2244
Gene Name
fibrinogen beta chain
Gene Aliases
HEL-S-78p
UniGene
Hs.300774
Gene Chromosome Location
Chr.4: 154562980-154572763 on Build GRCh38
Transcripts
NM_001184741.1 NM_005141.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4122944 c.370A>G p.R124G Chr.4: 154566552-154566552 on Build GRCh38
311170 c.479A>G p.K160R Chr.4: 154566661-154566661 on Build GRCh38
6663346 c.382G>T p.D128Y Chr.4: 154566564-154566564 on Build GRCh38
5731977 c.385G>A p.E129K Chr.4: 154566567-154566567 on Build GRCh38
5931486 c.326G>A p.G109E Chr.4: 154566508-154566508 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577497579 Chr.4: 154566792-154566792 on Build GRCh38
rs931648098 Chr.4: 154566722-154566722 on Build GRCh38
rs979168616 Chr.4: 154566730-154566730 on Build GRCh38
rs987818339 Chr.4: 154566758-154566758 on Build GRCh38
rs1025489049 Chr.4: 154566624-154566624 on Build GRCh38
rs898590076 Chr.4: 154566834-154566834 on Build GRCh38
rs754431566 Chr.4: 154566482-154566482 on Build GRCh38
rs766099099 Chr.4: 154566665-154566665 on Build GRCh38
rs749412950 Chr.4: 154566579-154566579 on Build GRCh38
rs905998442 Chr.4: 154566794-154566794 on Build GRCh38
rs544183073 Chr.4: 154566807-154566807 on Build GRCh38
rs769014989 Chr.4: 154566757-154566757 on Build GRCh38
rs776510812 Chr.4: 154566628-154566628 on Build GRCh38
rs750008000 Chr.4: 154566509-154566509 on Build GRCh38
rs761544987 Chr.4: 154566494-154566494 on Build GRCh38
rs199768390 Chr.4: 154566533-154566533 on Build GRCh38
rs2227407 Chr.4: 154566832-154566832 on Build GRCh38
rs755412339 Chr.4: 154566539-154566539 on Build GRCh38
rs756704290 Chr.4: 154566689-154566689 on Build GRCh38
rs570183893 Chr.4: 154566545-154566545 on Build GRCh38
rs770121845 Chr.4: 154566467-154566467 on Build GRCh38
rs757471901 Chr.4: 154566473-154566473 on Build GRCh38
rs777247170 Chr.4: 154566829-154566829 on Build GRCh38
rs142641543 Chr.4: 154566573-154566573 on Build GRCh38
rs770246669 Chr.4: 154566564-154566564 on Build GRCh38
rs1045925172 Chr.4: 154566793-154566793 on Build GRCh38
rs148045912 Chr.4: 154566549-154566549 on Build GRCh38
rs774502903 Chr.4: 154566489-154566489 on Build GRCh38
rs754492004 Chr.4: 154566667-154566667 on Build GRCh38
rs751143215 Chr.4: 154566525-154566525 on Build GRCh38
rs749303315 Chr.4: 154566454-154566454 on Build GRCh38
rs2227435 Chr.4: 154566735-154566735 on Build GRCh38
rs975853322 Chr.4: 154566678-154566678 on Build GRCh38
rs758347635 Chr.4: 154566440-154566446 on Build GRCh38
rs113610714 Chr.4: 154566661-154566661 on Build GRCh38
rs375927175 Chr.4: 154566786-154566786 on Build GRCh38
rs574535925 Chr.4: 154566852-154566852 on Build GRCh38
rs534169891 Chr.4: 154566761-154566761 on Build GRCh38
rs369442578 Chr.4: 154566485-154566485 on Build GRCh38
rs113118895 Chr.4: 154566766-154566766 on Build GRCh38
rs751016082 Chr.4: 154566666-154566666 on Build GRCh38
rs753190069 Chr.4: 154566686-154566686 on Build GRCh38
rs938763487 Chr.4: 154566787-154566787 on Build GRCh38
rs199596249 Chr.4: 154566658-154566658 on Build GRCh38
rs771243649 Chr.4: 154566583-154566583 on Build GRCh38
rs1018143803 Chr.4: 154566692-154566692 on Build GRCh38
rs778269805 Chr.4: 154566696-154566696 on Build GRCh38
rs748567943 Chr.4: 154566449-154566449 on Build GRCh38
rs1012836408 Chr.4: 154566808-154566808 on Build GRCh38
rs377635985 Chr.4: 154566680-154566680 on Build GRCh38
rs758080375 Chr.4: 154566514-154566514 on Build GRCh38
rs764952511 Chr.4: 154566642-154566642 on Build GRCh38
rs772160258 Chr.4: 154566622-154566622 on Build GRCh38
rs148864798 Chr.4: 154566492-154566492 on Build GRCh38
rs761740955 Chr.4: 154566637-154566637 on Build GRCh38
rs779147226 Chr.4: 154566714-154566714 on Build GRCh38
rs907066992 Chr.4: 154566781-154566781 on Build GRCh38
rs6055 Chr.4: 154566500-154566500 on Build GRCh38
rs1027216342 Chr.4: 154566820-154566820 on Build GRCh38
rs200237413 Chr.4: 154566444-154566444 on Build GRCh38
rs780249564 Chr.4: 154566445-154566445 on Build GRCh38
rs375136916 Chr.4: 154566480-154566480 on Build GRCh38
rs774263206 Chr.4: 154566584-154566584 on Build GRCh38
rs145524939 Chr.4: 154566491-154566491 on Build GRCh38
rs111502670 Chr.4: 154566673-154566673 on Build GRCh38
rs913472540 Chr.4: 154566762-154566762 on Build GRCh38
rs746024226 Chr.4: 154566552-154566552 on Build GRCh38
rs779413540 Chr.4: 154566517-154566517 on Build GRCh38
rs995767164 Chr.4: 154566493-154566493 on Build GRCh38
rs746105693 Chr.4: 154566723-154566723 on Build GRCh38
rs762504717 Chr.4: 154566659-154566659 on Build GRCh38
rs537550350 Chr.4: 154566736-154566736 on Build GRCh38
rs867633933 Chr.4: 154566601-154566601 on Build GRCh38
rs748690444 Chr.4: 154566553-154566553 on Build GRCh38
rs759710385 Chr.4: 154566589-154566589 on Build GRCh38
rs994118230 Chr.4: 154566836-154566836 on Build GRCh38
rs749728241 Chr.4: 154566698-154566698 on Build GRCh38
rs1041760497 Chr.4: 154566431-154566431 on Build GRCh38

View Full Product Details