Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 161480915-161481455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL5
Entrez Gene ID
56884
Gene Name
follistatin like 5
Gene Aliases
-
UniGene
Hs.32452
Gene Chromosome Location
Chr.4: 161383892-162164034 on Build GRCh38
Transcripts
NM_001128427.2 NM_001128428.2 NM_020116.4 XM_011532126.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1252860 c.1535A>G p.K512R Chr.4: 161481093-161481093 on Build GRCh38
4123227 c.1580T>C p.V527A Chr.4: 161481048-161481048 on Build GRCh38
6099524 c.1585G>T p.V529L Chr.4: 161481043-161481043 on Build GRCh38
5709148 c.1573C>G p.L525V Chr.4: 161481055-161481055 on Build GRCh38
6667584 c.1515A>G p.S505S Chr.4: 161481113-161481113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753515958 Chr.4: 161481146-161481146 on Build GRCh38
rs541125698 Chr.4: 161480968-161480968 on Build GRCh38
rs779560788 Chr.4: 161481029-161481029 on Build GRCh38
rs749910724 Chr.4: 161481161-161481161 on Build GRCh38
rs760462038 Chr.4: 161481140-161481140 on Build GRCh38
rs565207465 Chr.4: 161481421-161481421 on Build GRCh38
rs774271837 Chr.4: 161480983-161480983 on Build GRCh38
rs542859060 Chr.4: 161481238-161481238 on Build GRCh38
rs749243623 Chr.4: 161481285-161481286 on Build GRCh38
rs201852430 Chr.4: 161481203-161481203 on Build GRCh38
rs755755679 Chr.4: 161481032-161481032 on Build GRCh38
rs756822778 Chr.4: 161481064-161481064 on Build GRCh38
rs79444402 Chr.4: 161481244-161481244 on Build GRCh38
rs1023410825 Chr.4: 161481198-161481198 on Build GRCh38
rs764808067 Chr.4: 161481156-161481156 on Build GRCh38
rs912962061 Chr.4: 161481035-161481035 on Build GRCh38
rs769852932 Chr.4: 161481108-161481108 on Build GRCh38
rs747491394 Chr.4: 161481227-161481227 on Build GRCh38
rs144983464 Chr.4: 161481118-161481118 on Build GRCh38
rs758871689 Chr.4: 161481074-161481074 on Build GRCh38
rs1015172077 Chr.4: 161481219-161481219 on Build GRCh38
rs779619865 Chr.4: 161481065-161481065 on Build GRCh38
rs780548133 Chr.4: 161481181-161481181 on Build GRCh38
rs774618947 Chr.4: 161481286-161481287 on Build GRCh38
rs113349353 Chr.4: 161481346-161481346 on Build GRCh38
rs1050558651 Chr.4: 161481025-161481025 on Build GRCh38
rs1054736125 Chr.4: 161481419-161481419 on Build GRCh38
rs759439551 Chr.4: 161481113-161481113 on Build GRCh38
rs187455555 Chr.4: 161481122-161481122 on Build GRCh38
rs747346497 Chr.4: 161481081-161481081 on Build GRCh38
rs761456854 Chr.4: 161481147-161481147 on Build GRCh38
rs763720624 Chr.4: 161481141-161481141 on Build GRCh38
rs936129552 Chr.4: 161481402-161481402 on Build GRCh38
rs1016095011 Chr.4: 161481300-161481300 on Build GRCh38
rs988973081 Chr.4: 161481029-161481029 on Build GRCh38
rs768770120 Chr.4: 161481083-161481083 on Build GRCh38
rs780432267 Chr.4: 161481013-161481013 on Build GRCh38
rs751000924 Chr.4: 161481155-161481155 on Build GRCh38
rs758993616 Chr.4: 161481163-161481163 on Build GRCh38
rs766312402 Chr.4: 161480977-161480977 on Build GRCh38
rs115916272 Chr.4: 161481222-161481222 on Build GRCh38
rs748297344 Chr.4: 161481101-161481101 on Build GRCh38
rs147077925 Chr.4: 161481200-161481200 on Build GRCh38
rs183569275 Chr.4: 161481385-161481385 on Build GRCh38
rs1000458567 Chr.4: 161481097-161481097 on Build GRCh38
rs763754090 Chr.4: 161481036-161481036 on Build GRCh38
rs975066779 Chr.4: 161481374-161481374 on Build GRCh38
rs538974134 Chr.4: 161481073-161481073 on Build GRCh38
rs932014913 Chr.4: 161481246-161481246 on Build GRCh38
rs748350334 Chr.4: 161481212-161481212 on Build GRCh38
rs143585706 Chr.4: 161481043-161481043 on Build GRCh38
rs944790926 Chr.4: 161481380-161481380 on Build GRCh38
rs1011900475 Chr.4: 161481180-161481180 on Build GRCh38
rs759386499 Chr.4: 161480986-161480986 on Build GRCh38
rs986530839 Chr.4: 161481256-161481256 on Build GRCh38
rs1006086995 Chr.4: 161481291-161481291 on Build GRCh38
rs781331337 Chr.4: 161481208-161481208 on Build GRCh38
rs573628869 Chr.4: 161480966-161480966 on Build GRCh38
rs776850627 Chr.4: 161481096-161481096 on Build GRCh38
rs1041771670 Chr.4: 161481383-161481383 on Build GRCh38
rs964695915 Chr.4: 161481357-161481357 on Build GRCh38
rs192067562 Chr.4: 161481255-161481255 on Build GRCh38
rs772008065 Chr.4: 161481116-161481116 on Build GRCh38
rs202020180 Chr.4: 161481284-161481285 on Build GRCh38
rs752436601 Chr.4: 161481006-161481006 on Build GRCh38
rs774324832 Chr.4: 161481111-161481111 on Build GRCh38
rs974467436 Chr.4: 161480950-161480950 on Build GRCh38
rs767281636 Chr.4: 161480987-161480987 on Build GRCh38
rs12510971 Chr.4: 161480999-161480999 on Build GRCh38
rs571877451 Chr.4: 161481423-161481424 on Build GRCh38
rs912019479 Chr.4: 161481342-161481342 on Build GRCh38
rs890525909 Chr.4: 161481041-161481041 on Build GRCh38
rs920611444 Chr.4: 161480962-161480962 on Build GRCh38
rs780312167 Chr.4: 161481080-161481080 on Build GRCh38
rs913014446 Chr.4: 161481423-161481423 on Build GRCh38

View Full Product Details