Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 164957987-164958525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM218A
Entrez Gene ID
152756
Gene Name
family with sequence similarity 218 member A
Gene Aliases
C4orf39
UniGene
Hs.178648
Gene Chromosome Location
Chr.4: 164956948-164959121 on Build GRCh38
Transcripts
NM_153027.1 NM_001012414.2

Gene Symbol
TRIM61
Entrez Gene ID
391712
Gene Name
tripartite motif containing 61
Gene Aliases
RNF35
UniGene
Hs.654633
Gene Chromosome Location
Chr.4: 164954446-164977666 on Build GRCh38
Transcripts
NM_153027.1 NM_001012414.2

SNP Detail

SNP ID Chromosome Location
rs537356988 Chr.4: 164958335-164958335 on Build GRCh38
rs535011835 Chr.4: 164958477-164958477 on Build GRCh38
rs985812188 Chr.4: 164958409-164958409 on Build GRCh38
rs977193418 Chr.4: 164958386-164958386 on Build GRCh38
rs888871930 Chr.4: 164958093-164958093 on Build GRCh38
rs1021579374 Chr.4: 164958299-164958299 on Build GRCh38
rs943179931 Chr.4: 164958112-164958112 on Build GRCh38
rs958537507 Chr.4: 164958250-164958250 on Build GRCh38
rs930292314 Chr.4: 164958332-164958332 on Build GRCh38
rs930324712 Chr.4: 164958084-164958084 on Build GRCh38
rs892437074 Chr.4: 164958158-164958158 on Build GRCh38
rs994029560 Chr.4: 164958143-164958143 on Build GRCh38
rs972287343 Chr.4: 164958319-164958319 on Build GRCh38
rs906574433 Chr.4: 164958172-164958172 on Build GRCh38
rs570784464 Chr.4: 164958470-164958470 on Build GRCh38
rs559395896 Chr.4: 164958058-164958058 on Build GRCh38
rs796700425 Chr.4: 164958141-164958145 on Build GRCh38
rs981350794 Chr.4: 164958024-164958024 on Build GRCh38
rs897691072 Chr.4: 164958493-164958493 on Build GRCh38
rs889050294 Chr.4: 164958072-164958072 on Build GRCh38
rs558867533 Chr.4: 164958460-164958461 on Build GRCh38
rs941949464 Chr.4: 164958438-164958438 on Build GRCh38
rs966901845 Chr.4: 164958336-164958336 on Build GRCh38
rs910178973 Chr.4: 164958433-164958433 on Build GRCh38
rs764246089 Chr.4: 164958302-164958302 on Build GRCh38
rs932959649 Chr.4: 164958033-164958033 on Build GRCh38
rs1002176141 Chr.4: 164958214-164958214 on Build GRCh38
rs989708016 Chr.4: 164958279-164958279 on Build GRCh38
rs1011330872 Chr.4: 164958213-164958213 on Build GRCh38
rs10001548 Chr.4: 164958229-164958229 on Build GRCh38
rs772267120 Chr.4: 164958491-164958491 on Build GRCh38
rs140689608 Chr.4: 164958388-164958388 on Build GRCh38
rs1050357492 Chr.4: 164958046-164958046 on Build GRCh38
rs919524348 Chr.4: 164958323-164958323 on Build GRCh38
rs529958332 Chr.4: 164958248-164958248 on Build GRCh38
rs897620301 Chr.4: 164958118-164958118 on Build GRCh38
rs775530106 Chr.4: 164958191-164958191 on Build GRCh38
rs201464823 Chr.4: 164958064-164958064 on Build GRCh38
rs933028327 Chr.4: 164958396-164958396 on Build GRCh38
rs560935761 Chr.4: 164958247-164958247 on Build GRCh38
rs1048775785 Chr.4: 164958085-164958085 on Build GRCh38
rs1025008862 Chr.4: 164958169-164958169 on Build GRCh38
rs766541550 Chr.4: 164958023-164958024 on Build GRCh38
rs1031567012 Chr.4: 164958153-164958153 on Build GRCh38
rs1006108683 Chr.4: 164958101-164958101 on Build GRCh38
rs759823511 Chr.4: 164958028-164958029 on Build GRCh38
rs569837710 Chr.4: 164958311-164958311 on Build GRCh38
rs939575097 Chr.4: 164958026-164958026 on Build GRCh38
rs901720595 Chr.4: 164958134-164958134 on Build GRCh38
rs529888763 Chr.4: 164958117-164958117 on Build GRCh38

View Full Product Details