Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168237446-168237979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX60
Entrez Gene ID
55601
Gene Name
DEXD/H-box helicase 60
Gene Aliases
-
UniGene
Hs.591710
Gene Chromosome Location
Chr.4: 168216291-168318807 on Build GRCh38
Transcripts
NM_017631.5 XM_011532103.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
733166 c.4192C>T p.L1398L Chr.4: 168237768-168237768 on Build GRCh38
5351071 c.4218G>T p.M1406I Chr.4: 168237742-168237742 on Build GRCh38
6642068 c.4269+2T>C p.? Chr.4: 168237689-168237689 on Build GRCh38
3601841 c.4173A>C p.S1391S Chr.4: 168237787-168237787 on Build GRCh38
6216343 c.4176G>T p.V1392V Chr.4: 168237784-168237784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750806949 Chr.4: 168237814-168237814 on Build GRCh38
rs1049713078 Chr.4: 168237920-168237920 on Build GRCh38
rs932734862 Chr.4: 168237857-168237857 on Build GRCh38
rs1036219190 Chr.4: 168237913-168237913 on Build GRCh38
rs761427207 Chr.4: 168237731-168237731 on Build GRCh38
rs201353977 Chr.4: 168237691-168237691 on Build GRCh38
rs944052445 Chr.4: 168237547-168237547 on Build GRCh38
rs371200444 Chr.4: 168237760-168237760 on Build GRCh38
rs199827681 Chr.4: 168237841-168237841 on Build GRCh38
rs972933876 Chr.4: 168237472-168237472 on Build GRCh38
rs774931360 Chr.4: 168237708-168237708 on Build GRCh38
rs148385942 Chr.4: 168237781-168237781 on Build GRCh38
rs927229327 Chr.4: 168237588-168237588 on Build GRCh38
rs1050230916 Chr.4: 168237872-168237872 on Build GRCh38
rs758861055 Chr.4: 168237835-168237835 on Build GRCh38
rs904913567 Chr.4: 168237931-168237931 on Build GRCh38
rs1050698194 Chr.4: 168237632-168237632 on Build GRCh38
rs889946124 Chr.4: 168237834-168237834 on Build GRCh38
rs779317437 Chr.4: 168237694-168237694 on Build GRCh38
rs757461751 Chr.4: 168237796-168237796 on Build GRCh38
rs980930962 Chr.4: 168237756-168237756 on Build GRCh38
rs368653421 Chr.4: 168237665-168237665 on Build GRCh38
rs12506919 Chr.4: 168237921-168237921 on Build GRCh38
rs1006554246 Chr.4: 168237646-168237646 on Build GRCh38
rs767687017 Chr.4: 168237675-168237675 on Build GRCh38
rs758454014 Chr.4: 168237674-168237674 on Build GRCh38
rs199730154 Chr.4: 168237644-168237644 on Build GRCh38
rs773194993 Chr.4: 168237746-168237746 on Build GRCh38
rs41278577 Chr.4: 168237550-168237550 on Build GRCh38
rs766452761 Chr.4: 168237768-168237768 on Build GRCh38
rs768124041 Chr.4: 168237721-168237721 on Build GRCh38
rs993814765 Chr.4: 168237854-168237854 on Build GRCh38
rs142610127 Chr.4: 168237752-168237752 on Build GRCh38
rs756253380 Chr.4: 168237683-168237683 on Build GRCh38
rs752848935 Chr.4: 168237680-168237680 on Build GRCh38
rs899940009 Chr.4: 168237552-168237552 on Build GRCh38
rs746343096 Chr.4: 168237696-168237696 on Build GRCh38
rs146908767 Chr.4: 168237765-168237765 on Build GRCh38
rs1024658592 Chr.4: 168237741-168237741 on Build GRCh38
rs776356666 Chr.4: 168237722-168237722 on Build GRCh38
rs779227306 Chr.4: 168237808-168237808 on Build GRCh38
rs764138919 Chr.4: 168237784-168237784 on Build GRCh38
rs375342350 Chr.4: 168237656-168237656 on Build GRCh38
rs946732783 Chr.4: 168237692-168237692 on Build GRCh38
rs147721895 Chr.4: 168237745-168237745 on Build GRCh38
rs542743025 Chr.4: 168237479-168237479 on Build GRCh38
rs914183426 Chr.4: 168237811-168237811 on Build GRCh38
rs574540552 Chr.4: 168237943-168237943 on Build GRCh38
rs371938722 Chr.4: 168237847-168237847 on Build GRCh38
rs754086522 Chr.4: 168237790-168237790 on Build GRCh38
rs561107503 Chr.4: 168237499-168237499 on Build GRCh38
rs374919041 Chr.4: 168237773-168237773 on Build GRCh38
rs902558833 Chr.4: 168237753-168237753 on Build GRCh38
rs371974507 Chr.4: 168237652-168237652 on Build GRCh38
rs1024606434 Chr.4: 168237922-168237922 on Build GRCh38
rs531545429 Chr.4: 168237546-168237546 on Build GRCh38
rs941903008 Chr.4: 168237896-168237896 on Build GRCh38
rs779139974 Chr.4: 168237607-168237607 on Build GRCh38
rs772717360 Chr.4: 168237613-168237613 on Build GRCh38
rs887604454 Chr.4: 168237894-168237894 on Build GRCh38
rs1025383613 Chr.4: 168237532-168237532 on Build GRCh38
rs369230742 Chr.4: 168237840-168237840 on Build GRCh38
rs78723409 Chr.4: 168237941-168237941 on Build GRCh38
rs200068951 Chr.4: 168237718-168237718 on Build GRCh38
rs778062988 Chr.4: 168237690-168237690 on Build GRCh38

View Full Product Details