Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 169635597-169636126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN3
Entrez Gene ID
1182
Gene Name
chloride voltage-gated channel 3
Gene Aliases
CLC3, ClC-3
UniGene
Hs.481186
Gene Chromosome Location
Chr.4: 169620521-169723187 on Build GRCh38
Transcripts
NM_001243372.1 NM_001829.3 NM_173872.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
222967 c.109G>A p.D37N Chr.4: 169636037-169636037 on Build GRCh38
1729416 c.52A>T p.S18C Chr.4: 169635980-169635980 on Build GRCh38
447596 c.160+1G>T p.? Chr.4: 169636089-169636089 on Build GRCh38
6254433 c.12G>T p.E4D Chr.4: 169635940-169635940 on Build GRCh38
4123424 c.139T>C p.L47L Chr.4: 169636067-169636067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903364926 Chr.4: 169636035-169636035 on Build GRCh38
rs775886629 Chr.4: 169635901-169635901 on Build GRCh38
rs986532357 Chr.4: 169635861-169635861 on Build GRCh38
rs554007804 Chr.4: 169635873-169635873 on Build GRCh38
rs1041886056 Chr.4: 169635952-169635952 on Build GRCh38
rs1011783257 Chr.4: 169635636-169635636 on Build GRCh38
rs772334005 Chr.4: 169636009-169636009 on Build GRCh38
rs746091406 Chr.4: 169635899-169635899 on Build GRCh38
rs909172566 Chr.4: 169635787-169635787 on Build GRCh38
rs751440151 Chr.4: 169635961-169635961 on Build GRCh38
rs538791858 Chr.4: 169635837-169635837 on Build GRCh38
rs566774783 Chr.4: 169635638-169635638 on Build GRCh38
rs762388847 Chr.4: 169636062-169636062 on Build GRCh38
rs761226535 Chr.4: 169635902-169635902 on Build GRCh38
rs937381688 Chr.4: 169635871-169635871 on Build GRCh38
rs747298399 Chr.4: 169636038-169636038 on Build GRCh38
rs757213816 Chr.4: 169635978-169635978 on Build GRCh38
rs755491509 Chr.4: 169635897-169635897 on Build GRCh38
rs776947851 Chr.4: 169636051-169636051 on Build GRCh38
rs1043231677 Chr.4: 169636042-169636042 on Build GRCh38
rs143082055 Chr.4: 169635803-169635803 on Build GRCh38
rs761181927 Chr.4: 169635920-169635920 on Build GRCh38
rs559222364 Chr.4: 169635826-169635826 on Build GRCh38
rs766089486 Chr.4: 169636087-169636087 on Build GRCh38
rs904780443 Chr.4: 169635818-169635818 on Build GRCh38
rs1057457298 Chr.4: 169635969-169635969 on Build GRCh38
rs769080194 Chr.4: 169635875-169635875 on Build GRCh38
rs9996873 Chr.4: 169635695-169635695 on Build GRCh38
rs567634703 Chr.4: 169635701-169635701 on Build GRCh38
rs764854983 Chr.4: 169635926-169635926 on Build GRCh38
rs754831858 Chr.4: 169635966-169635966 on Build GRCh38
rs919799334 Chr.4: 169635896-169635897 on Build GRCh38
rs780940007 Chr.4: 169635970-169635970 on Build GRCh38
rs967081592 Chr.4: 169635668-169635668 on Build GRCh38
rs986437534 Chr.4: 169635778-169635778 on Build GRCh38
rs1022264560 Chr.4: 169635809-169635810 on Build GRCh38
rs938710480 Chr.4: 169635963-169635963 on Build GRCh38
rs746072762 Chr.4: 169636000-169636000 on Build GRCh38
rs762761846 Chr.4: 169635940-169635940 on Build GRCh38
rs769424677 Chr.4: 169636074-169636074 on Build GRCh38
rs751168388 Chr.4: 169635683-169635683 on Build GRCh38
rs772417603 Chr.4: 169635900-169635900 on Build GRCh38
rs887151260 Chr.4: 169635730-169635730 on Build GRCh38
rs1019382194 Chr.4: 169635862-169635862 on Build GRCh38
rs780368756 Chr.4: 169636022-169636022 on Build GRCh38
rs199640504 Chr.4: 169636081-169636081 on Build GRCh38
rs772615293 Chr.4: 169636101-169636101 on Build GRCh38
rs866530257 Chr.4: 169635667-169635667 on Build GRCh38
rs142998163 Chr.4: 169636045-169636045 on Build GRCh38
rs894617342 Chr.4: 169635789-169635789 on Build GRCh38
rs779026941 Chr.4: 169635983-169635983 on Build GRCh38
rs774723149 Chr.4: 169635809-169635809 on Build GRCh38
rs956391607 Chr.4: 169635851-169635855 on Build GRCh38
rs750029741 Chr.4: 169635935-169635935 on Build GRCh38
rs542838107 Chr.4: 169635823-169635823 on Build GRCh38
rs752690413 Chr.4: 169635973-169635973 on Build GRCh38
rs373294356 Chr.4: 169636078-169636078 on Build GRCh38
rs748721807 Chr.4: 169636052-169636052 on Build GRCh38
rs549147161 Chr.4: 169635696-169635696 on Build GRCh38
rs192705380 Chr.4: 169635918-169635918 on Build GRCh38
rs975282259 Chr.4: 169635889-169635889 on Build GRCh38
rs573754599 Chr.4: 169635901-169635902 on Build GRCh38
rs1045806932 Chr.4: 169635827-169635827 on Build GRCh38
rs916012724 Chr.4: 169635980-169635980 on Build GRCh38
rs766254279 Chr.4: 169635951-169635951 on Build GRCh38
rs948864639 Chr.4: 169636039-169636039 on Build GRCh38
rs747452518 Chr.4: 169635879-169635879 on Build GRCh38
rs893355435 Chr.4: 169635903-169635903 on Build GRCh38
rs1057174174 Chr.4: 169635757-169635757 on Build GRCh38
rs537982005 Chr.4: 169635799-169635799 on Build GRCh38
rs71622351 Chr.4: 169635625-169635625 on Build GRCh38
rs769086873 Chr.4: 169635904-169635904 on Build GRCh38
rs377531867 Chr.4: 169636099-169636099 on Build GRCh38

View Full Product Details