Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 170060570-170061082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADAT
Entrez Gene ID
51166
Gene Name
aminoadipate aminotransferase
Gene Aliases
KAT2, KATII, KYAT2
UniGene
Hs.529735
Gene Chromosome Location
Chr.4: 170060222-170091699 on Build GRCh38
Transcripts
NM_001286682.1 NM_001286683.1 NM_016228.3 NM_182662.1 XM_006714231.2 XM_011532020.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6860560 c.1237-10_1237-9insT p.? Chr.4: 170060978-170060979 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs981348526 Chr.4: 170060771-170060772 on Build GRCh38
rs995116644 Chr.4: 170060829-170060829 on Build GRCh38
rs541493410 Chr.4: 170060671-170060671 on Build GRCh38
rs915818606 Chr.4: 170060704-170060704 on Build GRCh38
rs969928231 Chr.4: 170061022-170061022 on Build GRCh38
rs1036659968 Chr.4: 170060916-170060916 on Build GRCh38
rs946017472 Chr.4: 170060830-170060830 on Build GRCh38
rs992903266 Chr.4: 170060989-170060989 on Build GRCh38
rs558994689 Chr.4: 170060973-170060973 on Build GRCh38
rs750856764 Chr.4: 170061014-170061014 on Build GRCh38
rs538945896 Chr.4: 170060770-170060770 on Build GRCh38
rs1040358933 Chr.4: 170060831-170060831 on Build GRCh38
rs760285261 Chr.4: 170061031-170061031 on Build GRCh38
rs1027807648 Chr.4: 170060856-170060856 on Build GRCh38
rs913499928 Chr.4: 170060826-170060826 on Build GRCh38
rs932044757 Chr.4: 170060845-170060845 on Build GRCh38
rs879930008 Chr.4: 170060772-170060787 on Build GRCh38
rs369301970 Chr.4: 170060890-170060890 on Build GRCh38
rs769515661 Chr.4: 170060933-170060933 on Build GRCh38
rs1006137757 Chr.4: 170060597-170060597 on Build GRCh38
rs948671423 Chr.4: 170060723-170060723 on Build GRCh38
rs186211908 Chr.4: 170060904-170060904 on Build GRCh38
rs1042855589 Chr.4: 170060743-170060744 on Build GRCh38
rs554734932 Chr.4: 170060860-170060860 on Build GRCh38
rs992727600 Chr.4: 170060649-170060649 on Build GRCh38
rs980516740 Chr.4: 170060785-170060785 on Build GRCh38
rs1053624591 Chr.4: 170060773-170060773 on Build GRCh38
rs990773908 Chr.4: 170060802-170060802 on Build GRCh38
rs775157880 Chr.4: 170060979-170060979 on Build GRCh38
rs571564328 Chr.4: 170060759-170060759 on Build GRCh38
rs192025116 Chr.4: 170060989-170060989 on Build GRCh38
rs890426694 Chr.4: 170060783-170060783 on Build GRCh38
rs536876521 Chr.4: 170060825-170060825 on Build GRCh38
rs1050538473 Chr.4: 170060867-170060867 on Build GRCh38
rs774493607 Chr.4: 170060990-170060990 on Build GRCh38
rs982553693 Chr.4: 170060603-170060603 on Build GRCh38
rs116231186 Chr.4: 170060796-170060796 on Build GRCh38
rs774840072 Chr.4: 170060842-170060842 on Build GRCh38
rs773074451 Chr.4: 170060952-170060952 on Build GRCh38
rs748794589 Chr.4: 170060665-170060666 on Build GRCh38
rs139230340 Chr.4: 170060961-170060961 on Build GRCh38
rs149113985 Chr.4: 170061054-170061055 on Build GRCh38
rs776284073 Chr.4: 170060631-170060631 on Build GRCh38
rs561797935 Chr.4: 170060795-170060795 on Build GRCh38
rs201901211 Chr.4: 170060978-170060979 on Build GRCh38
rs747704435 Chr.4: 170060908-170060908 on Build GRCh38
rs969091260 Chr.4: 170060774-170060774 on Build GRCh38
rs1014844248 Chr.4: 170060602-170060602 on Build GRCh38
rs34202278 Chr.4: 170060632-170060632 on Build GRCh38

View Full Product Details