Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 170061752-170062261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADAT
Entrez Gene ID
51166
Gene Name
aminoadipate aminotransferase
Gene Aliases
KAT2, KATII, KYAT2
UniGene
Hs.529735
Gene Chromosome Location
Chr.4: 170060222-170091699 on Build GRCh38
Transcripts
NM_001286682.1 NM_001286683.1 NM_016228.3 NM_182662.1 XM_006714231.2 XM_011532020.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1633557 c.1206C>A p.F402L Chr.4: 170061922-170061922 on Build GRCh38
7320967 c.1165G>A p.V389I Chr.4: 170061963-170061963 on Build GRCh38
4421313 c.1159T>C p.F387L Chr.4: 170061969-170061969 on Build GRCh38
1633558 c.1205T>A p.F402Y Chr.4: 170061923-170061923 on Build GRCh38
3781782 c.1135-7C>G p.? Chr.4: 170062000-170062000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186589353 Chr.4: 170062115-170062115 on Build GRCh38
rs764126506 Chr.4: 170061844-170061844 on Build GRCh38
rs546201219 Chr.4: 170062147-170062147 on Build GRCh38
rs148517792 Chr.4: 170062174-170062174 on Build GRCh38
rs949251720 Chr.4: 170061783-170061783 on Build GRCh38
rs183344666 Chr.4: 170062072-170062072 on Build GRCh38
rs767495658 Chr.4: 170061963-170061963 on Build GRCh38
rs780525079 Chr.4: 170061894-170061894 on Build GRCh38
rs530659731 Chr.4: 170061895-170061895 on Build GRCh38
rs369670390 Chr.4: 170062042-170062042 on Build GRCh38
rs765444553 Chr.4: 170061987-170061987 on Build GRCh38
rs1045196398 Chr.4: 170062231-170062231 on Build GRCh38
rs780752607 Chr.4: 170062108-170062108 on Build GRCh38
rs371494823 Chr.4: 170061885-170061885 on Build GRCh38
rs369053395 Chr.4: 170061882-170061882 on Build GRCh38
rs781727621 Chr.4: 170062023-170062023 on Build GRCh38
rs773056396 Chr.4: 170061928-170061928 on Build GRCh38
rs1054031905 Chr.4: 170061961-170061961 on Build GRCh38
rs948340234 Chr.4: 170062192-170062192 on Build GRCh38
rs559753344 Chr.4: 170062151-170062151 on Build GRCh38
rs563864565 Chr.4: 170062116-170062116 on Build GRCh38
rs376931614 Chr.4: 170061957-170061957 on Build GRCh38
rs1045245780 Chr.4: 170061955-170061955 on Build GRCh38
rs1019170860 Chr.4: 170061785-170061785 on Build GRCh38
rs903316067 Chr.4: 170062033-170062033 on Build GRCh38
rs965920208 Chr.4: 170061790-170061790 on Build GRCh38
rs375265742 Chr.4: 170062014-170062014 on Build GRCh38
rs998103699 Chr.4: 170062047-170062047 on Build GRCh38
rs770888623 Chr.4: 170061940-170061940 on Build GRCh38
rs553269600 Chr.4: 170061823-170061823 on Build GRCh38
rs574686394 Chr.4: 170061832-170061832 on Build GRCh38
rs766683086 Chr.4: 170062011-170062011 on Build GRCh38
rs1007843755 Chr.4: 170062146-170062146 on Build GRCh38
rs758737405 Chr.4: 170062219-170062219 on Build GRCh38
rs757357837 Chr.4: 170061867-170061867 on Build GRCh38
rs753985591 Chr.4: 170061851-170061851 on Build GRCh38
rs749090252 Chr.4: 170061937-170061937 on Build GRCh38
rs564484199 Chr.4: 170061925-170061925 on Build GRCh38
rs762398883 Chr.4: 170061848-170061849 on Build GRCh38
rs758831171 Chr.4: 170061879-170061879 on Build GRCh38
rs759938840 Chr.4: 170062119-170062119 on Build GRCh38
rs776516272 Chr.4: 170061964-170061964 on Build GRCh38
rs755533124 Chr.4: 170061891-170061891 on Build GRCh38
rs992778888 Chr.4: 170062185-170062185 on Build GRCh38
rs755371345 Chr.4: 170062013-170062013 on Build GRCh38
rs762011420 Chr.4: 170061984-170061984 on Build GRCh38
rs747594491 Chr.4: 170062029-170062029 on Build GRCh38
rs940914684 Chr.4: 170061913-170061913 on Build GRCh38
rs34877246 Chr.4: 170062012-170062013 on Build GRCh38
rs548647616 Chr.4: 170062178-170062178 on Build GRCh38
rs919350538 Chr.4: 170062187-170062187 on Build GRCh38
rs368940133 Chr.4: 170061841-170061841 on Build GRCh38
rs533355016 Chr.4: 170062089-170062090 on Build GRCh38
rs769538121 Chr.4: 170062156-170062156 on Build GRCh38
rs747522798 Chr.4: 170061883-170061883 on Build GRCh38
rs952149108 Chr.4: 170061881-170061881 on Build GRCh38
rs751983389 Chr.4: 170062012-170062012 on Build GRCh38
rs758742764 Chr.4: 170062002-170062002 on Build GRCh38
rs200697371 Chr.4: 170061875-170061875 on Build GRCh38
rs139598358 Chr.4: 170061960-170061960 on Build GRCh38
rs948247416 Chr.4: 170061943-170061943 on Build GRCh38
rs765462648 Chr.4: 170061874-170061874 on Build GRCh38
rs541616015 Chr.4: 170061858-170061858 on Build GRCh38
rs528047884 Chr.4: 170062127-170062127 on Build GRCh38

View Full Product Details