Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 170066301-170066838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADAT
Entrez Gene ID
51166
Gene Name
aminoadipate aminotransferase
Gene Aliases
KAT2, KATII, KYAT2
UniGene
Hs.529735
Gene Chromosome Location
Chr.4: 170060222-170091699 on Build GRCh38
Transcripts
NM_001286682.1 NM_001286683.1 NM_016228.3 NM_182662.1 XM_006714231.2 XM_011532020.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4877612 c.983A>G p.N328S Chr.4: 170066458-170066458 on Build GRCh38
6589230 c.984C>T p.N328N Chr.4: 170066457-170066457 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs141571124 Chr.4: 170066765-170066776 on Build GRCh38
rs41278617 Chr.4: 170066750-170066750 on Build GRCh38
rs781631913 Chr.4: 170066536-170066536 on Build GRCh38
rs921398428 Chr.4: 170066450-170066450 on Build GRCh38
rs879628663 Chr.4: 170066392-170066392 on Build GRCh38
rs969029404 Chr.4: 170066806-170066806 on Build GRCh38
rs374625874 Chr.4: 170066403-170066403 on Build GRCh38
rs761180610 Chr.4: 170066529-170066529 on Build GRCh38
rs1022300547 Chr.4: 170066798-170066798 on Build GRCh38
rs774414364 Chr.4: 170066512-170066512 on Build GRCh38
rs370870683 Chr.4: 170066524-170066524 on Build GRCh38
rs1480544 Chr.4: 170066485-170066485 on Build GRCh38
rs779862124 Chr.4: 170066468-170066468 on Build GRCh38
rs116082202 Chr.4: 170066462-170066462 on Build GRCh38
rs780582925 Chr.4: 170066642-170066642 on Build GRCh38
rs766326292 Chr.4: 170066328-170066328 on Build GRCh38
rs1014256285 Chr.4: 170066573-170066573 on Build GRCh38
rs755909309 Chr.4: 170066595-170066596 on Build GRCh38
rs796220191 Chr.4: 170066333-170066333 on Build GRCh38
rs753132944 Chr.4: 170066395-170066395 on Build GRCh38
rs895858375 Chr.4: 170066547-170066547 on Build GRCh38
rs778399666 Chr.4: 170066460-170066460 on Build GRCh38
rs772212212 Chr.4: 170066518-170066518 on Build GRCh38
rs746236198 Chr.4: 170066546-170066546 on Build GRCh38
rs550008061 Chr.4: 170066620-170066620 on Build GRCh38
rs561260112 Chr.4: 170066548-170066548 on Build GRCh38
rs919736134 Chr.4: 170066614-170066614 on Build GRCh38
rs563254595 Chr.4: 170066594-170066594 on Build GRCh38
rs548986259 Chr.4: 170066522-170066522 on Build GRCh38
rs369026657 Chr.4: 170066464-170066464 on Build GRCh38
rs1033283839 Chr.4: 170066664-170066664 on Build GRCh38
rs781161935 Chr.4: 170066483-170066483 on Build GRCh38
rs538732065 Chr.4: 170066753-170066753 on Build GRCh38
rs759781914 Chr.4: 170066716-170066716 on Build GRCh38
rs147615281 Chr.4: 170066416-170066416 on Build GRCh38
rs527720783 Chr.4: 170066513-170066513 on Build GRCh38
rs1013128763 Chr.4: 170066779-170066780 on Build GRCh38
rs1000052607 Chr.4: 170066623-170066623 on Build GRCh38
rs887175758 Chr.4: 170066745-170066745 on Build GRCh38
rs545144600 Chr.4: 170066473-170066473 on Build GRCh38
rs774697043 Chr.4: 170066389-170066389 on Build GRCh38
rs759788081 Chr.4: 170066391-170066391 on Build GRCh38
rs370313181 Chr.4: 170066373-170066373 on Build GRCh38
rs764693858 Chr.4: 170066530-170066530 on Build GRCh38
rs988457870 Chr.4: 170066325-170066325 on Build GRCh38
rs754504488 Chr.4: 170066402-170066402 on Build GRCh38
rs754848971 Chr.4: 170066477-170066477 on Build GRCh38
rs544424541 Chr.4: 170066394-170066394 on Build GRCh38
rs756896158 Chr.4: 170066418-170066418 on Build GRCh38

View Full Product Details