Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174643769-174644301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRA3
Entrez Gene ID
8001
Gene Name
glycine receptor alpha 3
Gene Aliases
-
UniGene
Hs.413099
Gene Chromosome Location
Chr.4: 174642011-174829568 on Build GRCh38
Transcripts
NM_001042543.2 NM_006529.3 XM_011532268.2 XM_017008630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1671439 c.1297C>T p.R433* Chr.4: 174643884-174643884 on Build GRCh38
3602083 c.1199C>T p.P400L Chr.4: 174643982-174643982 on Build GRCh38
7181936 c.1200A>T p.P400P Chr.4: 174643981-174643981 on Build GRCh38
5344675 c.1293C>A p.I431I Chr.4: 174643888-174643888 on Build GRCh38
3602082 c.1206C>T p.G402G Chr.4: 174643975-174643975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759915545 Chr.4: 174643881-174643881 on Build GRCh38
rs796080466 Chr.4: 174644126-174644126 on Build GRCh38
rs1023128987 Chr.4: 174644008-174644008 on Build GRCh38
rs1040155477 Chr.4: 174643870-174643870 on Build GRCh38
rs780098337 Chr.4: 174643924-174643924 on Build GRCh38
rs1016567434 Chr.4: 174643978-174643978 on Build GRCh38
rs761345475 Chr.4: 174644004-174644004 on Build GRCh38
rs544493104 Chr.4: 174644198-174644198 on Build GRCh38
rs940284958 Chr.4: 174644172-174644172 on Build GRCh38
rs755939109 Chr.4: 174644167-174644167 on Build GRCh38
rs765924811 Chr.4: 174643931-174643931 on Build GRCh38
rs573092808 Chr.4: 174644129-174644129 on Build GRCh38
rs754973814 Chr.4: 174644009-174644009 on Build GRCh38
rs1045476641 Chr.4: 174644049-174644049 on Build GRCh38
rs532546694 Chr.4: 174643797-174643797 on Build GRCh38
rs75893041 Chr.4: 174644033-174644033 on Build GRCh38
rs749515822 Chr.4: 174643866-174643866 on Build GRCh38
rs766164934 Chr.4: 174644028-174644028 on Build GRCh38
rs762874196 Chr.4: 174644026-174644026 on Build GRCh38
rs759002807 Chr.4: 174644073-174644074 on Build GRCh38
rs375970694 Chr.4: 174643900-174643900 on Build GRCh38
rs964822223 Chr.4: 174643849-174643849 on Build GRCh38
rs773489264 Chr.4: 174643838-174643838 on Build GRCh38
rs779111192 Chr.4: 174643945-174643945 on Build GRCh38
rs145802010 Chr.4: 174644042-174644042 on Build GRCh38
rs554679165 Chr.4: 174643934-174643934 on Build GRCh38
rs750173498 Chr.4: 174644022-174644022 on Build GRCh38
rs112221905 Chr.4: 174644133-174644133 on Build GRCh38
rs757337154 Chr.4: 174644141-174644141 on Build GRCh38
rs575491802 Chr.4: 174644189-174644189 on Build GRCh38
rs990020566 Chr.4: 174643829-174643829 on Build GRCh38
rs148421929 Chr.4: 174643892-174643892 on Build GRCh38
rs142571571 Chr.4: 174643915-174643915 on Build GRCh38
rs532523180 Chr.4: 174644218-174644218 on Build GRCh38
rs915217485 Chr.4: 174643802-174643802 on Build GRCh38
rs1051719325 Chr.4: 174644164-174644164 on Build GRCh38
rs376118696 Chr.4: 174644056-174644056 on Build GRCh38
rs1038547770 Chr.4: 174644174-174644174 on Build GRCh38
rs746211010 Chr.4: 174644062-174644062 on Build GRCh38
rs745747356 Chr.4: 174643790-174643790 on Build GRCh38
rs55885329 Chr.4: 174644140-174644140 on Build GRCh38
rs758247302 Chr.4: 174643920-174643920 on Build GRCh38
rs548476104 Chr.4: 174643935-174643935 on Build GRCh38
rs753486096 Chr.4: 174643910-174643910 on Build GRCh38
rs373319986 Chr.4: 174644183-174644183 on Build GRCh38
rs761280535 Chr.4: 174643975-174643975 on Build GRCh38
rs4235212 Chr.4: 174644094-174644094 on Build GRCh38
rs746289463 Chr.4: 174643951-174643951 on Build GRCh38
rs1055333321 Chr.4: 174643983-174643983 on Build GRCh38
rs1017485474 Chr.4: 174644131-174644131 on Build GRCh38
rs563561979 Chr.4: 174644205-174644205 on Build GRCh38
rs749462691 Chr.4: 174643936-174643936 on Build GRCh38
rs772841058 Chr.4: 174643883-174643883 on Build GRCh38
rs1012660544 Chr.4: 174643993-174643993 on Build GRCh38
rs150967429 Chr.4: 174643970-174643970 on Build GRCh38
rs1031481752 Chr.4: 174644075-174644075 on Build GRCh38
rs893090396 Chr.4: 174644124-174644124 on Build GRCh38
rs942274067 Chr.4: 174643972-174643972 on Build GRCh38
rs949517674 Chr.4: 174644040-174644040 on Build GRCh38
rs772442290 Chr.4: 174643964-174643964 on Build GRCh38
rs372873478 Chr.4: 174644035-174644035 on Build GRCh38
rs369207686 Chr.4: 174643911-174643911 on Build GRCh38
rs372292473 Chr.4: 174643812-174643812 on Build GRCh38
rs371337324 Chr.4: 174643846-174643846 on Build GRCh38
rs747482564 Chr.4: 174644100-174644100 on Build GRCh38
rs898201586 Chr.4: 174644187-174644187 on Build GRCh38
rs748223691 Chr.4: 174643930-174643930 on Build GRCh38
rs764989600 Chr.4: 174644018-174644018 on Build GRCh38
rs771142846 Chr.4: 174643880-174643880 on Build GRCh38
rs776315842 Chr.4: 174643996-174643996 on Build GRCh38
rs753987637 Chr.4: 174644054-174644054 on Build GRCh38
rs768235434 Chr.4: 174643976-174643976 on Build GRCh38
rs764904885 Chr.4: 174643914-174643914 on Build GRCh38
rs553067062 Chr.4: 174644108-174644108 on Build GRCh38
rs369604752 Chr.4: 174644067-174644067 on Build GRCh38
rs201290451 Chr.4: 174643909-174643909 on Build GRCh38
rs1053178715 Chr.4: 174644116-174644116 on Build GRCh38
rs568373129 Chr.4: 174644034-174644034 on Build GRCh38
rs760317437 Chr.4: 174643906-174643906 on Build GRCh38
rs570403797 Chr.4: 174644057-174644057 on Build GRCh38
rs779025643 Chr.4: 174644175-174644175 on Build GRCh38
rs898132805 Chr.4: 174643980-174643980 on Build GRCh38
rs781103156 Chr.4: 174643804-174643804 on Build GRCh38
rs771215209 Chr.4: 174643940-174643940 on Build GRCh38
rs1030432796 Chr.4: 174644157-174644157 on Build GRCh38
rs767185308 Chr.4: 174644043-174644043 on Build GRCh38
rs144082170 Chr.4: 174643836-174643836 on Build GRCh38

View Full Product Details