Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 177438667-177439103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGA
Entrez Gene ID
175
Gene Name
aspartylglucosaminidase
Gene Aliases
AGU, ASRG, GA
UniGene
Hs.207776
Gene Chromosome Location
Chr.4: 177430774-177442503 on Build GRCh38
Transcripts
NM_000027.3 NM_001171988.1 NR_033655.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3373442 c.489C>T p.C163C Chr.4: 177438763-177438763 on Build GRCh38
5572094 c.504G>A p.W168* Chr.4: 177438748-177438748 on Build GRCh38
140503 c.457G>A p.A153T Chr.4: 177438795-177438795 on Build GRCh38
6348989 c.482G>T p.R161L Chr.4: 177438770-177438770 on Build GRCh38
6894406 c.430G>A p.E144K Chr.4: 177438822-177438822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575273988 Chr.4: 177439063-177439063 on Build GRCh38
rs1035465111 Chr.4: 177438708-177438708 on Build GRCh38
rs919741792 Chr.4: 177438970-177438970 on Build GRCh38
rs759303473 Chr.4: 177438892-177438892 on Build GRCh38
rs6829436 Chr.4: 177438991-177438991 on Build GRCh38
rs199682344 Chr.4: 177438817-177438817 on Build GRCh38
rs369809419 Chr.4: 177438706-177438706 on Build GRCh38
rs777637637 Chr.4: 177438771-177438771 on Build GRCh38
rs973498288 Chr.4: 177439075-177439075 on Build GRCh38
rs767409598 Chr.4: 177438761-177438761 on Build GRCh38
rs2228119 Chr.4: 177438806-177438806 on Build GRCh38
rs765893106 Chr.4: 177438897-177438897 on Build GRCh38
rs773819054 Chr.4: 177438857-177438857 on Build GRCh38
rs386833426 Chr.4: 177438865-177438865 on Build GRCh38
rs1057517329 Chr.4: 177438762-177438762 on Build GRCh38
rs748440983 Chr.4: 177438842-177438842 on Build GRCh38
rs374995967 Chr.4: 177438883-177438883 on Build GRCh38
rs945449841 Chr.4: 177439074-177439074 on Build GRCh38
rs931153394 Chr.4: 177438983-177438983 on Build GRCh38
rs772742442 Chr.4: 177438891-177438892 on Build GRCh38
rs192195150 Chr.4: 177438770-177438770 on Build GRCh38
rs899560320 Chr.4: 177438921-177438921 on Build GRCh38
rs6855346 Chr.4: 177439042-177439042 on Build GRCh38
rs776555596 Chr.4: 177438718-177438718 on Build GRCh38
rs919288111 Chr.4: 177438868-177438868 on Build GRCh38
rs1001103607 Chr.4: 177438871-177438871 on Build GRCh38
rs902873735 Chr.4: 177438951-177438951 on Build GRCh38
rs530298258 Chr.4: 177438861-177438861 on Build GRCh38
rs1039649070 Chr.4: 177438920-177438920 on Build GRCh38
rs370323493 Chr.4: 177438902-177438902 on Build GRCh38
rs184031039 Chr.4: 177438758-177438758 on Build GRCh38
rs1021215661 Chr.4: 177438936-177438936 on Build GRCh38
rs770618714 Chr.4: 177438876-177438876 on Build GRCh38
rs961109623 Chr.4: 177438941-177438941 on Build GRCh38
rs991950983 Chr.4: 177438798-177438798 on Build GRCh38
rs745976989 Chr.4: 177438779-177438779 on Build GRCh38
rs386833430 Chr.4: 177438749-177438749 on Build GRCh38
rs752551444 Chr.4: 177438895-177438895 on Build GRCh38
rs766165178 Chr.4: 177438743-177438743 on Build GRCh38
rs761357861 Chr.4: 177438718-177438719 on Build GRCh38
rs121964904 Chr.4: 177438764-177438764 on Build GRCh38
rs762507077 Chr.4: 177438739-177438739 on Build GRCh38
rs567125055 Chr.4: 177438910-177438910 on Build GRCh38
rs386833427 Chr.4: 177438848-177438848 on Build GRCh38
rs77927123 Chr.4: 177439012-177439012 on Build GRCh38
rs1023243767 Chr.4: 177439066-177439066 on Build GRCh38
rs150633580 Chr.4: 177438719-177438719 on Build GRCh38
rs386833428 Chr.4: 177438813-177438813 on Build GRCh38
rs766030582 Chr.4: 177438726-177438726 on Build GRCh38
rs184625943 Chr.4: 177439068-177439068 on Build GRCh38
rs146381591 Chr.4: 177438816-177438816 on Build GRCh38
rs953707707 Chr.4: 177438882-177438882 on Build GRCh38
rs373800568 Chr.4: 177439027-177439027 on Build GRCh38
rs144349742 Chr.4: 177438824-177438824 on Build GRCh38
rs767066628 Chr.4: 177438893-177438893 on Build GRCh38
rs943541481 Chr.4: 177438917-177438917 on Build GRCh38
rs972508281 Chr.4: 177438968-177438968 on Build GRCh38
rs534506747 Chr.4: 177438932-177438932 on Build GRCh38
rs769429451 Chr.4: 177438839-177438840 on Build GRCh38
rs762407762 Chr.4: 177438894-177438895 on Build GRCh38
rs370524263 Chr.4: 177438838-177438838 on Build GRCh38
rs1011768926 Chr.4: 177438923-177438923 on Build GRCh38
rs202033374 Chr.4: 177438795-177438795 on Build GRCh38
rs1029008254 Chr.4: 177438877-177438877 on Build GRCh38
rs937988761 Chr.4: 177439020-177439020 on Build GRCh38
rs771619882 Chr.4: 177438911-177438911 on Build GRCh38
rs770449168 Chr.4: 177438850-177438850 on Build GRCh38
rs554486000 Chr.4: 177439070-177439070 on Build GRCh38
rs752614801 Chr.4: 177439022-177439022 on Build GRCh38

View Full Product Details