Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183317756-183318257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN22
Entrez Gene ID
53842
Gene Name
claudin 22
Gene Aliases
CLDN21
UniGene
Hs.333179
Gene Chromosome Location
Chr.4: 183318067-183320774 on Build GRCh38
Transcripts
NM_001111319.1 NM_024949.5 XM_011532269.2

Gene Symbol
WWC2
Entrez Gene ID
80014
Gene Name
WW and C2 domain containing 2
Gene Aliases
BOMB
UniGene
Hs.333179
Gene Chromosome Location
Chr.4: 183098677-183320777 on Build GRCh38
Transcripts
NM_001111319.1 NM_024949.5 XM_011532269.2

SNP Detail

SNP ID Chromosome Location
rs770185337 Chr.4: 183317828-183317828 on Build GRCh38
rs897787308 Chr.4: 183318216-183318216 on Build GRCh38
rs886094905 Chr.4: 183318111-183318111 on Build GRCh38
rs567418069 Chr.4: 183317860-183317860 on Build GRCh38
rs920848360 Chr.4: 183317936-183317936 on Build GRCh38
rs1010138780 Chr.4: 183317785-183317785 on Build GRCh38
rs180836649 Chr.4: 183318176-183318176 on Build GRCh38
rs572445343 Chr.4: 183317806-183317806 on Build GRCh38
rs979750892 Chr.4: 183318143-183318143 on Build GRCh38
rs1041603584 Chr.4: 183318124-183318124 on Build GRCh38
rs781202881 Chr.4: 183317809-183317809 on Build GRCh38
rs1042059197 Chr.4: 183317818-183317818 on Build GRCh38
rs950195179 Chr.4: 183317956-183317956 on Build GRCh38
rs533992504 Chr.4: 183318034-183318034 on Build GRCh38
rs1044554670 Chr.4: 183317993-183317993 on Build GRCh38
rs967944815 Chr.4: 183317873-183317873 on Build GRCh38
rs1024850933 Chr.4: 183318215-183318215 on Build GRCh38
rs191029395 Chr.4: 183318131-183318131 on Build GRCh38
rs979368806 Chr.4: 183317875-183317875 on Build GRCh38
rs1024296525 Chr.4: 183317913-183317913 on Build GRCh38
rs748133229 Chr.4: 183318028-183318028 on Build GRCh38
rs1000040175 Chr.4: 183317850-183317850 on Build GRCh38
rs956194722 Chr.4: 183318187-183318187 on Build GRCh38
rs910947700 Chr.4: 183318020-183318020 on Build GRCh38
rs904351693 Chr.4: 183317833-183317833 on Build GRCh38
rs904454649 Chr.4: 183318125-183318125 on Build GRCh38
rs550510379 Chr.4: 183317808-183317808 on Build GRCh38
rs185536982 Chr.4: 183317930-183317930 on Build GRCh38
rs1014340815 Chr.4: 183318205-183318205 on Build GRCh38
rs986274459 Chr.4: 183318015-183318015 on Build GRCh38
rs962388428 Chr.4: 183317919-183317919 on Build GRCh38
rs995957514 Chr.4: 183318133-183318133 on Build GRCh38
rs541028320 Chr.4: 183317885-183317885 on Build GRCh38
rs894342773 Chr.4: 183318186-183318186 on Build GRCh38
rs1055619746 Chr.4: 183318138-183318138 on Build GRCh38
rs558754936 Chr.4: 183318066-183318066 on Build GRCh38
rs952178826 Chr.4: 183317966-183317966 on Build GRCh38
rs140129511 Chr.4: 183317861-183317861 on Build GRCh38
rs371804641 Chr.4: 183317802-183317802 on Build GRCh38
rs972341278 Chr.4: 183317935-183317935 on Build GRCh38
rs773198390 Chr.4: 183318232-183318232 on Build GRCh38
rs922283821 Chr.4: 183318056-183318056 on Build GRCh38
rs1014373720 Chr.4: 183317912-183317912 on Build GRCh38
rs934713274 Chr.4: 183317910-183317910 on Build GRCh38
rs573012244 Chr.4: 183317938-183317938 on Build GRCh38
rs945740855 Chr.4: 183318067-183318067 on Build GRCh38
rs1028813352 Chr.4: 183318134-183318134 on Build GRCh38

View Full Product Details