Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183445433-183445976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN2AIP
Entrez Gene ID
55602
Gene Name
CDKN2A interacting protein
Gene Aliases
CARF
UniGene
Hs.592508
Gene Chromosome Location
Chr.4: 183444591-183449064 on Build GRCh38
Transcripts
NM_001317343.1 NM_017632.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
481091 c.295A>G p.K99E Chr.4: 183445557-183445557 on Build GRCh38
1725015 c.383_384insA p.G129fs*5 Chr.4: 183445645-183445646 on Build GRCh38
3730333 c.343T>G p.Y115D Chr.4: 183445605-183445605 on Build GRCh38
1053610 c.358G>A p.E120K Chr.4: 183445620-183445620 on Build GRCh38
1053609 c.274T>C p.Y92H Chr.4: 183445536-183445536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774669140 Chr.4: 183445485-183445485 on Build GRCh38
rs750140199 Chr.4: 183445720-183445724 on Build GRCh38
rs936397925 Chr.4: 183445711-183445711 on Build GRCh38
rs1042188656 Chr.4: 183445459-183445459 on Build GRCh38
rs748434981 Chr.4: 183445554-183445554 on Build GRCh38
rs1000956513 Chr.4: 183445483-183445483 on Build GRCh38
rs920992887 Chr.4: 183445719-183445719 on Build GRCh38
rs562786754 Chr.4: 183445591-183445591 on Build GRCh38
rs774924031 Chr.4: 183445574-183445574 on Build GRCh38
rs397713406 Chr.4: 183445481-183445482 on Build GRCh38
rs760306455 Chr.4: 183445510-183445510 on Build GRCh38
rs775189235 Chr.4: 183445509-183445509 on Build GRCh38
rs531711240 Chr.4: 183445592-183445592 on Build GRCh38
rs923687614 Chr.4: 183445702-183445702 on Build GRCh38
rs757706166 Chr.4: 183445696-183445696 on Build GRCh38
rs1052960308 Chr.4: 183445776-183445776 on Build GRCh38
rs764939439 Chr.4: 183445611-183445611 on Build GRCh38
rs1012159416 Chr.4: 183445524-183445524 on Build GRCh38
rs576993790 Chr.4: 183445516-183445516 on Build GRCh38
rs778181086 Chr.4: 183445513-183445513 on Build GRCh38
rs902230193 Chr.4: 183445473-183445473 on Build GRCh38
rs756211383 Chr.4: 183445682-183445682 on Build GRCh38
rs1043250993 Chr.4: 183445744-183445744 on Build GRCh38
rs542248144 Chr.4: 183445607-183445607 on Build GRCh38
rs1041882908 Chr.4: 183445701-183445701 on Build GRCh38
rs35858568 Chr.4: 183445477-183445478 on Build GRCh38
rs906436812 Chr.4: 183445804-183445804 on Build GRCh38
rs200057043 Chr.4: 183445743-183445743 on Build GRCh38
rs776091820 Chr.4: 183445609-183445609 on Build GRCh38
rs952854579 Chr.4: 183445896-183445897 on Build GRCh38
rs757110805 Chr.4: 183445716-183445716 on Build GRCh38
rs753566867 Chr.4: 183445608-183445610 on Build GRCh38
rs570034596 Chr.4: 183445914-183445914 on Build GRCh38
rs758251245 Chr.4: 183445649-183445649 on Build GRCh38
rs1002760501 Chr.4: 183445584-183445584 on Build GRCh38
rs200915094 Chr.4: 183445478-183445478 on Build GRCh38
rs755155806 Chr.4: 183445542-183445542 on Build GRCh38
rs760411117 Chr.4: 183445720-183445720 on Build GRCh38
rs1025468818 Chr.4: 183445539-183445539 on Build GRCh38
rs143816918 Chr.4: 183445803-183445803 on Build GRCh38
rs765236634 Chr.4: 183445526-183445526 on Build GRCh38
rs551218956 Chr.4: 183445603-183445603 on Build GRCh38
rs964035681 Chr.4: 183445618-183445618 on Build GRCh38
rs779381798 Chr.4: 183445562-183445562 on Build GRCh38
rs537887747 Chr.4: 183445756-183445756 on Build GRCh38
rs1004500482 Chr.4: 183445934-183445935 on Build GRCh38
rs1046495197 Chr.4: 183445808-183445808 on Build GRCh38
rs550024803 Chr.4: 183445893-183445893 on Build GRCh38
rs971111270 Chr.4: 183445812-183445812 on Build GRCh38
rs749682911 Chr.4: 183445567-183445567 on Build GRCh38
rs1027115775 Chr.4: 183445889-183445889 on Build GRCh38
rs925877860 Chr.4: 183445772-183445772 on Build GRCh38
rs780312991 Chr.4: 183445878-183445878 on Build GRCh38
rs571001762 Chr.4: 183445807-183445807 on Build GRCh38
rs778381400 Chr.4: 183445717-183445718 on Build GRCh38
rs141073811 Chr.4: 183445558-183445558 on Build GRCh38
rs200183773 Chr.4: 183445602-183445602 on Build GRCh38
rs771140793 Chr.4: 183445569-183445569 on Build GRCh38
rs11282266 Chr.4: 183445476-183445477 on Build GRCh38
rs1053945880 Chr.4: 183445733-183445733 on Build GRCh38
rs1052348592 Chr.4: 183445504-183445505 on Build GRCh38
rs369246349 Chr.4: 183445543-183445543 on Build GRCh38
rs146932991 Chr.4: 183445655-183445655 on Build GRCh38
rs369471031 Chr.4: 183445503-183445503 on Build GRCh38
rs779132351 Chr.4: 183445700-183445700 on Build GRCh38
rs527775428 Chr.4: 183445802-183445802 on Build GRCh38
rs201633995 Chr.4: 183445476-183445476 on Build GRCh38
rs905247516 Chr.4: 183445849-183445849 on Build GRCh38
rs752852591 Chr.4: 183445676-183445676 on Build GRCh38
rs200329502 Chr.4: 183445692-183445692 on Build GRCh38
rs761674330 Chr.4: 183445610-183445610 on Build GRCh38
rs376417441 Chr.4: 183445523-183445523 on Build GRCh38
rs750436686 Chr.4: 183445623-183445623 on Build GRCh38
rs73870523 Chr.4: 183445522-183445522 on Build GRCh38
rs533392580 Chr.4: 183445851-183445851 on Build GRCh38
rs766870406 Chr.4: 183445507-183445507 on Build GRCh38
rs749402413 Chr.4: 183445695-183445695 on Build GRCh38
rs138352332 Chr.4: 183445538-183445538 on Build GRCh38
rs904423312 Chr.4: 183445487-183445487 on Build GRCh38
rs894835997 Chr.4: 183445773-183445773 on Build GRCh38
rs1011856827 Chr.4: 183445784-183445784 on Build GRCh38
rs1025442577 Chr.4: 183445898-183445898 on Build GRCh38
rs770954239 Chr.4: 183445654-183445654 on Build GRCh38
rs764034598 Chr.4: 183445494-183445494 on Build GRCh38
rs908246633 Chr.4: 183445648-183445648 on Build GRCh38

View Full Product Details