Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184089805-184090301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP6
Entrez Gene ID
133121
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 6
Gene Aliases
NPP6
UniGene
Hs.297814
Gene Chromosome Location
Chr.4: 184088706-184217961 on Build GRCh38
Transcripts
NM_153343.3

SNP Detail

SNP ID Chromosome Location
rs1020613981 Chr.4: 184089837-184089837 on Build GRCh38
rs189893262 Chr.4: 184089854-184089854 on Build GRCh38
rs1046145019 Chr.4: 184090215-184090215 on Build GRCh38
rs189155055 Chr.4: 184089959-184089959 on Build GRCh38
rs911356498 Chr.4: 184090077-184090077 on Build GRCh38
rs951889176 Chr.4: 184089964-184089964 on Build GRCh38
rs573480708 Chr.4: 184090261-184090261 on Build GRCh38
rs578040410 Chr.4: 184089968-184089968 on Build GRCh38
rs961958214 Chr.4: 184089999-184089999 on Build GRCh38
rs147664047 Chr.4: 184090019-184090019 on Build GRCh38
rs150069681 Chr.4: 184089841-184089841 on Build GRCh38
rs534754226 Chr.4: 184089917-184089917 on Build GRCh38
rs936175241 Chr.4: 184090078-184090078 on Build GRCh38
rs938302978 Chr.4: 184089898-184089898 on Build GRCh38
rs764819326 Chr.4: 184090061-184090061 on Build GRCh38
rs182366819 Chr.4: 184089869-184089869 on Build GRCh38
rs530390386 Chr.4: 184089984-184089984 on Build GRCh38
rs114285165 Chr.4: 184089976-184089976 on Build GRCh38
rs1001991544 Chr.4: 184090264-184090264 on Build GRCh38
rs887696444 Chr.4: 184090048-184090048 on Build GRCh38
rs979117904 Chr.4: 184090219-184090219 on Build GRCh38
rs557463042 Chr.4: 184090018-184090018 on Build GRCh38
rs931299017 Chr.4: 184089987-184089987 on Build GRCh38
rs1020687604 Chr.4: 184090123-184090123 on Build GRCh38
rs946354538 Chr.4: 184090211-184090211 on Build GRCh38
rs755742310 Chr.4: 184089893-184089893 on Build GRCh38
rs557298663 Chr.4: 184090175-184090175 on Build GRCh38
rs901699723 Chr.4: 184090069-184090069 on Build GRCh38
rs966457020 Chr.4: 184090201-184090201 on Build GRCh38
rs918114819 Chr.4: 184089946-184089946 on Build GRCh38
rs540712028 Chr.4: 184090265-184090265 on Build GRCh38
rs1045760887 Chr.4: 184089926-184089926 on Build GRCh38
rs769489953 Chr.4: 184090194-184090195 on Build GRCh38
rs376711266 Chr.4: 184090197-184090197 on Build GRCh38
rs1050339902 Chr.4: 184089838-184089838 on Build GRCh38
rs990995793 Chr.4: 184089933-184089933 on Build GRCh38
rs891780382 Chr.4: 184089886-184089886 on Build GRCh38
rs1017423068 Chr.4: 184090063-184090063 on Build GRCh38
rs193038452 Chr.4: 184090022-184090022 on Build GRCh38
rs1007921889 Chr.4: 184090058-184090058 on Build GRCh38
rs112102971 Chr.4: 184090087-184090087 on Build GRCh38
rs142325674 Chr.4: 184090263-184090263 on Build GRCh38
rs758904470 Chr.4: 184089911-184089911 on Build GRCh38

View Full Product Details