Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184649257-184649799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP3
Entrez Gene ID
836
Gene Name
caspase 3
Gene Aliases
CPP32, CPP32B, SCA-1
UniGene
Hs.141125
Gene Chromosome Location
Chr.4: 184627696-184649475 on Build GRCh38
Transcripts
NM_004346.3 NM_032991.2 XM_011532301.1 NM_001300767.1 NM_001300768.1 NM_152683.3 XM_011531719.1 XM_011531720.1 XM_011531721.2 XM_011531723.1 XM_011531724.2 XM_011531725.1 XM_011531726.2 XM_011531729.1 XM_011531730.2 XM_017007864.1 XM_017007865.1 XM_017007866.1 XM_017007867.1 XM_017007868.1 XM_017007869.1 XM_017007870.1 XM_017007871.1 XM_017007872.1 XM_017007873.1 XM_017007874.1 XM_017007875.1

Gene Symbol
PRIMPOL
Entrez Gene ID
201973
Gene Name
primase and DNA directed polymerase
Gene Aliases
CCDC111, MYP22
UniGene
Hs.481307
Gene Chromosome Location
Chr.4: 184649613-184694959 on Build GRCh38
Transcripts
NM_004346.3 NM_032991.2 XM_011532301.1 NM_001300767.1 NM_001300768.1 NM_152683.3 XM_011531719.1 XM_011531720.1 XM_011531721.2 XM_011531723.1 XM_011531724.2 XM_011531725.1 XM_011531726.2 XM_011531729.1 XM_011531730.2 XM_017007864.1 XM_017007865.1 XM_017007866.1 XM_017007867.1 XM_017007868.1 XM_017007869.1 XM_017007870.1 XM_017007871.1 XM_017007872.1 XM_017007873.1 XM_017007874.1 XM_017007875.1

SNP Detail

SNP ID Chromosome Location
rs954470965 Chr.4: 184649703-184649703 on Build GRCh38
rs981077811 Chr.4: 184649387-184649387 on Build GRCh38
rs1051069868 Chr.4: 184649451-184649451 on Build GRCh38
rs918961702 Chr.4: 184649704-184649704 on Build GRCh38
rs970291622 Chr.4: 184649314-184649314 on Build GRCh38
rs957568730 Chr.4: 184649654-184649654 on Build GRCh38
rs1033214976 Chr.4: 184649337-184649337 on Build GRCh38
rs114746204 Chr.4: 184649521-184649521 on Build GRCh38
rs569952684 Chr.4: 184649671-184649671 on Build GRCh38
rs550204147 Chr.4: 184649662-184649662 on Build GRCh38
rs977392124 Chr.4: 184649417-184649417 on Build GRCh38
rs1028592254 Chr.4: 184649679-184649679 on Build GRCh38
rs999919521 Chr.4: 184649419-184649419 on Build GRCh38
rs35372903 Chr.4: 184649279-184649279 on Build GRCh38
rs918906108 Chr.4: 184649392-184649392 on Build GRCh38
rs916114580 Chr.4: 184649368-184649368 on Build GRCh38
rs535609691 Chr.4: 184649686-184649686 on Build GRCh38
rs990162580 Chr.4: 184649499-184649521 on Build GRCh38
rs4647600 Chr.4: 184649745-184649745 on Build GRCh38
rs932805237 Chr.4: 184649432-184649432 on Build GRCh38
rs901728603 Chr.4: 184649437-184649437 on Build GRCh38
rs965630899 Chr.4: 184649410-184649410 on Build GRCh38
rs991757967 Chr.4: 184649349-184649349 on Build GRCh38
rs949986009 Chr.4: 184649463-184649463 on Build GRCh38
rs562734902 Chr.4: 184649469-184649469 on Build GRCh38
rs1053138223 Chr.4: 184649450-184649450 on Build GRCh38
rs943159138 Chr.4: 184649415-184649415 on Build GRCh38
rs1044373921 Chr.4: 184649364-184649364 on Build GRCh38
rs529278405 Chr.4: 184649541-184649541 on Build GRCh38
rs886257172 Chr.4: 184649630-184649630 on Build GRCh38
rs925991219 Chr.4: 184649444-184649444 on Build GRCh38
rs895800445 Chr.4: 184649455-184649455 on Build GRCh38
rs531533981 Chr.4: 184649479-184649479 on Build GRCh38
rs1027781677 Chr.4: 184649353-184649353 on Build GRCh38
rs981675711 Chr.4: 184649672-184649672 on Build GRCh38
rs749761007 Chr.4: 184649341-184649341 on Build GRCh38
rs759064251 Chr.4: 184649612-184649624 on Build GRCh38
rs1027426557 Chr.4: 184649491-184649491 on Build GRCh38
rs951861966 Chr.4: 184649717-184649717 on Build GRCh38
rs988890245 Chr.4: 184649569-184649569 on Build GRCh38
rs950362523 Chr.4: 184649707-184649707 on Build GRCh38
rs554245353 Chr.4: 184649722-184649722 on Build GRCh38
rs903192832 Chr.4: 184649763-184649763 on Build GRCh38
rs1014166023 Chr.4: 184649309-184649309 on Build GRCh38
rs116609630 Chr.4: 184649542-184649542 on Build GRCh38
rs11722179 Chr.4: 184649325-184649325 on Build GRCh38
rs917288690 Chr.4: 184649585-184649585 on Build GRCh38
rs929071614 Chr.4: 184649396-184649396 on Build GRCh38
rs909088109 Chr.4: 184649411-184649411 on Build GRCh38
rs934754550 Chr.4: 184649565-184649565 on Build GRCh38
rs113420705 Chr.4: 184649399-184649399 on Build GRCh38
rs950057551 Chr.4: 184649598-184649598 on Build GRCh38
rs62339863 Chr.4: 184649313-184649313 on Build GRCh38
rs1004630060 Chr.4: 184649636-184649636 on Build GRCh38
rs11737863 Chr.4: 184649299-184649299 on Build GRCh38
rs965682813 Chr.4: 184649538-184649538 on Build GRCh38
rs943230517 Chr.4: 184649725-184649725 on Build GRCh38
rs905701598 Chr.4: 184649473-184649473 on Build GRCh38
rs1033471561 Chr.4: 184649616-184649616 on Build GRCh38
rs998161124 Chr.4: 184649447-184649447 on Build GRCh38
rs1004577737 Chr.4: 184649514-184649514 on Build GRCh38
rs779449324 Chr.4: 184649753-184649753 on Build GRCh38
rs996206313 Chr.4: 184649673-184649673 on Build GRCh38
rs1033217703 Chr.4: 184649705-184649705 on Build GRCh38
rs533571190 Chr.4: 184649754-184649754 on Build GRCh38
rs926095972 Chr.4: 184649562-184649562 on Build GRCh38
rs957768768 Chr.4: 184649342-184649342 on Build GRCh38
rs903182976 Chr.4: 184649456-184649456 on Build GRCh38
rs78471921 Chr.4: 184649734-184649734 on Build GRCh38
rs1044320685 Chr.4: 184649602-184649602 on Build GRCh38
rs971062912 Chr.4: 184649370-184649370 on Build GRCh38
rs184780686 Chr.4: 184649561-184649561 on Build GRCh38
rs1013471603 Chr.4: 184649655-184649655 on Build GRCh38
rs553766081 Chr.4: 184649755-184649755 on Build GRCh38
rs547734642 Chr.4: 184649549-184649549 on Build GRCh38
rs905749474 Chr.4: 184649605-184649605 on Build GRCh38
rs533708265 Chr.4: 184649576-184649576 on Build GRCh38
rs956183502 Chr.4: 184649711-184649711 on Build GRCh38
rs548741388 Chr.4: 184649484-184649484 on Build GRCh38
rs954240363 Chr.4: 184649546-184649546 on Build GRCh38
rs1028959453 Chr.4: 184649544-184649544 on Build GRCh38
rs374692744 Chr.4: 184649427-184649427 on Build GRCh38
rs974958580 Chr.4: 184649736-184649736 on Build GRCh38
rs1040398121 Chr.4: 184649478-184649478 on Build GRCh38

View Full Product Details