Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184672095-184672603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRIMPOL
Entrez Gene ID
201973
Gene Name
primase and DNA directed polymerase
Gene Aliases
CCDC111, MYP22
UniGene
Hs.481307
Gene Chromosome Location
Chr.4: 184649613-184694959 on Build GRCh38
Transcripts
NM_001300767.1 NM_001300768.1 NM_152683.3 XM_011531719.1 XM_011531720.1 XM_011531721.2 XM_011531723.1 XM_011531724.2 XM_011531725.1 XM_011531726.2 XM_011531729.1 XM_011531730.2 XM_017007864.1 XM_017007865.1 XM_017007866.1 XM_017007867.1 XM_017007868.1 XM_017007869.1 XM_017007870.1 XM_017007871.1 XM_017007872.1 XM_017007873.1 XM_017007874.1 XM_017007875.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1428856 c.560A>T p.N187I Chr.4: 184672176-184672176 on Build GRCh38
5040905 c.728G>C p.W243S Chr.4: 184672344-184672344 on Build GRCh38
6621311 c.844G>A p.G282R Chr.4: 184672460-184672460 on Build GRCh38
5179145 c.602G>A p.G201D Chr.4: 184672218-184672218 on Build GRCh38
3825595 c.607G>A p.E203K Chr.4: 184672223-184672223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762906128 Chr.4: 184672350-184672350 on Build GRCh38
rs780695010 Chr.4: 184672437-184672437 on Build GRCh38
rs775686581 Chr.4: 184672359-184672359 on Build GRCh38
rs746457375 Chr.4: 184672296-184672296 on Build GRCh38
rs768393267 Chr.4: 184672312-184672312 on Build GRCh38
rs1050915128 Chr.4: 184672282-184672282 on Build GRCh38
rs771771068 Chr.4: 184672361-184672361 on Build GRCh38
rs761843804 Chr.4: 184672138-184672138 on Build GRCh38
rs749057543 Chr.4: 184672459-184672459 on Build GRCh38
rs769224374 Chr.4: 184672237-184672237 on Build GRCh38
rs745573429 Chr.4: 184672269-184672270 on Build GRCh38
rs756220697 Chr.4: 184672151-184672151 on Build GRCh38
rs760643017 Chr.4: 184672364-184672364 on Build GRCh38
rs750777798 Chr.4: 184672408-184672408 on Build GRCh38
rs774154627 Chr.4: 184672464-184672464 on Build GRCh38
rs763403867 Chr.4: 184672495-184672495 on Build GRCh38
rs753996813 Chr.4: 184672376-184672376 on Build GRCh38
rs1036846832 Chr.4: 184672402-184672402 on Build GRCh38
rs760380380 Chr.4: 184672197-184672197 on Build GRCh38
rs376832310 Chr.4: 184672165-184672165 on Build GRCh38
rs145554992 Chr.4: 184672434-184672434 on Build GRCh38
rs751676662 Chr.4: 184672147-184672147 on Build GRCh38
rs763963677 Chr.4: 184672131-184672131 on Build GRCh38
rs921054222 Chr.4: 184672265-184672265 on Build GRCh38
rs770893069 Chr.4: 184672460-184672460 on Build GRCh38
rs72689278 Chr.4: 184672564-184672564 on Build GRCh38
rs771699834 Chr.4: 184672195-184672195 on Build GRCh38
rs367648499 Chr.4: 184672493-184672493 on Build GRCh38
rs763390444 Chr.4: 184672125-184672125 on Build GRCh38
rs779743283 Chr.4: 184672293-184672293 on Build GRCh38
rs34772069 Chr.4: 184672397-184672397 on Build GRCh38
rs767584476 Chr.4: 184672229-184672229 on Build GRCh38
rs370944751 Chr.4: 184672175-184672175 on Build GRCh38
rs62339868 Chr.4: 184672127-184672127 on Build GRCh38
rs746395188 Chr.4: 184672283-184672283 on Build GRCh38
rs755045895 Chr.4: 184672148-184672148 on Build GRCh38
rs893003038 Chr.4: 184672368-184672368 on Build GRCh38
rs761480385 Chr.4: 184672213-184672213 on Build GRCh38
rs933737058 Chr.4: 184672270-184672270 on Build GRCh38
rs748032943 Chr.4: 184672331-184672331 on Build GRCh38
rs765062426 Chr.4: 184672146-184672146 on Build GRCh38
rs372516005 Chr.4: 184672533-184672533 on Build GRCh38
rs577346176 Chr.4: 184672544-184672544 on Build GRCh38
rs910094289 Chr.4: 184672158-184672158 on Build GRCh38
rs780686146 Chr.4: 184672420-184672420 on Build GRCh38
rs377687682 Chr.4: 184672225-184672225 on Build GRCh38
rs769589734 Chr.4: 184672337-184672337 on Build GRCh38
rs999868327 Chr.4: 184672497-184672497 on Build GRCh38
rs371116864 Chr.4: 184672246-184672246 on Build GRCh38
rs958289534 Chr.4: 184672523-184672523 on Build GRCh38
rs776142895 Chr.4: 184672329-184672329 on Build GRCh38
rs141459769 Chr.4: 184672233-184672233 on Build GRCh38
rs1032645595 Chr.4: 184672516-184672516 on Build GRCh38
rs374221499 Chr.4: 184672431-184672431 on Build GRCh38
rs374268046 Chr.4: 184672207-184672207 on Build GRCh38
rs202080437 Chr.4: 184672210-184672211 on Build GRCh38
rs774021595 Chr.4: 184672176-184672177 on Build GRCh38
rs887285358 Chr.4: 184672517-184672517 on Build GRCh38
rs958237253 Chr.4: 184672145-184672145 on Build GRCh38
rs72689277 Chr.4: 184672545-184672545 on Build GRCh38
rs749564018 Chr.4: 184672159-184672159 on Build GRCh38
rs202179118 Chr.4: 184672416-184672416 on Build GRCh38
rs371359916 Chr.4: 184672126-184672126 on Build GRCh38
rs1028109368 Chr.4: 184672480-184672480 on Build GRCh38
rs114174581 Chr.4: 184672377-184672377 on Build GRCh38
rs768154834 Chr.4: 184672206-184672206 on Build GRCh38
rs774998238 Chr.4: 184672339-184672339 on Build GRCh38
rs764446905 Chr.4: 184672371-184672371 on Build GRCh38
rs750869342 Chr.4: 184672494-184672494 on Build GRCh38
rs377593791 Chr.4: 184672128-184672128 on Build GRCh38
rs762157728 Chr.4: 184672486-184672486 on Build GRCh38
rs776711191 Chr.4: 184672478-184672478 on Build GRCh38
rs779624305 Chr.4: 184672250-184672250 on Build GRCh38
rs771519815 Chr.4: 184672164-184672164 on Build GRCh38
rs777270041 Chr.4: 184672454-184672454 on Build GRCh38
rs553051581 Chr.4: 184672575-184672575 on Build GRCh38
rs764197991 Chr.4: 184672238-184672238 on Build GRCh38
rs778228425 Chr.4: 184672155-184672155 on Build GRCh38
rs975142479 Chr.4: 184672558-184672558 on Build GRCh38
rs191920610 Chr.4: 184672532-184672532 on Build GRCh38
rs1035077169 Chr.4: 184672510-184672510 on Build GRCh38
rs753511492 Chr.4: 184672133-184672133 on Build GRCh38
rs781451291 Chr.4: 184672149-184672149 on Build GRCh38
rs371793156 Chr.4: 184672529-184672529 on Build GRCh38
rs758909238 Chr.4: 184672291-184672291 on Build GRCh38
rs769392440 Chr.4: 184672451-184672451 on Build GRCh38
rs760869885 Chr.4: 184672468-184672468 on Build GRCh38
rs754272222 Chr.4: 184672240-184672240 on Build GRCh38
rs747759813 Chr.4: 184672450-184672450 on Build GRCh38
rs767049420 Chr.4: 184672507-184672507 on Build GRCh38
rs148306598 Chr.4: 184672222-184672222 on Build GRCh38
rs549819226 Chr.4: 184672351-184672351 on Build GRCh38
rs752019453 Chr.4: 184672508-184672508 on Build GRCh38
rs924429272 Chr.4: 184672332-184672332 on Build GRCh38
rs775082409 Chr.4: 184672196-184672196 on Build GRCh38

View Full Product Details