Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185396519-185397028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD37
Entrez Gene ID
353322
Gene Name
ankyrin repeat domain 37
Gene Aliases
Lrp2bp
UniGene
Hs.508154
Gene Chromosome Location
Chr.4: 185396686-185400245 on Build GRCh38
Transcripts
NM_181726.3 XM_017008176.1 XM_005263125.4 XM_006714265.3

Gene Symbol
LRP2BP
Entrez Gene ID
55805
Gene Name
LRP2 binding protein
Gene Aliases
-
UniGene
Hs.558513
Gene Chromosome Location
Chr.4: 185363878-185397160 on Build GRCh38
Transcripts
NM_181726.3 XM_017008176.1 XM_005263125.4 XM_006714265.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5018234 c.3G>C p.M1I Chr.4: 185396926-185396926 on Build GRCh38
159082 c.25G>A p.E9K Chr.4: 185396948-185396948 on Build GRCh38
6600720 c.21C>T p.N7N Chr.4: 185396944-185396944 on Build GRCh38
6420627 c.11T>G p.L4R Chr.4: 185396934-185396934 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770063120 Chr.4: 185396927-185396927 on Build GRCh38
rs538205808 Chr.4: 185396655-185396655 on Build GRCh38
rs1044379672 Chr.4: 185396973-185396973 on Build GRCh38
rs939942718 Chr.4: 185396727-185396727 on Build GRCh38
rs916167848 Chr.4: 185396644-185396644 on Build GRCh38
rs1027920119 Chr.4: 185396539-185396539 on Build GRCh38
rs368102461 Chr.4: 185396842-185396842 on Build GRCh38
rs901769290 Chr.4: 185396684-185396684 on Build GRCh38
rs991626648 Chr.4: 185396734-185396734 on Build GRCh38
rs757111355 Chr.4: 185396897-185396897 on Build GRCh38
rs1017414423 Chr.4: 185396681-185396681 on Build GRCh38
rs375548764 Chr.4: 185396903-185396903 on Build GRCh38
rs919495355 Chr.4: 185396782-185396782 on Build GRCh38
rs562203200 Chr.4: 185396972-185396972 on Build GRCh38
rs777933221 Chr.4: 185396694-185396694 on Build GRCh38
rs984801009 Chr.4: 185397002-185397002 on Build GRCh38
rs972074976 Chr.4: 185396748-185396748 on Build GRCh38
rs759890579 Chr.4: 185396978-185396979 on Build GRCh38
rs887858444 Chr.4: 185396672-185396672 on Build GRCh38
rs1021270592 Chr.4: 185396693-185396693 on Build GRCh38
rs1023205437 Chr.4: 185396560-185396560 on Build GRCh38
rs368015093 Chr.4: 185396947-185396947 on Build GRCh38
rs1003046025 Chr.4: 185396732-185396732 on Build GRCh38
rs903277808 Chr.4: 185396686-185396686 on Build GRCh38
rs759998381 Chr.4: 185396879-185396879 on Build GRCh38
rs746896112 Chr.4: 185396843-185396843 on Build GRCh38
rs745658221 Chr.4: 185396985-185396985 on Build GRCh38
rs1049050073 Chr.4: 185396820-185396820 on Build GRCh38
rs962729693 Chr.4: 185396857-185396857 on Build GRCh38
rs897802116 Chr.4: 185396666-185396666 on Build GRCh38
rs536545116 Chr.4: 185396965-185396966 on Build GRCh38
rs763683249 Chr.4: 185396892-185396892 on Build GRCh38
rs375186975 Chr.4: 185396858-185396858 on Build GRCh38
rs1049209264 Chr.4: 185396670-185396670 on Build GRCh38
rs779818036 Chr.4: 185396995-185396995 on Build GRCh38
rs1000933647 Chr.4: 185396707-185396707 on Build GRCh38
rs991616849 Chr.4: 185396643-185396643 on Build GRCh38
rs909302855 Chr.4: 185396826-185396826 on Build GRCh38
rs971607204 Chr.4: 185396731-185396731 on Build GRCh38
rs925023426 Chr.4: 185396632-185396632 on Build GRCh38
rs1036690892 Chr.4: 185396733-185396733 on Build GRCh38
rs778091654 Chr.4: 185397000-185397000 on Build GRCh38
rs976160577 Chr.4: 185396581-185396581 on Build GRCh38
rs556691658 Chr.4: 185396801-185396801 on Build GRCh38
rs377231072 Chr.4: 185396661-185396661 on Build GRCh38
rs753237761 Chr.4: 185396974-185396974 on Build GRCh38
rs957663123 Chr.4: 185396550-185396550 on Build GRCh38
rs959129180 Chr.4: 185396953-185396953 on Build GRCh38
rs371823357 Chr.4: 185396844-185396844 on Build GRCh38
rs540139187 Chr.4: 185396867-185396867 on Build GRCh38
rs971595941 Chr.4: 185396565-185396566 on Build GRCh38
rs530573280 Chr.4: 185396548-185396548 on Build GRCh38
rs966878150 Chr.4: 185396611-185396611 on Build GRCh38
rs758385831 Chr.4: 185396989-185396989 on Build GRCh38
rs997417634 Chr.4: 185396687-185396687 on Build GRCh38
rs763359900 Chr.4: 185396873-185396873 on Build GRCh38
rs959076996 Chr.4: 185396722-185396722 on Build GRCh38
rs1040048170 Chr.4: 185396676-185396676 on Build GRCh38
rs781068208 Chr.4: 185396917-185396917 on Build GRCh38
rs1023257451 Chr.4: 185396730-185396730 on Build GRCh38
rs927707479 Chr.4: 185396593-185396593 on Build GRCh38
rs901698461 Chr.4: 185396871-185396871 on Build GRCh38
rs532006092 Chr.4: 185396928-185396928 on Build GRCh38
rs1032093109 Chr.4: 185396714-185396714 on Build GRCh38
rs753436386 Chr.4: 185396895-185396895 on Build GRCh38
rs371815282 Chr.4: 185396919-185396919 on Build GRCh38
rs552289467 Chr.4: 185396957-185396957 on Build GRCh38
rs1011928520 Chr.4: 185396954-185396954 on Build GRCh38
rs771379699 Chr.4: 185396868-185396868 on Build GRCh38
rs527814388 Chr.4: 185396574-185396575 on Build GRCh38
rs200611006 Chr.4: 185396967-185396967 on Build GRCh38
rs998874872 Chr.4: 185396540-185396540 on Build GRCh38
rs981600487 Chr.4: 185396571-185396571 on Build GRCh38
rs929284967 Chr.4: 185396669-185396669 on Build GRCh38
rs916051614 Chr.4: 185396735-185396735 on Build GRCh38
rs768104857 Chr.4: 185396979-185396979 on Build GRCh38
rs111856363 Chr.4: 185396713-185396713 on Build GRCh38
rs202070767 Chr.4: 185396880-185396880 on Build GRCh38
rs934493413 Chr.4: 185396908-185396908 on Build GRCh38
rs771521687 Chr.4: 185396877-185396877 on Build GRCh38
rs752441156 Chr.4: 185396663-185396663 on Build GRCh38
rs577153984 Chr.4: 185396678-185396678 on Build GRCh38
rs776065978 Chr.4: 185396968-185396968 on Build GRCh38
rs368996821 Chr.4: 185396916-185396916 on Build GRCh38
rs979225880 Chr.4: 185396629-185396629 on Build GRCh38
rs372155536 Chr.4: 185396886-185396886 on Build GRCh38
rs902428618 Chr.4: 185396910-185396910 on Build GRCh38
rs1007391159 Chr.4: 185396554-185396554 on Build GRCh38
rs750137008 Chr.4: 185396905-185396905 on Build GRCh38
rs749430460 Chr.4: 185396942-185396942 on Build GRCh38
rs369509687 Chr.4: 185396997-185396997 on Build GRCh38
rs375151146 Chr.4: 185396979-185396979 on Build GRCh38
rs1017571976 Chr.4: 185396561-185396561 on Build GRCh38
rs1042344422 Chr.4: 185396883-185396883 on Build GRCh38
rs538560344 Chr.4: 185396806-185396806 on Build GRCh38
rs1050902374 Chr.4: 185396887-185396887 on Build GRCh38
rs936209871 Chr.4: 185396639-185396639 on Build GRCh38
rs539950386 Chr.4: 185396682-185396682 on Build GRCh38
rs948737818 Chr.4: 185396935-185396935 on Build GRCh38
rs929361102 Chr.4: 185396796-185396796 on Build GRCh38
rs575553162 Chr.4: 185396805-185396805 on Build GRCh38
rs999320876 Chr.4: 185396691-185396691 on Build GRCh38
rs764895901 Chr.4: 185396879-185396880 on Build GRCh38
rs1020587344 Chr.4: 185396589-185396589 on Build GRCh38
rs554102963 Chr.4: 185396665-185396665 on Build GRCh38
rs368944108 Chr.4: 185396836-185396836 on Build GRCh38
rs756302916 Chr.4: 185396998-185396998 on Build GRCh38
rs760468797 Chr.4: 185396889-185396889 on Build GRCh38
rs761416661 Chr.4: 185396977-185396977 on Build GRCh38
rs981741444 Chr.4: 185396753-185396753 on Build GRCh38
rs965910935 Chr.4: 185396576-185396576 on Build GRCh38
rs893150185 Chr.4: 185396711-185396711 on Build GRCh38

View Full Product Details